Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2141046-2141555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_001007139.5 NM_001127598.2 NM_001291862.2 NR_028043.2 NR_133657.1 NR_003512.3

Gene Symbol
IGF2-AS
Entrez Gene ID
51214
Gene Name
IGF2 antisense RNA
Gene Aliases
IGF2-AS1, IGF2AS, PEG8
UniGene
Hs.716962
Gene Chromosome Location
Chr.11: 2140512-2148666 on Build GRCh38
Transcripts
NM_001007139.5 NM_001127598.2 NM_001291862.2 NR_028043.2 NR_133657.1 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_001007139.5 NM_001127598.2 NM_001291862.2 NR_028043.2 NR_133657.1 NR_003512.3

SNP Detail

SNP ID Chromosome Location
rs966299910 Chr.11: 2141389-2141389 on Build GRCh38
rs764369247 Chr.11: 2141186-2141187 on Build GRCh38
rs182981173 Chr.11: 2141268-2141268 on Build GRCh38
rs923802782 Chr.11: 2141103-2141103 on Build GRCh38
rs900035996 Chr.11: 2141343-2141343 on Build GRCh38
rs892771270 Chr.11: 2141310-2141310 on Build GRCh38
rs1029737745 Chr.11: 2141370-2141370 on Build GRCh38
rs3213215 Chr.11: 2141196-2141196 on Build GRCh38
rs975953806 Chr.11: 2141411-2141411 on Build GRCh38
rs565529874 Chr.11: 2141260-2141260 on Build GRCh38
rs569877042 Chr.11: 2141321-2141321 on Build GRCh38
rs894049225 Chr.11: 2141072-2141072 on Build GRCh38
rs753016197 Chr.11: 2141245-2141245 on Build GRCh38
rs1049760336 Chr.11: 2141143-2141143 on Build GRCh38
rs932610328 Chr.11: 2141140-2141140 on Build GRCh38
rs947002927 Chr.11: 2141329-2141329 on Build GRCh38
rs963003523 Chr.11: 2141514-2141514 on Build GRCh38
rs186687164 Chr.11: 2141374-2141374 on Build GRCh38
rs998256070 Chr.11: 2141118-2141118 on Build GRCh38
rs35463443 Chr.11: 2141343-2141344 on Build GRCh38
rs547404153 Chr.11: 2141248-2141248 on Build GRCh38
rs187259775 Chr.11: 2141121-2141121 on Build GRCh38
rs765524123 Chr.11: 2141144-2141144 on Build GRCh38
rs1021922179 Chr.11: 2141291-2141291 on Build GRCh38
rs548458805 Chr.11: 2141280-2141280 on Build GRCh38
rs1035655060 Chr.11: 2141232-2141232 on Build GRCh38
rs577298891 Chr.11: 2141435-2141435 on Build GRCh38
rs915757627 Chr.11: 2141238-2141238 on Build GRCh38
rs1004663547 Chr.11: 2141386-2141386 on Build GRCh38
rs1018463515 Chr.11: 2141106-2141106 on Build GRCh38
rs1040348379 Chr.11: 2141332-2141332 on Build GRCh38
rs571287076 Chr.11: 2141170-2141170 on Build GRCh38
rs534789662 Chr.11: 2141495-2141495 on Build GRCh38
rs964152877 Chr.11: 2141114-2141114 on Build GRCh38
rs559118406 Chr.11: 2141376-2141376 on Build GRCh38
rs982797553 Chr.11: 2141158-2141158 on Build GRCh38
rs991790532 Chr.11: 2141203-2141203 on Build GRCh38
rs761361694 Chr.11: 2141451-2141451 on Build GRCh38
rs1011537033 Chr.11: 2141086-2141086 on Build GRCh38
rs998308472 Chr.11: 2141356-2141356 on Build GRCh38
rs563741963 Chr.11: 2141202-2141202 on Build GRCh38
rs931894342 Chr.11: 2141502-2141502 on Build GRCh38
rs951425867 Chr.11: 2141125-2141125 on Build GRCh38
rs768215216 Chr.11: 2141244-2141244 on Build GRCh38
rs924354615 Chr.11: 2141254-2141254 on Build GRCh38
rs1003787724 Chr.11: 2141209-2141209 on Build GRCh38
rs934348321 Chr.11: 2141255-2141255 on Build GRCh38

View Full Product Details