Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4586711-4587218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52I2
Entrez Gene ID
143502
Gene Name
olfactory receptor family 52 subfamily I member 2
Gene Aliases
OR11-12, OR52I1
UniGene
Hs.553607
Gene Chromosome Location
Chr.11: 4586791-4587905 on Build GRCh38
Transcripts
NM_001005170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1578437 c.401_402insTTTTGT p.V134_H135insFV Chr.11: 4587213-4587214 on Build GRCh38
688078 c.286C>T p.L96L Chr.11: 4587098-4587098 on Build GRCh38
1627982 c.368G>T p.S123I Chr.11: 4587180-4587180 on Build GRCh38
688080 c.183G>T p.L61L Chr.11: 4586995-4586995 on Build GRCh38
1180650 c.4T>A p.C2S Chr.11: 4586816-4586816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2115929 Chr.11: 4586872-4586872 on Build GRCh38
rs754731746 Chr.11: 4587150-4587150 on Build GRCh38
rs768040052 Chr.11: 4586919-4586919 on Build GRCh38
rs763163575 Chr.11: 4586974-4586974 on Build GRCh38
rs879160554 Chr.11: 4586931-4586931 on Build GRCh38
rs762289287 Chr.11: 4586771-4586771 on Build GRCh38
rs557924667 Chr.11: 4587044-4587044 on Build GRCh38
rs144837372 Chr.11: 4587137-4587137 on Build GRCh38
rs115128348 Chr.11: 4587168-4587168 on Build GRCh38
rs748812191 Chr.11: 4586916-4586916 on Build GRCh38
rs775005094 Chr.11: 4586857-4586857 on Build GRCh38
rs753574270 Chr.11: 4587085-4587085 on Build GRCh38
rs557100327 Chr.11: 4587023-4587023 on Build GRCh38
rs753605884 Chr.11: 4587149-4587149 on Build GRCh38
rs965316914 Chr.11: 4586912-4586912 on Build GRCh38
rs199862163 Chr.11: 4587015-4587015 on Build GRCh38
rs1017678692 Chr.11: 4587192-4587192 on Build GRCh38
rs542100049 Chr.11: 4586756-4586756 on Build GRCh38
rs776415569 Chr.11: 4586833-4586833 on Build GRCh38
rs769345391 Chr.11: 4586841-4586841 on Build GRCh38
rs142676846 Chr.11: 4586973-4586973 on Build GRCh38
rs200571706 Chr.11: 4586831-4586831 on Build GRCh38
rs763065318 Chr.11: 4587059-4587059 on Build GRCh38
rs757119323 Chr.11: 4586908-4586908 on Build GRCh38
rs561474051 Chr.11: 4587096-4587096 on Build GRCh38
rs752852896 Chr.11: 4586887-4586887 on Build GRCh38
rs781069460 Chr.11: 4587008-4587008 on Build GRCh38
rs773300843 Chr.11: 4587189-4587189 on Build GRCh38
rs7128702 Chr.11: 4586886-4586886 on Build GRCh38
rs771376286 Chr.11: 4586937-4586937 on Build GRCh38
rs56002758 Chr.11: 4587163-4587163 on Build GRCh38
rs369462656 Chr.11: 4586836-4586836 on Build GRCh38
rs748142759 Chr.11: 4586817-4586817 on Build GRCh38
rs141485776 Chr.11: 4587129-4587129 on Build GRCh38
rs572477890 Chr.11: 4586813-4586814 on Build GRCh38
rs759935380 Chr.11: 4586964-4586964 on Build GRCh38
rs138816802 Chr.11: 4586900-4586900 on Build GRCh38
rs746820900 Chr.11: 4586832-4586832 on Build GRCh38
rs777156929 Chr.11: 4586962-4586962 on Build GRCh38
rs1028183841 Chr.11: 4586822-4586822 on Build GRCh38
rs562772899 Chr.11: 4586801-4586801 on Build GRCh38
rs371482534 Chr.11: 4587037-4587037 on Build GRCh38
rs370188965 Chr.11: 4586930-4586930 on Build GRCh38
rs754609849 Chr.11: 4587086-4587086 on Build GRCh38
rs200581685 Chr.11: 4586862-4586862 on Build GRCh38
rs756367816 Chr.11: 4586794-4586794 on Build GRCh38
rs772187053 Chr.11: 4587188-4587188 on Build GRCh38
rs1028234831 Chr.11: 4587089-4587089 on Build GRCh38
rs12793957 Chr.11: 4587032-4587032 on Build GRCh38
rs770334488 Chr.11: 4587118-4587118 on Build GRCh38
rs368491751 Chr.11: 4587105-4587105 on Build GRCh38
rs575239969 Chr.11: 4586851-4586851 on Build GRCh38
rs766648684 Chr.11: 4586876-4586876 on Build GRCh38
rs375883595 Chr.11: 4586901-4586901 on Build GRCh38
rs377292270 Chr.11: 4586776-4586776 on Build GRCh38
rs780547834 Chr.11: 4587169-4587169 on Build GRCh38
rs141405424 Chr.11: 4586939-4586939 on Build GRCh38
rs773535424 Chr.11: 4586863-4586863 on Build GRCh38
rs368990302 Chr.11: 4587030-4587030 on Build GRCh38
rs2201338 Chr.11: 4587175-4587175 on Build GRCh38
rs753992648 Chr.11: 4586806-4586806 on Build GRCh38
rs766255269 Chr.11: 4587146-4587146 on Build GRCh38
rs779615355 Chr.11: 4586913-4586913 on Build GRCh38
rs968388159 Chr.11: 4586748-4586748 on Build GRCh38
rs568452202 Chr.11: 4586911-4586911 on Build GRCh38
rs775898224 Chr.11: 4587050-4587050 on Build GRCh38
rs192487695 Chr.11: 4586860-4586860 on Build GRCh38
rs140702655 Chr.11: 4587016-4587016 on Build GRCh38
rs766129992 Chr.11: 4587084-4587084 on Build GRCh38
rs373674464 Chr.11: 4587001-4587001 on Build GRCh38
rs953907960 Chr.11: 4587095-4587095 on Build GRCh38
rs199924123 Chr.11: 4587160-4587160 on Build GRCh38
rs373352913 Chr.11: 4586896-4586896 on Build GRCh38
rs746471070 Chr.11: 4587167-4587167 on Build GRCh38
rs893682633 Chr.11: 4586784-4586784 on Build GRCh38
rs149411057 Chr.11: 4587130-4587130 on Build GRCh38
rs763808672 Chr.11: 4586968-4586968 on Build GRCh38
rs761886686 Chr.11: 4587066-4587066 on Build GRCh38
rs749629745 Chr.11: 4587172-4587172 on Build GRCh38
rs112052046 Chr.11: 4586838-4586838 on Build GRCh38
rs764052362 Chr.11: 4586985-4586985 on Build GRCh38
rs369842491 Chr.11: 4586788-4586788 on Build GRCh38
rs780829699 Chr.11: 4586909-4586909 on Build GRCh38
rs754150906 Chr.11: 4586879-4586879 on Build GRCh38
rs757921773 Chr.11: 4587107-4587107 on Build GRCh38
rs373274567 Chr.11: 4586793-4586793 on Build GRCh38
rs764332350 Chr.11: 4587062-4587062 on Build GRCh38
rs762235219 Chr.11: 4586989-4586989 on Build GRCh38
rs761158944 Chr.11: 4586871-4586871 on Build GRCh38
rs751598870 Chr.11: 4586989-4586992 on Build GRCh38
rs550578100 Chr.11: 4586763-4586763 on Build GRCh38
rs757351110 Chr.11: 4587096-4587096 on Build GRCh38
rs145090174 Chr.11: 4587109-4587109 on Build GRCh38
rs770267003 Chr.11: 4587045-4587045 on Build GRCh38
rs140789551 Chr.11: 4587176-4587176 on Build GRCh38
rs140300674 Chr.11: 4587065-4587065 on Build GRCh38
rs760581992 Chr.11: 4587144-4587144 on Build GRCh38
rs764806515 Chr.11: 4587159-4587159 on Build GRCh38
rs748372319 Chr.11: 4587184-4587184 on Build GRCh38
rs200926516 Chr.11: 4587074-4587074 on Build GRCh38
rs746350049 Chr.11: 4587115-4587115 on Build GRCh38
rs749627578 Chr.11: 4587123-4587123 on Build GRCh38
rs61744242 Chr.11: 4587012-4587012 on Build GRCh38
rs764313125 Chr.11: 4586814-4586815 on Build GRCh38
rs562405324 Chr.11: 4586783-4586783 on Build GRCh38
rs138555035 Chr.11: 4587038-4587038 on Build GRCh38
rs773834429 Chr.11: 4586925-4586925 on Build GRCh38
rs779103720 Chr.11: 4586810-4586810 on Build GRCh38
rs755175718 Chr.11: 4586807-4586807 on Build GRCh38
rs374452023 Chr.11: 4587078-4587078 on Build GRCh38
rs776024852 Chr.11: 4587122-4587122 on Build GRCh38

View Full Product Details