Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 28110028-28110546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL15
Entrez Gene ID
196074
Gene Name
methyltransferase like 15
Gene Aliases
METT5D1
UniGene
Hs.243326
Gene Chromosome Location
Chr.11: 28108251-28527041 on Build GRCh38
Transcripts
NM_001113528.1 NM_001297775.1 NM_152636.2 XM_011519935.2 XM_011519940.2 XM_011519941.2 XM_017017297.1 XM_017017298.1 XM_017017300.1 XM_017017301.1 XM_017017303.1

SNP Detail

SNP ID Chromosome Location
rs966839000 Chr.11: 28110084-28110084 on Build GRCh38
rs1046017628 Chr.11: 28110306-28110306 on Build GRCh38
rs960616767 Chr.11: 28110099-28110099 on Build GRCh38
rs978297869 Chr.11: 28110090-28110090 on Build GRCh38
rs956656232 Chr.11: 28110221-28110221 on Build GRCh38
rs958483124 Chr.11: 28110355-28110355 on Build GRCh38
rs987429088 Chr.11: 28110192-28110192 on Build GRCh38
rs144306638 Chr.11: 28110505-28110505 on Build GRCh38
rs554518500 Chr.11: 28110364-28110364 on Build GRCh38
rs1029250395 Chr.11: 28110366-28110366 on Build GRCh38
rs567338139 Chr.11: 28110225-28110225 on Build GRCh38
rs964308056 Chr.11: 28110490-28110490 on Build GRCh38
rs979961670 Chr.11: 28110184-28110184 on Build GRCh38
rs949065303 Chr.11: 28110345-28110353 on Build GRCh38
rs1012607670 Chr.11: 28110399-28110399 on Build GRCh38
rs183224984 Chr.11: 28110134-28110134 on Build GRCh38
rs191165694 Chr.11: 28110229-28110229 on Build GRCh38
rs931158720 Chr.11: 28110276-28110276 on Build GRCh38
rs187850202 Chr.11: 28110191-28110191 on Build GRCh38
rs907894061 Chr.11: 28110415-28110415 on Build GRCh38
rs922173114 Chr.11: 28110159-28110159 on Build GRCh38
rs1004467810 Chr.11: 28110352-28110352 on Build GRCh38
rs1032952529 Chr.11: 28110115-28110115 on Build GRCh38
rs566290881 Chr.11: 28110165-28110165 on Build GRCh38
rs759813979 Chr.11: 28110088-28110088 on Build GRCh38
rs1031050260 Chr.11: 28110425-28110425 on Build GRCh38
rs76221023 Chr.11: 28110169-28110169 on Build GRCh38
rs886145952 Chr.11: 28110064-28110064 on Build GRCh38
rs572646985 Chr.11: 28110464-28110464 on Build GRCh38
rs370388614 Chr.11: 28110368-28110368 on Build GRCh38
rs968317959 Chr.11: 28110421-28110421 on Build GRCh38
rs1054998144 Chr.11: 28110282-28110282 on Build GRCh38
rs962052557 Chr.11: 28110438-28110438 on Build GRCh38
rs187944940 Chr.11: 28110496-28110496 on Build GRCh38
rs954916910 Chr.11: 28110376-28110376 on Build GRCh38
rs975033165 Chr.11: 28110146-28110146 on Build GRCh38
rs923927821 Chr.11: 28110215-28110215 on Build GRCh38
rs919778026 Chr.11: 28110248-28110248 on Build GRCh38
rs896380162 Chr.11: 28110386-28110386 on Build GRCh38
rs749905759 Chr.11: 28110411-28110411 on Build GRCh38
rs1013223668 Chr.11: 28110294-28110294 on Build GRCh38
rs1032091205 Chr.11: 28110349-28110349 on Build GRCh38
rs764400641 Chr.11: 28110457-28110457 on Build GRCh38
rs1023904438 Chr.11: 28110151-28110151 on Build GRCh38
rs766401947 Chr.11: 28110404-28110406 on Build GRCh38
rs571156397 Chr.11: 28110290-28110290 on Build GRCh38
rs1000711443 Chr.11: 28110423-28110423 on Build GRCh38
rs920113353 Chr.11: 28110312-28110312 on Build GRCh38
rs764636088 Chr.11: 28110353-28110353 on Build GRCh38
rs1034868940 Chr.11: 28110093-28110093 on Build GRCh38
rs755582865 Chr.11: 28110450-28110450 on Build GRCh38
rs986770453 Chr.11: 28110451-28110451 on Build GRCh38
rs35686304 Chr.11: 28110143-28110143 on Build GRCh38
rs61888968 Chr.11: 28110319-28110319 on Build GRCh38
rs182178041 Chr.11: 28110246-28110246 on Build GRCh38
rs139572677 Chr.11: 28110336-28110336 on Build GRCh38
rs112785689 Chr.11: 28110434-28110434 on Build GRCh38

View Full Product Details