Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44077617-44078122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
UniGene
Hs.126706
Gene Chromosome Location
Chr.11: 44066118-44084237 on Build GRCh38
Transcripts
NM_001127219.1 NM_032592.3 XM_005253170.3 XM_005253172.2 XM_006718348.2 XM_011520406.2 XM_011520407.1 XM_011520410.2 XM_017018421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32704 c.662G>A p.G221E Chr.11: 44077852-44077852 on Build GRCh38
4032807 c.709G>A p.A237T Chr.11: 44077899-44077899 on Build GRCh38
5027381 c.718G>A p.E240K Chr.11: 44077908-44077908 on Build GRCh38
7095629 c.728C>G p.S243C Chr.11: 44077918-44077918 on Build GRCh38
69505 c.661G>C p.G221R Chr.11: 44077851-44077851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754359923 Chr.11: 44077865-44077865 on Build GRCh38
rs772042129 Chr.11: 44077965-44077965 on Build GRCh38
rs749754237 Chr.11: 44077805-44077805 on Build GRCh38
rs749889420 Chr.11: 44077880-44077880 on Build GRCh38
rs1048799885 Chr.11: 44077689-44077689 on Build GRCh38
rs1056669609 Chr.11: 44077727-44077727 on Build GRCh38
rs768328446 Chr.11: 44077829-44077829 on Build GRCh38
rs563395676 Chr.11: 44077877-44077877 on Build GRCh38
rs548871599 Chr.11: 44077974-44077974 on Build GRCh38
rs771675736 Chr.11: 44077821-44077821 on Build GRCh38
rs556636504 Chr.11: 44077659-44077660 on Build GRCh38
rs955785109 Chr.11: 44077836-44077836 on Build GRCh38
rs756821862 Chr.11: 44077811-44077811 on Build GRCh38
rs201006757 Chr.11: 44077800-44077800 on Build GRCh38
rs770788917 Chr.11: 44077947-44077947 on Build GRCh38
rs3134907 Chr.11: 44078038-44078038 on Build GRCh38
rs930064013 Chr.11: 44077891-44077891 on Build GRCh38
rs748126449 Chr.11: 44077934-44077934 on Build GRCh38
rs763300645 Chr.11: 44077767-44077767 on Build GRCh38
rs35514614 Chr.11: 44077852-44077852 on Build GRCh38
rs376215031 Chr.11: 44077664-44077664 on Build GRCh38
rs745527857 Chr.11: 44077819-44077819 on Build GRCh38
rs778339991 Chr.11: 44077815-44077815 on Build GRCh38
rs895008157 Chr.11: 44077744-44077744 on Build GRCh38
rs763846243 Chr.11: 44077991-44077991 on Build GRCh38
rs983164540 Chr.11: 44077898-44077898 on Build GRCh38
rs746857839 Chr.11: 44077826-44077826 on Build GRCh38
rs775277283 Chr.11: 44077820-44077820 on Build GRCh38
rs1012731461 Chr.11: 44078008-44078008 on Build GRCh38
rs1011983733 Chr.11: 44077761-44077761 on Build GRCh38
rs766570260 Chr.11: 44077838-44077838 on Build GRCh38
rs567573520 Chr.11: 44077992-44077992 on Build GRCh38
rs774981438 Chr.11: 44077824-44077824 on Build GRCh38
rs573006076 Chr.11: 44077769-44077769 on Build GRCh38
rs151237226 Chr.11: 44078055-44078055 on Build GRCh38
rs774532219 Chr.11: 44077844-44077844 on Build GRCh38
rs939250464 Chr.11: 44077963-44077963 on Build GRCh38
rs145349955 Chr.11: 44077863-44077863 on Build GRCh38
rs753864353 Chr.11: 44078027-44078027 on Build GRCh38
rs76818320 Chr.11: 44077640-44077640 on Build GRCh38
rs780782552 Chr.11: 44077933-44077933 on Build GRCh38
rs201529252 Chr.11: 44077889-44077889 on Build GRCh38
rs759731314 Chr.11: 44077951-44077951 on Build GRCh38
rs376247485 Chr.11: 44077914-44077914 on Build GRCh38
rs746623811 Chr.11: 44077910-44077910 on Build GRCh38
rs759857734 Chr.11: 44077848-44077848 on Build GRCh38
rs574221319 Chr.11: 44077741-44077741 on Build GRCh38
rs753058356 Chr.11: 44077798-44077798 on Build GRCh38
rs369723942 Chr.11: 44077847-44077847 on Build GRCh38
rs764353314 Chr.11: 44077864-44077864 on Build GRCh38
rs769803998 Chr.11: 44077939-44077939 on Build GRCh38
rs757813899 Chr.11: 44077884-44077884 on Build GRCh38
rs756413120 Chr.11: 44077799-44077799 on Build GRCh38
rs191252184 Chr.11: 44078062-44078062 on Build GRCh38
rs188070248 Chr.11: 44077702-44077702 on Build GRCh38
rs1047065474 Chr.11: 44077654-44077654 on Build GRCh38
rs200253447 Chr.11: 44077943-44077943 on Build GRCh38
rs761785309 Chr.11: 44077834-44077834 on Build GRCh38
rs757698561 Chr.11: 44077872-44077872 on Build GRCh38
rs543608955 Chr.11: 44077661-44077661 on Build GRCh38
rs1022089768 Chr.11: 44077778-44077778 on Build GRCh38
rs372496715 Chr.11: 44077831-44077831 on Build GRCh38
rs377437583 Chr.11: 44077783-44077783 on Build GRCh38
rs773150498 Chr.11: 44077748-44077748 on Build GRCh38
rs767802451 Chr.11: 44077795-44077795 on Build GRCh38
rs939299891 Chr.11: 44077711-44077711 on Build GRCh38

View Full Product Details