Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47161785-47162279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf49
Entrez Gene ID
79096
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
UniGene
Hs.368296
Gene Chromosome Location
Chr.11: 46936524-47164381 on Build GRCh38
Transcripts
NM_001003676.2 NM_001003677.2 NM_001003678.2 NM_001278222.1 NM_024113.4 NR_103471.1 NR_103472.1 XM_006718315.1 XM_011520364.2 XM_011520365.2 XM_017018277.1 XM_017018278.1 XM_017018279.1 XM_017018280.1 XM_017018281.1 XM_017018282.1

SNP Detail

SNP ID Chromosome Location
rs891970796 Chr.11: 47161810-47161810 on Build GRCh38
rs75741689 Chr.11: 47162140-47162140 on Build GRCh38
rs957037678 Chr.11: 47162231-47162231 on Build GRCh38
rs759585367 Chr.11: 47162081-47162081 on Build GRCh38
rs14717 Chr.11: 47162022-47162022 on Build GRCh38
rs148649527 Chr.11: 47161967-47161967 on Build GRCh38
rs979824479 Chr.11: 47161978-47161978 on Build GRCh38
rs14718 Chr.11: 47162106-47162106 on Build GRCh38
rs895532194 Chr.11: 47161824-47161824 on Build GRCh38
rs926947709 Chr.11: 47162010-47162010 on Build GRCh38
rs879621554 Chr.11: 47162009-47162010 on Build GRCh38
rs897973767 Chr.11: 47161841-47161841 on Build GRCh38
rs989194485 Chr.11: 47161929-47161929 on Build GRCh38
rs952197150 Chr.11: 47161891-47161891 on Build GRCh38
rs559462134 Chr.11: 47161910-47161910 on Build GRCh38
rs78112569 Chr.11: 47162139-47162139 on Build GRCh38
rs897602742 Chr.11: 47162082-47162082 on Build GRCh38
rs1040924324 Chr.11: 47162063-47162063 on Build GRCh38
rs996824521 Chr.11: 47162065-47162065 on Build GRCh38
rs747647388 Chr.11: 47161906-47161906 on Build GRCh38
rs535336596 Chr.11: 47162047-47162047 on Build GRCh38
rs1014976673 Chr.11: 47162090-47162090 on Build GRCh38
rs549597650 Chr.11: 47162031-47162031 on Build GRCh38
rs192397029 Chr.11: 47161907-47161907 on Build GRCh38
rs972513064 Chr.11: 47161923-47161923 on Build GRCh38
rs774658358 Chr.11: 47161934-47161935 on Build GRCh38
rs1054212227 Chr.11: 47162019-47162019 on Build GRCh38
rs1050490202 Chr.11: 47162067-47162067 on Build GRCh38
rs995440665 Chr.11: 47161875-47161875 on Build GRCh38
rs563963949 Chr.11: 47161867-47161867 on Build GRCh38
rs761924615 Chr.11: 47162221-47162221 on Build GRCh38
rs777225897 Chr.11: 47161970-47161970 on Build GRCh38
rs961203802 Chr.11: 47161919-47161919 on Build GRCh38
rs902068814 Chr.11: 47161834-47161834 on Build GRCh38
rs1021031980 Chr.11: 47162207-47162207 on Build GRCh38
rs544016237 Chr.11: 47162214-47162214 on Build GRCh38
rs995011292 Chr.11: 47162125-47162125 on Build GRCh38
rs183173046 Chr.11: 47162252-47162252 on Build GRCh38
rs966089456 Chr.11: 47161941-47161941 on Build GRCh38
rs539236886 Chr.11: 47162027-47162027 on Build GRCh38
rs989790684 Chr.11: 47162242-47162242 on Build GRCh38
rs1054667121 Chr.11: 47161808-47161808 on Build GRCh38
rs962080903 Chr.11: 47162135-47162135 on Build GRCh38
rs117749721 Chr.11: 47161876-47161876 on Build GRCh38
rs1015479640 Chr.11: 47161839-47161839 on Build GRCh38
rs971375989 Chr.11: 47162185-47162185 on Build GRCh38
rs79545080 Chr.11: 47161825-47161825 on Build GRCh38
rs1006145371 Chr.11: 47162075-47162075 on Build GRCh38
rs77535491 Chr.11: 47162012-47162012 on Build GRCh38
rs912955940 Chr.11: 47162021-47162021 on Build GRCh38
rs745388579 Chr.11: 47162068-47162068 on Build GRCh38
rs1018932438 Chr.11: 47162147-47162147 on Build GRCh38
rs749141719 Chr.11: 47161997-47161997 on Build GRCh38
rs1055428566 Chr.11: 47161814-47161814 on Build GRCh38
rs569590319 Chr.11: 47162032-47162032 on Build GRCh38
rs557565641 Chr.11: 47162170-47162170 on Build GRCh38
rs1040661184 Chr.11: 47161826-47161826 on Build GRCh38
rs935612386 Chr.11: 47162014-47162014 on Build GRCh38
rs967225128 Chr.11: 47162011-47162011 on Build GRCh38
rs1009600828 Chr.11: 47162199-47162199 on Build GRCh38
rs902120911 Chr.11: 47162064-47162064 on Build GRCh38
rs776375687 Chr.11: 47161966-47161966 on Build GRCh38
rs534614366 Chr.11: 47162151-47162151 on Build GRCh38
rs531632494 Chr.11: 47162195-47162195 on Build GRCh38

View Full Product Details