Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47260291-47260806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1H3
Entrez Gene ID
10062
Gene Name
nuclear receptor subfamily 1 group H member 3
Gene Aliases
LXR-a, LXRA, RLD-1
UniGene
Hs.438863
Gene Chromosome Location
Chr.11: 47248300-47269033 on Build GRCh38
Transcripts
NM_001130101.2 NM_001130102.2 NM_001251934.1 NM_001251935.1 NM_005693.3 XM_005252705.1 XM_005252706.1 XM_005252707.4 XM_005252709.1 XM_005252710.1 XM_005252713.3 XM_005252715.2 XM_005252716.3 XM_005252718.3 XM_006718112.1 XM_006718113.1 XM_006718115.1 XM_006718116.1 XM_011519805.2 XM_011519806.1 XM_011519807.1 XM_011519808.2 XM_017017056.1 XM_017017057.1 XM_017017058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1981342 c.439C>T p.R147C Chr.11: 47260615-47260615 on Build GRCh38
4745665 c.476G>A p.R159H Chr.11: 47260652-47260652 on Build GRCh38
927376 c.490C>T p.R164W Chr.11: 47260666-47260666 on Build GRCh38
6551971 c.363C>T p.F121F Chr.11: 47260539-47260539 on Build GRCh38
22025 c.390G>T p.A130A Chr.11: 47260566-47260566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560145458 Chr.11: 47260384-47260384 on Build GRCh38
rs768053453 Chr.11: 47260663-47260663 on Build GRCh38
rs34806350 Chr.11: 47260566-47260566 on Build GRCh38
rs749306694 Chr.11: 47260510-47260510 on Build GRCh38
rs781012781 Chr.11: 47260493-47260493 on Build GRCh38
rs946699019 Chr.11: 47260619-47260619 on Build GRCh38
rs892307963 Chr.11: 47260761-47260761 on Build GRCh38
rs761438448 Chr.11: 47260391-47260391 on Build GRCh38
rs1038626979 Chr.11: 47260671-47260671 on Build GRCh38
rs773330714 Chr.11: 47260398-47260398 on Build GRCh38
rs753126526 Chr.11: 47260666-47260666 on Build GRCh38
rs751781969 Chr.11: 47260413-47260413 on Build GRCh38
rs780959887 Chr.11: 47260569-47260569 on Build GRCh38
rs148847768 Chr.11: 47260589-47260589 on Build GRCh38
rs762962632 Chr.11: 47260637-47260637 on Build GRCh38
rs753876556 Chr.11: 47260448-47260448 on Build GRCh38
rs776816368 Chr.11: 47260617-47260617 on Build GRCh38
rs757527591 Chr.11: 47260717-47260718 on Build GRCh38
rs754635334 Chr.11: 47260491-47260491 on Build GRCh38
rs1043752731 Chr.11: 47260643-47260643 on Build GRCh38
rs891251541 Chr.11: 47260732-47260732 on Build GRCh38
rs745644666 Chr.11: 47260607-47260607 on Build GRCh38
rs761767428 Chr.11: 47260618-47260618 on Build GRCh38
rs753342549 Chr.11: 47260694-47260694 on Build GRCh38
rs146426911 Chr.11: 47260414-47260414 on Build GRCh38
rs200218097 Chr.11: 47260565-47260565 on Build GRCh38
rs35378256 Chr.11: 47260368-47260368 on Build GRCh38
rs139930107 Chr.11: 47260371-47260371 on Build GRCh38
rs1030802497 Chr.11: 47260759-47260759 on Build GRCh38
rs770865360 Chr.11: 47260599-47260599 on Build GRCh38
rs145675661 Chr.11: 47260746-47260746 on Build GRCh38
rs747876791 Chr.11: 47260578-47260578 on Build GRCh38
rs756143353 Chr.11: 47260365-47260365 on Build GRCh38
rs906576811 Chr.11: 47260338-47260338 on Build GRCh38
rs1011211613 Chr.11: 47260771-47260771 on Build GRCh38
rs779563962 Chr.11: 47260378-47260378 on Build GRCh38
rs879613993 Chr.11: 47260764-47260764 on Build GRCh38
rs777323885 Chr.11: 47260693-47260693 on Build GRCh38
rs763028207 Chr.11: 47260729-47260729 on Build GRCh38
rs746731016 Chr.11: 47260379-47260379 on Build GRCh38
rs755441726 Chr.11: 47260568-47260568 on Build GRCh38
rs899806576 Chr.11: 47260674-47260674 on Build GRCh38
rs374430731 Chr.11: 47260521-47260521 on Build GRCh38
rs757354481 Chr.11: 47260450-47260450 on Build GRCh38
rs770789264 Chr.11: 47260517-47260517 on Build GRCh38
rs748753598 Chr.11: 47260591-47260591 on Build GRCh38
rs932690369 Chr.11: 47260701-47260701 on Build GRCh38
rs774079609 Chr.11: 47260544-47260544 on Build GRCh38
rs775386021 Chr.11: 47260507-47260507 on Build GRCh38
rs377628830 Chr.11: 47260459-47260459 on Build GRCh38
rs145779080 Chr.11: 47260728-47260728 on Build GRCh38
rs939465465 Chr.11: 47260345-47260345 on Build GRCh38
rs371886672 Chr.11: 47260375-47260375 on Build GRCh38
rs762626495 Chr.11: 47260401-47260401 on Build GRCh38
rs764255803 Chr.11: 47260545-47260545 on Build GRCh38
rs747336913 Chr.11: 47260480-47260480 on Build GRCh38
rs756770158 Chr.11: 47260703-47260703 on Build GRCh38
rs766886186 Chr.11: 47260646-47260646 on Build GRCh38
rs199680544 Chr.11: 47260524-47260524 on Build GRCh38
rs577011989 Chr.11: 47260461-47260461 on Build GRCh38
rs369828876 Chr.11: 47260385-47260385 on Build GRCh38
rs778191731 Chr.11: 47260360-47260362 on Build GRCh38
rs747639895 Chr.11: 47260494-47260494 on Build GRCh38
rs112784092 Chr.11: 47260402-47260402 on Build GRCh38
rs759974111 Chr.11: 47260652-47260652 on Build GRCh38
rs371384896 Chr.11: 47260508-47260508 on Build GRCh38
rs374064171 Chr.11: 47260397-47260397 on Build GRCh38
rs755762647 Chr.11: 47260688-47260688 on Build GRCh38
rs34974176 Chr.11: 47260509-47260509 on Build GRCh38
rs773299515 Chr.11: 47260635-47260635 on Build GRCh38
rs764307084 Chr.11: 47260443-47260443 on Build GRCh38
rs760894618 Chr.11: 47260427-47260427 on Build GRCh38
rs776890461 Chr.11: 47260547-47260547 on Build GRCh38
rs765714521 Chr.11: 47260457-47260457 on Build GRCh38
rs752046081 Chr.11: 47260651-47260651 on Build GRCh38
rs771898659 Chr.11: 47260612-47260612 on Build GRCh38
rs370060636 Chr.11: 47260600-47260600 on Build GRCh38
rs2279238 Chr.11: 47260473-47260473 on Build GRCh38
rs745840776 Chr.11: 47260415-47260415 on Build GRCh38
rs765448154 Chr.11: 47260623-47260623 on Build GRCh38
rs567560964 Chr.11: 47260374-47260374 on Build GRCh38
rs201229981 Chr.11: 47260561-47260561 on Build GRCh38
rs765354464 Chr.11: 47260558-47260558 on Build GRCh38
rs761873817 Chr.11: 47260552-47260552 on Build GRCh38
rs145536921 Chr.11: 47260587-47260587 on Build GRCh38

View Full Product Details