Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56093139-56093658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8I2
Entrez Gene ID
120586
Gene Name
olfactory receptor family 8 subfamily I member 2
Gene Aliases
OR11-170
UniGene
Hs.554524
Gene Chromosome Location
Chr.11: 56093308-56094240 on Build GRCh38
Transcripts
NM_001003750.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033774 c.323T>C p.V108A Chr.11: 56093630-56093630 on Build GRCh38
4710293 c.271T>C p.S91P Chr.11: 56093578-56093578 on Build GRCh38
4033772 c.32T>C p.V11A Chr.11: 56093339-56093339 on Build GRCh38
4791871 c.194A>T p.N65I Chr.11: 56093501-56093501 on Build GRCh38
6485603 c.23A>G p.E8G Chr.11: 56093330-56093330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761184596 Chr.11: 56093454-56093454 on Build GRCh38
rs778478595 Chr.11: 56093367-56093367 on Build GRCh38
rs768568510 Chr.11: 56093423-56093423 on Build GRCh38
rs770610043 Chr.11: 56093540-56093540 on Build GRCh38
rs201519232 Chr.11: 56093331-56093331 on Build GRCh38
rs757449135 Chr.11: 56093397-56093397 on Build GRCh38
rs568277388 Chr.11: 56093308-56093308 on Build GRCh38
rs779093803 Chr.11: 56093494-56093494 on Build GRCh38
rs750471721 Chr.11: 56093500-56093500 on Build GRCh38
rs760926386 Chr.11: 56093544-56093544 on Build GRCh38
rs762424867 Chr.11: 56093531-56093531 on Build GRCh38
rs535039269 Chr.11: 56093439-56093439 on Build GRCh38
rs745618560 Chr.11: 56093315-56093315 on Build GRCh38
rs764883357 Chr.11: 56093264-56093264 on Build GRCh38
rs985928503 Chr.11: 56093363-56093363 on Build GRCh38
rs750505231 Chr.11: 56093555-56093555 on Build GRCh38
rs746816606 Chr.11: 56093512-56093512 on Build GRCh38
rs1013826771 Chr.11: 56093557-56093557 on Build GRCh38
rs766593939 Chr.11: 56093565-56093565 on Build GRCh38
rs775169529 Chr.11: 56093327-56093327 on Build GRCh38
rs761448342 Chr.11: 56093574-56093574 on Build GRCh38
rs867452723 Chr.11: 56093429-56093429 on Build GRCh38
rs748797357 Chr.11: 56093581-56093581 on Build GRCh38
rs764865092 Chr.11: 56093340-56093340 on Build GRCh38
rs751781705 Chr.11: 56093567-56093567 on Build GRCh38
rs780200138 Chr.11: 56093508-56093508 on Build GRCh38
rs138846423 Chr.11: 56093572-56093572 on Build GRCh38
rs758590179 Chr.11: 56093502-56093502 on Build GRCh38
rs748166736 Chr.11: 56093235-56093235 on Build GRCh38
rs759907028 Chr.11: 56093385-56093389 on Build GRCh38
rs144263438 Chr.11: 56093409-56093409 on Build GRCh38
rs756838054 Chr.11: 56093262-56093262 on Build GRCh38
rs762341594 Chr.11: 56093550-56093550 on Build GRCh38
rs562847672 Chr.11: 56093206-56093206 on Build GRCh38
rs749388797 Chr.11: 56093305-56093305 on Build GRCh38
rs539009203 Chr.11: 56093357-56093357 on Build GRCh38
rs143920392 Chr.11: 56093176-56093176 on Build GRCh38
rs765764668 Chr.11: 56093487-56093488 on Build GRCh38
rs746053712 Chr.11: 56093407-56093407 on Build GRCh38
rs754445015 Chr.11: 56093478-56093478 on Build GRCh38
rs183167361 Chr.11: 56093573-56093573 on Build GRCh38
rs771665296 Chr.11: 56093258-56093258 on Build GRCh38
rs767878120 Chr.11: 56093351-56093351 on Build GRCh38
rs141191744 Chr.11: 56093554-56093554 on Build GRCh38
rs764661137 Chr.11: 56093461-56093461 on Build GRCh38
rs200412583 Chr.11: 56093406-56093406 on Build GRCh38
rs774397067 Chr.11: 56093230-56093230 on Build GRCh38
rs753379937 Chr.11: 56093261-56093261 on Build GRCh38
rs533510191 Chr.11: 56093215-56093215 on Build GRCh38
rs748033528 Chr.11: 56093519-56093519 on Build GRCh38
rs762327242 Chr.11: 56093621-56093621 on Build GRCh38
rs377712344 Chr.11: 56093470-56093470 on Build GRCh38
rs755279483 Chr.11: 56093347-56093347 on Build GRCh38
rs774775935 Chr.11: 56093313-56093313 on Build GRCh38
rs773911459 Chr.11: 56093543-56093543 on Build GRCh38
rs867307617 Chr.11: 56093420-56093420 on Build GRCh38
rs574279295 Chr.11: 56093518-56093518 on Build GRCh38
rs770392973 Chr.11: 56093582-56093582 on Build GRCh38
rs201473727 Chr.11: 56093541-56093541 on Build GRCh38
rs568504554 Chr.11: 56093338-56093338 on Build GRCh38
rs773173351 Chr.11: 56093352-56093352 on Build GRCh38
rs755441893 Chr.11: 56093274-56093274 on Build GRCh38
rs750120848 Chr.11: 56093344-56093344 on Build GRCh38
rs773934404 Chr.11: 56093585-56093585 on Build GRCh38
rs776062641 Chr.11: 56093337-56093337 on Build GRCh38
rs916746372 Chr.11: 56093244-56093244 on Build GRCh38
rs771950754 Chr.11: 56093318-56093318 on Build GRCh38
rs199834660 Chr.11: 56093527-56093527 on Build GRCh38
rs200387875 Chr.11: 56093447-56093447 on Build GRCh38
rs1049313097 Chr.11: 56093223-56093223 on Build GRCh38
rs138233109 Chr.11: 56093415-56093415 on Build GRCh38
rs560401423 Chr.11: 56093598-56093598 on Build GRCh38
rs202108392 Chr.11: 56093360-56093360 on Build GRCh38
rs534144921 Chr.11: 56093444-56093444 on Build GRCh38
rs373532000 Chr.11: 56093269-56093269 on Build GRCh38
rs756603204 Chr.11: 56093299-56093299 on Build GRCh38
rs757860785 Chr.11: 56093480-56093480 on Build GRCh38
rs768480288 Chr.11: 56093515-56093515 on Build GRCh38
rs578075631 Chr.11: 56093578-56093578 on Build GRCh38
rs776616051 Chr.11: 56093426-56093426 on Build GRCh38
rs1040032893 Chr.11: 56093173-56093173 on Build GRCh38
rs545372561 Chr.11: 56093594-56093594 on Build GRCh38
rs111785740 Chr.11: 56093539-56093539 on Build GRCh38
rs1842700 Chr.11: 56093304-56093304 on Build GRCh38
rs1040360836 Chr.11: 56093349-56093349 on Build GRCh38
rs545507497 Chr.11: 56093234-56093234 on Build GRCh38
rs866650900 Chr.11: 56093516-56093516 on Build GRCh38
rs994352839 Chr.11: 56093498-56093498 on Build GRCh38
rs1048732153 Chr.11: 56093475-56093475 on Build GRCh38
rs560591857 Chr.11: 56093242-56093242 on Build GRCh38
rs928935468 Chr.11: 56093440-56093440 on Build GRCh38
rs377349344 Chr.11: 56093547-56093547 on Build GRCh38
rs866256608 Chr.11: 56093436-56093436 on Build GRCh38
rs145057566 Chr.11: 56093556-56093556 on Build GRCh38
rs550211029 Chr.11: 56093335-56093335 on Build GRCh38
rs566533595 Chr.11: 56093432-56093432 on Build GRCh38
rs756029254 Chr.11: 56093577-56093577 on Build GRCh38
rs946847027 Chr.11: 56093245-56093245 on Build GRCh38
rs754285611 Chr.11: 56093548-56093548 on Build GRCh38
rs753090740 Chr.11: 56093453-56093453 on Build GRCh38
rs993242254 Chr.11: 56093202-56093202 on Build GRCh38
rs901874021 Chr.11: 56093385-56093385 on Build GRCh38
rs765958780 Chr.11: 56093448-56093448 on Build GRCh38
rs748740477 Chr.11: 56093296-56093296 on Build GRCh38
rs563969347 Chr.11: 56093291-56093291 on Build GRCh38
rs771853397 Chr.11: 56093609-56093609 on Build GRCh38
rs777077171 Chr.11: 56093610-56093610 on Build GRCh38
rs374664682 Chr.11: 56093376-56093376 on Build GRCh38
rs370749171 Chr.11: 56093628-56093628 on Build GRCh38
rs148743160 Chr.11: 56093530-56093530 on Build GRCh38
rs954103662 Chr.11: 56093619-56093619 on Build GRCh38
rs746795179 Chr.11: 56093422-56093422 on Build GRCh38
rs113726715 Chr.11: 56093277-56093277 on Build GRCh38
rs1033809086 Chr.11: 56093248-56093248 on Build GRCh38

View Full Product Details