Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63291457-63291955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A10
Entrez Gene ID
387775
Gene Name
solute carrier family 22 member 10
Gene Aliases
OAT5, hOAT5
UniGene
Hs.188982
Gene Chromosome Location
Chr.11: 63268257-63311983 on Build GRCh38
Transcripts
NM_001039752.3 NR_134874.1 XM_011545016.2 XM_017017699.1 XM_017017700.1 XM_017017701.1 XM_017017702.1 XM_017017703.1 XM_017017704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5525574 c.445C>T p.Q149* Chr.11: 63291582-63291582 on Build GRCh38
1127701 c.445C>G p.Q149E Chr.11: 63291582-63291582 on Build GRCh38
467175 c.430C>A p.L144M Chr.11: 63291567-63291567 on Build GRCh38
3869801 c.406G>A p.D136N Chr.11: 63291543-63291543 on Build GRCh38
3739199 c.439G>C p.V147L Chr.11: 63291576-63291576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777563215 Chr.11: 63291744-63291744 on Build GRCh38
rs770035485 Chr.11: 63291644-63291644 on Build GRCh38
rs533209121 Chr.11: 63291611-63291611 on Build GRCh38
rs753164140 Chr.11: 63291490-63291490 on Build GRCh38
rs754621564 Chr.11: 63291613-63291613 on Build GRCh38
rs1025917506 Chr.11: 63291697-63291697 on Build GRCh38
rs967713445 Chr.11: 63291485-63291485 on Build GRCh38
rs780888319 Chr.11: 63291514-63291514 on Build GRCh38
rs181411561 Chr.11: 63291756-63291756 on Build GRCh38
rs537247627 Chr.11: 63291858-63291858 on Build GRCh38
rs776731612 Chr.11: 63291642-63291642 on Build GRCh38
rs998771867 Chr.11: 63291612-63291612 on Build GRCh38
rs1001201015 Chr.11: 63291486-63291486 on Build GRCh38
rs754640506 Chr.11: 63291681-63291681 on Build GRCh38
rs777688777 Chr.11: 63291629-63291629 on Build GRCh38
rs1012764979 Chr.11: 63291881-63291881 on Build GRCh38
rs994532236 Chr.11: 63291915-63291915 on Build GRCh38
rs777728607 Chr.11: 63291589-63291589 on Build GRCh38
rs915349409 Chr.11: 63291739-63291739 on Build GRCh38
rs752221477 Chr.11: 63291695-63291695 on Build GRCh38
rs764487325 Chr.11: 63291658-63291658 on Build GRCh38
rs746115266 Chr.11: 63291643-63291643 on Build GRCh38
rs773333998 Chr.11: 63291545-63291545 on Build GRCh38
rs762363840 Chr.11: 63291671-63291671 on Build GRCh38
rs1042108606 Chr.11: 63291762-63291762 on Build GRCh38
rs903522171 Chr.11: 63291832-63291832 on Build GRCh38
rs950410025 Chr.11: 63291931-63291931 on Build GRCh38
rs539065414 Chr.11: 63291884-63291884 on Build GRCh38
rs779849366 Chr.11: 63291519-63291519 on Build GRCh38
rs770927881 Chr.11: 63291547-63291547 on Build GRCh38
rs1022729253 Chr.11: 63291895-63291895 on Build GRCh38
rs546545462 Chr.11: 63291716-63291716 on Build GRCh38
rs975882247 Chr.11: 63291587-63291587 on Build GRCh38
rs973060583 Chr.11: 63291689-63291689 on Build GRCh38
rs185713384 Chr.11: 63291757-63291757 on Build GRCh38
rs546492938 Chr.11: 63291563-63291563 on Build GRCh38
rs778723128 Chr.11: 63291528-63291528 on Build GRCh38
rs1017438593 Chr.11: 63291501-63291501 on Build GRCh38
rs374973806 Chr.11: 63291512-63291512 on Build GRCh38
rs1042443269 Chr.11: 63291752-63291752 on Build GRCh38
rs189445162 Chr.11: 63291483-63291483 on Build GRCh38
rs752635326 Chr.11: 63291708-63291708 on Build GRCh38
rs746803380 Chr.11: 63291670-63291670 on Build GRCh38
rs1025968411 Chr.11: 63291928-63291928 on Build GRCh38
rs779655194 Chr.11: 63291608-63291608 on Build GRCh38
rs576833911 Chr.11: 63291876-63291876 on Build GRCh38
rs755690939 Chr.11: 63291599-63291599 on Build GRCh38
rs763406066 Chr.11: 63291652-63291652 on Build GRCh38
rs898942251 Chr.11: 63291913-63291913 on Build GRCh38
rs776728920 Chr.11: 63291559-63291559 on Build GRCh38
rs1042390516 Chr.11: 63291606-63291606 on Build GRCh38
rs943551067 Chr.11: 63291726-63291726 on Build GRCh38
rs895689951 Chr.11: 63291837-63291837 on Build GRCh38
rs759028657 Chr.11: 63291526-63291526 on Build GRCh38
rs371560094 Chr.11: 63291661-63291661 on Build GRCh38
rs753360774 Chr.11: 63291679-63291679 on Build GRCh38
rs200029760 Chr.11: 63291539-63291539 on Build GRCh38
rs375444926 Chr.11: 63291624-63291624 on Build GRCh38
rs943481369 Chr.11: 63291484-63291484 on Build GRCh38
rs748070754 Chr.11: 63291535-63291535 on Build GRCh38
rs562547733 Chr.11: 63291729-63291729 on Build GRCh38
rs771968167 Chr.11: 63291559-63291562 on Build GRCh38
rs564599400 Chr.11: 63291591-63291591 on Build GRCh38
rs926860119 Chr.11: 63291693-63291693 on Build GRCh38
rs771018026 Chr.11: 63291641-63291641 on Build GRCh38
rs768351001 Chr.11: 63291527-63291527 on Build GRCh38
rs367972898 Chr.11: 63291852-63291852 on Build GRCh38
rs775775394 Chr.11: 63291645-63291645 on Build GRCh38
rs912084017 Chr.11: 63291722-63291722 on Build GRCh38
rs963068436 Chr.11: 63291680-63291680 on Build GRCh38
rs765840805 Chr.11: 63291674-63291674 on Build GRCh38
rs758392231 Chr.11: 63291625-63291625 on Build GRCh38
rs753074350 Chr.11: 63291580-63291580 on Build GRCh38
rs764698167 Chr.11: 63291584-63291584 on Build GRCh38
rs764933906 Chr.11: 63291694-63291694 on Build GRCh38
rs770702112 Chr.11: 63291689-63291690 on Build GRCh38
rs1057050827 Chr.11: 63291830-63291830 on Build GRCh38
rs369803443 Chr.11: 63291630-63291630 on Build GRCh38
rs1056630795 Chr.11: 63291737-63291737 on Build GRCh38
rs755889445 Chr.11: 63291517-63291517 on Build GRCh38
rs751401653 Chr.11: 63291801-63291801 on Build GRCh38
rs987730658 Chr.11: 63291717-63291717 on Build GRCh38
rs372861021 Chr.11: 63291616-63291616 on Build GRCh38
rs139776727 Chr.11: 63291868-63291868 on Build GRCh38
rs749931152 Chr.11: 63291515-63291515 on Build GRCh38
rs567943536 Chr.11: 63291795-63291795 on Build GRCh38
rs765550162 Chr.11: 63291575-63291575 on Build GRCh38

View Full Product Details