Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63302246-63302743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A10
Entrez Gene ID
387775
Gene Name
solute carrier family 22 member 10
Gene Aliases
OAT5, hOAT5
UniGene
Hs.188982
Gene Chromosome Location
Chr.11: 63268257-63311983 on Build GRCh38
Transcripts
NM_001039752.3 NR_134874.1 XM_011545016.2 XM_017017699.1 XM_017017700.1 XM_017017701.1 XM_017017702.1 XM_017017703.1 XM_017017704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585962 c.1071-1G>C p.? Chr.11: 63302328-63302328 on Build GRCh38
6585963 c.1104G>C p.M368I Chr.11: 63302362-63302362 on Build GRCh38
5503751 c.1264G>A p.A422T Chr.11: 63302522-63302522 on Build GRCh38
213243 c.1144C>G p.Q382E Chr.11: 63302402-63302402 on Build GRCh38
5519048 c.1213C>T p.R405C Chr.11: 63302471-63302471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998508445 Chr.11: 63302270-63302270 on Build GRCh38
rs754132420 Chr.11: 63302402-63302402 on Build GRCh38
rs752691780 Chr.11: 63302495-63302495 on Build GRCh38
rs778666185 Chr.11: 63302556-63302556 on Build GRCh38
rs147574056 Chr.11: 63302599-63302599 on Build GRCh38
rs987472951 Chr.11: 63302436-63302436 on Build GRCh38
rs768420092 Chr.11: 63302455-63302455 on Build GRCh38
rs963497323 Chr.11: 63302417-63302417 on Build GRCh38
rs765549452 Chr.11: 63302494-63302494 on Build GRCh38
rs766698401 Chr.11: 63302345-63302345 on Build GRCh38
rs528553487 Chr.11: 63302273-63302273 on Build GRCh38
rs1044631533 Chr.11: 63302642-63302642 on Build GRCh38
rs778271110 Chr.11: 63302467-63302467 on Build GRCh38
rs374377774 Chr.11: 63302389-63302389 on Build GRCh38
rs372246351 Chr.11: 63302693-63302693 on Build GRCh38
rs867469910 Chr.11: 63302523-63302523 on Build GRCh38
rs56260618 Chr.11: 63302434-63302434 on Build GRCh38
rs1038272778 Chr.11: 63302399-63302399 on Build GRCh38
rs964230298 Chr.11: 63302274-63302274 on Build GRCh38
rs777585249 Chr.11: 63302514-63302514 on Build GRCh38
rs773117715 Chr.11: 63302373-63302373 on Build GRCh38
rs537005860 Chr.11: 63302692-63302692 on Build GRCh38
rs777792699 Chr.11: 63302577-63302577 on Build GRCh38
rs370062357 Chr.11: 63302472-63302472 on Build GRCh38
rs994928251 Chr.11: 63302418-63302418 on Build GRCh38
rs758240714 Chr.11: 63302527-63302527 on Build GRCh38
rs551924935 Chr.11: 63302583-63302583 on Build GRCh38
rs982935754 Chr.11: 63302360-63302360 on Build GRCh38
rs761650141 Chr.11: 63302366-63302366 on Build GRCh38
rs766664148 Chr.11: 63302394-63302394 on Build GRCh38
rs377229708 Chr.11: 63302435-63302435 on Build GRCh38
rs778102652 Chr.11: 63302303-63302303 on Build GRCh38
rs768066793 Chr.11: 63302358-63302358 on Build GRCh38
rs577393298 Chr.11: 63302322-63302322 on Build GRCh38
rs748749134 Chr.11: 63302346-63302346 on Build GRCh38
rs779625761 Chr.11: 63302445-63302445 on Build GRCh38
rs200791944 Chr.11: 63302323-63302323 on Build GRCh38
rs756419947 Chr.11: 63302538-63302538 on Build GRCh38
rs199596564 Chr.11: 63302422-63302422 on Build GRCh38
rs754564922 Chr.11: 63302547-63302547 on Build GRCh38
rs564171817 Chr.11: 63302395-63302395 on Build GRCh38
rs369357843 Chr.11: 63302536-63302536 on Build GRCh38
rs747426166 Chr.11: 63302316-63302316 on Build GRCh38
rs754126496 Chr.11: 63302377-63302378 on Build GRCh38
rs1014381542 Chr.11: 63302439-63302439 on Build GRCh38
rs759666150 Chr.11: 63302489-63302489 on Build GRCh38
rs770701717 Chr.11: 63302548-63302549 on Build GRCh38
rs200687543 Chr.11: 63302546-63302546 on Build GRCh38
rs779012357 Chr.11: 63302427-63302427 on Build GRCh38
rs554365412 Chr.11: 63302654-63302654 on Build GRCh38
rs368792154 Chr.11: 63302345-63302345 on Build GRCh38
rs973720978 Chr.11: 63302660-63302660 on Build GRCh38
rs755178608 Chr.11: 63302422-63302423 on Build GRCh38
rs375615317 Chr.11: 63302529-63302529 on Build GRCh38
rs763886129 Chr.11: 63302639-63302639 on Build GRCh38
rs149029722 Chr.11: 63302684-63302684 on Build GRCh38
rs779424180 Chr.11: 63302286-63302286 on Build GRCh38
rs755042227 Chr.11: 63302284-63302284 on Build GRCh38
rs933758922 Chr.11: 63302351-63302351 on Build GRCh38
rs752807143 Chr.11: 63302453-63302453 on Build GRCh38
rs145117732 Chr.11: 63302423-63302423 on Build GRCh38
rs747742215 Chr.11: 63302470-63302470 on Build GRCh38
rs755024978 Chr.11: 63302405-63302405 on Build GRCh38
rs758457005 Chr.11: 63302478-63302478 on Build GRCh38
rs899347489 Chr.11: 63302656-63302656 on Build GRCh38
rs746944982 Chr.11: 63302579-63302579 on Build GRCh38
rs558279779 Chr.11: 63302683-63302683 on Build GRCh38
rs770859964 Chr.11: 63302474-63302474 on Build GRCh38
rs781531138 Chr.11: 63302530-63302530 on Build GRCh38
rs534745391 Chr.11: 63302615-63302615 on Build GRCh38
rs111953413 Chr.11: 63302475-63302475 on Build GRCh38
rs747567947 Chr.11: 63302564-63302564 on Build GRCh38
rs370921588 Chr.11: 63302300-63302300 on Build GRCh38
rs896055944 Chr.11: 63302289-63302289 on Build GRCh38
rs199585843 Chr.11: 63302412-63302412 on Build GRCh38
rs771870426 Chr.11: 63302471-63302471 on Build GRCh38
rs746900840 Chr.11: 63302545-63302546 on Build GRCh38
rs750628200 Chr.11: 63302534-63302534 on Build GRCh38
rs367719841 Chr.11: 63302433-63302433 on Build GRCh38
rs760583126 Chr.11: 63302377-63302377 on Build GRCh38
rs896093388 Chr.11: 63302614-63302614 on Build GRCh38
rs774276642 Chr.11: 63302361-63302361 on Build GRCh38
rs367929551 Chr.11: 63302390-63302390 on Build GRCh38
rs763125020 Chr.11: 63302526-63302526 on Build GRCh38
rs748898778 Chr.11: 63302451-63302451 on Build GRCh38
rs767548539 Chr.11: 63302371-63302371 on Build GRCh38

View Full Product Details