Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119101785-119102295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPAGT1
Entrez Gene ID
1798
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Gene Aliases
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
UniGene
Hs.524081
Gene Chromosome Location
Chr.11: 119096503-119102414 on Build GRCh38
Transcripts
NM_001382.3 XM_005271422.2 XM_011542648.1 XM_017017293.1 XM_017017294.1 XM_017017295.1

SNP Detail

SNP ID Chromosome Location
rs747470304 Chr.11: 119101942-119101942 on Build GRCh38
rs917380151 Chr.11: 119101957-119101957 on Build GRCh38
rs954086404 Chr.11: 119102213-119102213 on Build GRCh38
rs750633011 Chr.11: 119102271-119102271 on Build GRCh38
rs949853296 Chr.11: 119101981-119101981 on Build GRCh38
rs527857760 Chr.11: 119102083-119102085 on Build GRCh38
rs746259017 Chr.11: 119101809-119101809 on Build GRCh38
rs979019953 Chr.11: 119101938-119101938 on Build GRCh38
rs903353023 Chr.11: 119102218-119102218 on Build GRCh38
rs777081154 Chr.11: 119102235-119102235 on Build GRCh38
rs370219953 Chr.11: 119101854-119101854 on Build GRCh38
rs540454811 Chr.11: 119101807-119101807 on Build GRCh38
rs769347070 Chr.11: 119102234-119102234 on Build GRCh38
rs772260853 Chr.11: 119101811-119101811 on Build GRCh38
rs886047760 Chr.11: 119101920-119101920 on Build GRCh38
rs79071301 Chr.11: 119101955-119101955 on Build GRCh38
rs759377136 Chr.11: 119102256-119102256 on Build GRCh38
rs1044526199 Chr.11: 119101984-119101984 on Build GRCh38
rs527882164 Chr.11: 119102102-119102102 on Build GRCh38
rs372189618 Chr.11: 119102227-119102227 on Build GRCh38
rs766043849 Chr.11: 119102059-119102059 on Build GRCh38
rs934879553 Chr.11: 119101946-119101946 on Build GRCh38
rs141116509 Chr.11: 119102004-119102004 on Build GRCh38
rs773815465 Chr.11: 119102128-119102128 on Build GRCh38
rs914926915 Chr.11: 119102179-119102179 on Build GRCh38
rs894472819 Chr.11: 119101924-119101924 on Build GRCh38
rs573965223 Chr.11: 119101939-119101939 on Build GRCh38
rs748267269 Chr.11: 119101977-119101977 on Build GRCh38
rs552734184 Chr.11: 119102185-119102185 on Build GRCh38
rs1018799344 Chr.11: 119102171-119102171 on Build GRCh38
rs994502116 Chr.11: 119102016-119102016 on Build GRCh38
rs776172400 Chr.11: 119101907-119101907 on Build GRCh38
rs565232740 Chr.11: 119101950-119101950 on Build GRCh38
rs530006160 Chr.11: 119101990-119101990 on Build GRCh38
rs950570003 Chr.11: 119101839-119101839 on Build GRCh38
rs886366088 Chr.11: 119102111-119102111 on Build GRCh38
rs901656686 Chr.11: 119102183-119102183 on Build GRCh38
rs1041840928 Chr.11: 119101872-119101872 on Build GRCh38
rs548289286 Chr.11: 119101998-119101998 on Build GRCh38
rs773222359 Chr.11: 119101967-119101967 on Build GRCh38
rs1053539826 Chr.11: 119101835-119101835 on Build GRCh38
rs577067503 Chr.11: 119101888-119101888 on Build GRCh38
rs765463233 Chr.11: 119102263-119102263 on Build GRCh38
rs932590228 Chr.11: 119102170-119102170 on Build GRCh38
rs954175826 Chr.11: 119101920-119101920 on Build GRCh38
rs1015416115 Chr.11: 119102028-119102028 on Build GRCh38
rs767173143 Chr.11: 119102258-119102258 on Build GRCh38
rs965506125 Chr.11: 119101865-119101865 on Build GRCh38
rs921221632 Chr.11: 119102168-119102168 on Build GRCh38
rs921302222 Chr.11: 119101890-119101890 on Build GRCh38
rs1018758333 Chr.11: 119101856-119101856 on Build GRCh38
rs562876563 Chr.11: 119101960-119101960 on Build GRCh38
rs951875984 Chr.11: 119101864-119101864 on Build GRCh38
rs982880569 Chr.11: 119102025-119102026 on Build GRCh38
rs932346172 Chr.11: 119101822-119101822 on Build GRCh38
rs1003061278 Chr.11: 119101911-119101911 on Build GRCh38
rs150086804 Chr.11: 119102199-119102199 on Build GRCh38
rs1032945499 Chr.11: 119102232-119102232 on Build GRCh38
rs1048720664 Chr.11: 119102057-119102057 on Build GRCh38
rs191838170 Chr.11: 119101874-119101874 on Build GRCh38
rs1004776855 Chr.11: 119102113-119102113 on Build GRCh38

View Full Product Details