Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19739745-19740160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO4
Entrez Gene ID
255104
Gene Name
transmembrane and coiled-coil domains 4
Gene Aliases
-
UniGene
Hs.656313
Gene Chromosome Location
Chr.1: 19682213-19800385 on Build GRCh38
Transcripts
NM_181719.4 XM_005245822.4 XM_005245824.4 XM_005245825.4 XM_005245827.3 XM_006710551.3 XM_011541179.2 XM_011541180.2 XM_011541181.1 XM_011541185.2 XM_011541186.2 XM_017000913.1 XM_017000914.1 XM_017000915.1 XM_017000916.1 XM_017000917.1 XM_017000918.1 XM_017000919.1 XM_017000920.1 XM_017000921.1 XM_017000922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7394042 c.1091A>G p.N364S Chr.1: 19739912-19739912 on Build GRCh38
6209551 c.1179+8G>C p.? Chr.1: 19739816-19739816 on Build GRCh38
6802986 c.1127G>A p.R376Q Chr.1: 19739876-19739876 on Build GRCh38
4798537 c.1173G>A p.R391R Chr.1: 19739830-19739830 on Build GRCh38
6209567 c.1179+9G>T p.? Chr.1: 19739815-19739815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150663799 Chr.1: 19739832-19739832 on Build GRCh38
rs1035491299 Chr.1: 19739862-19739862 on Build GRCh38
rs770007334 Chr.1: 19739794-19739794 on Build GRCh38
rs775455291 Chr.1: 19739990-19739990 on Build GRCh38
rs199714913 Chr.1: 19739828-19739828 on Build GRCh38
rs145282048 Chr.1: 19739958-19739958 on Build GRCh38
rs6426824 Chr.1: 19739992-19739992 on Build GRCh38
rs752590581 Chr.1: 19739926-19739926 on Build GRCh38
rs774756780 Chr.1: 19739809-19739809 on Build GRCh38
rs959206775 Chr.1: 19740084-19740084 on Build GRCh38
rs200221198 Chr.1: 19739782-19739782 on Build GRCh38
rs547648652 Chr.1: 19740103-19740103 on Build GRCh38
rs545681408 Chr.1: 19739881-19739882 on Build GRCh38
rs769487627 Chr.1: 19739971-19739971 on Build GRCh38
rs765529375 Chr.1: 19739998-19739998 on Build GRCh38
rs762885224 Chr.1: 19739845-19739845 on Build GRCh38
rs776197717 Chr.1: 19739831-19739831 on Build GRCh38
rs765006414 Chr.1: 19739836-19739836 on Build GRCh38
rs767511747 Chr.1: 19739920-19739920 on Build GRCh38
rs1021111777 Chr.1: 19739998-19739998 on Build GRCh38
rs748387505 Chr.1: 19739789-19739789 on Build GRCh38
rs552239386 Chr.1: 19739955-19739955 on Build GRCh38
rs372565126 Chr.1: 19739839-19739839 on Build GRCh38
rs41303893 Chr.1: 19739887-19739887 on Build GRCh38
rs759192879 Chr.1: 19739916-19739916 on Build GRCh38
rs767185206 Chr.1: 19739987-19739987 on Build GRCh38
rs947443679 Chr.1: 19740065-19740065 on Build GRCh38
rs866612147 Chr.1: 19739822-19739822 on Build GRCh38
rs550102765 Chr.1: 19739975-19739975 on Build GRCh38
rs1009693372 Chr.1: 19739774-19739774 on Build GRCh38
rs1043456997 Chr.1: 19740074-19740074 on Build GRCh38
rs145902797 Chr.1: 19739897-19739897 on Build GRCh38
rs575894699 Chr.1: 19739904-19739904 on Build GRCh38
rs766155683 Chr.1: 19739905-19739905 on Build GRCh38
rs779319006 Chr.1: 19739879-19739879 on Build GRCh38
rs984192258 Chr.1: 19739917-19739917 on Build GRCh38
rs769673760 Chr.1: 19739800-19739800 on Build GRCh38
rs752786721 Chr.1: 19739854-19739854 on Build GRCh38
rs1033445767 Chr.1: 19740051-19740051 on Build GRCh38
rs750813714 Chr.1: 19740096-19740096 on Build GRCh38
rs753350080 Chr.1: 19739997-19739997 on Build GRCh38
rs773823888 Chr.1: 19739986-19739986 on Build GRCh38
rs753916968 Chr.1: 19739993-19739993 on Build GRCh38
rs767276118 Chr.1: 19740038-19740038 on Build GRCh38
rs944173905 Chr.1: 19739907-19739907 on Build GRCh38
rs778002131 Chr.1: 19739877-19739877 on Build GRCh38
rs760588280 Chr.1: 19739991-19739991 on Build GRCh38
rs777912210 Chr.1: 19739796-19739796 on Build GRCh38
rs200629068 Chr.1: 19739890-19739890 on Build GRCh38
rs564170029 Chr.1: 19739957-19739957 on Build GRCh38
rs371295326 Chr.1: 19739799-19739799 on Build GRCh38
rs889196477 Chr.1: 19740019-19740019 on Build GRCh38
rs574581882 Chr.1: 19739876-19739876 on Build GRCh38
rs144670706 Chr.1: 19739946-19739946 on Build GRCh38
rs369229688 Chr.1: 19739978-19739978 on Build GRCh38
rs754941185 Chr.1: 19739850-19739850 on Build GRCh38
rs909502079 Chr.1: 19739940-19739940 on Build GRCh38
rs59966976 Chr.1: 19739912-19739912 on Build GRCh38
rs770632490 Chr.1: 19739977-19739977 on Build GRCh38
rs6426825 Chr.1: 19739997-19739997 on Build GRCh38
rs374658138 Chr.1: 19739878-19739878 on Build GRCh38
rs576885219 Chr.1: 19739813-19739813 on Build GRCh38
rs755202204 Chr.1: 19739944-19739944 on Build GRCh38
rs755623318 Chr.1: 19739780-19739780 on Build GRCh38
rs746269000 Chr.1: 19739960-19739960 on Build GRCh38
rs766175122 Chr.1: 19739847-19739847 on Build GRCh38
rs531595830 Chr.1: 19739973-19739973 on Build GRCh38
rs147905112 Chr.1: 19740113-19740113 on Build GRCh38
rs750582126 Chr.1: 19740004-19740004 on Build GRCh38
rs772635020 Chr.1: 19739888-19739888 on Build GRCh38
rs769401993 Chr.1: 19739894-19739894 on Build GRCh38
rs775815849 Chr.1: 19739889-19739889 on Build GRCh38
rs780751086 Chr.1: 19739966-19739966 on Build GRCh38
rs751509507 Chr.1: 19739849-19739849 on Build GRCh38
rs142990957 Chr.1: 19739785-19739785 on Build GRCh38
rs920561211 Chr.1: 19740112-19740112 on Build GRCh38
rs758653993 Chr.1: 19739964-19739964 on Build GRCh38
rs747500995 Chr.1: 19739969-19739969 on Build GRCh38
rs959259158 Chr.1: 19739807-19739807 on Build GRCh38

View Full Product Details