Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23875090-23875614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNR2
Entrez Gene ID
1269
Gene Name
cannabinoid receptor 2
Gene Aliases
CB-2, CB2, CX5
UniGene
Hs.73037
Gene Chromosome Location
Chr.1: 23870526-23913327 on Build GRCh38
Transcripts
NM_001841.2 XM_011540629.2 XM_017000261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3486189 c.158C>T p.A53V Chr.1: 23875460-23875460 on Build GRCh38
533639 c.407G>A p.R136H Chr.1: 23875211-23875211 on Build GRCh38
110222 c.42G>A p.K14K Chr.1: 23875576-23875576 on Build GRCh38
3486180 c.468C>T p.I156I Chr.1: 23875150-23875150 on Build GRCh38
679674 c.291C>T p.F97F Chr.1: 23875327-23875327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879761216 Chr.1: 23875429-23875430 on Build GRCh38
rs751752206 Chr.1: 23875552-23875552 on Build GRCh38
rs755418976 Chr.1: 23875215-23875215 on Build GRCh38
rs200068439 Chr.1: 23875351-23875351 on Build GRCh38
rs749046000 Chr.1: 23875559-23875559 on Build GRCh38
rs773803426 Chr.1: 23875398-23875398 on Build GRCh38
rs916048109 Chr.1: 23875213-23875213 on Build GRCh38
rs141989735 Chr.1: 23875212-23875212 on Build GRCh38
rs199931185 Chr.1: 23875574-23875574 on Build GRCh38
rs199631465 Chr.1: 23875211-23875211 on Build GRCh38
rs41311993 Chr.1: 23875221-23875221 on Build GRCh38
rs147653938 Chr.1: 23875465-23875465 on Build GRCh38
rs144279977 Chr.1: 23875327-23875327 on Build GRCh38
rs764723293 Chr.1: 23875201-23875201 on Build GRCh38
rs769975822 Chr.1: 23875155-23875155 on Build GRCh38
rs199510108 Chr.1: 23875435-23875435 on Build GRCh38
rs758496486 Chr.1: 23875534-23875534 on Build GRCh38
rs764457545 Chr.1: 23875420-23875420 on Build GRCh38
rs111832243 Chr.1: 23875454-23875454 on Build GRCh38
rs776572645 Chr.1: 23875187-23875187 on Build GRCh38
rs750007574 Chr.1: 23875203-23875203 on Build GRCh38
rs80131168 Chr.1: 23875148-23875148 on Build GRCh38
rs762323628 Chr.1: 23875528-23875528 on Build GRCh38
rs769611593 Chr.1: 23875431-23875431 on Build GRCh38
rs1006481868 Chr.1: 23875231-23875231 on Build GRCh38
rs748062853 Chr.1: 23875246-23875258 on Build GRCh38
rs770989594 Chr.1: 23875232-23875232 on Build GRCh38
rs775178577 Chr.1: 23875586-23875586 on Build GRCh38
rs200666402 Chr.1: 23875488-23875488 on Build GRCh38
rs151085647 Chr.1: 23875576-23875576 on Build GRCh38
rs778488890 Chr.1: 23875114-23875114 on Build GRCh38
rs866049734 Chr.1: 23875335-23875335 on Build GRCh38
rs201650984 Chr.1: 23875368-23875368 on Build GRCh38
rs2501432 Chr.1: 23875430-23875430 on Build GRCh38
rs769849565 Chr.1: 23875383-23875383 on Build GRCh38
rs776450616 Chr.1: 23875246-23875246 on Build GRCh38
rs1009950229 Chr.1: 23875313-23875313 on Build GRCh38
rs780269365 Chr.1: 23875128-23875128 on Build GRCh38
rs756628656 Chr.1: 23875294-23875294 on Build GRCh38
rs543855954 Chr.1: 23875349-23875349 on Build GRCh38
rs765174269 Chr.1: 23875262-23875262 on Build GRCh38
rs1021275620 Chr.1: 23875408-23875408 on Build GRCh38
rs955203366 Chr.1: 23875359-23875359 on Build GRCh38
rs746669490 Chr.1: 23875591-23875591 on Build GRCh38
rs199615766 Chr.1: 23875486-23875486 on Build GRCh38
rs749285206 Chr.1: 23875318-23875318 on Build GRCh38
rs756316007 Chr.1: 23875371-23875371 on Build GRCh38
rs139756160 Chr.1: 23875289-23875289 on Build GRCh38
rs781772998 Chr.1: 23875144-23875144 on Build GRCh38
rs745320779 Chr.1: 23875571-23875571 on Build GRCh38
rs542890327 Chr.1: 23875178-23875178 on Build GRCh38
rs201802474 Chr.1: 23875324-23875324 on Build GRCh38
rs199785089 Chr.1: 23875222-23875222 on Build GRCh38
rs762850150 Chr.1: 23875354-23875354 on Build GRCh38
rs764487664 Chr.1: 23875477-23875477 on Build GRCh38
rs200980366 Chr.1: 23875299-23875299 on Build GRCh38
rs773937494 Chr.1: 23875440-23875440 on Build GRCh38
rs373993382 Chr.1: 23875159-23875159 on Build GRCh38
rs750464246 Chr.1: 23875531-23875531 on Build GRCh38
rs779046064 Chr.1: 23875423-23875423 on Build GRCh38
rs750020880 Chr.1: 23875117-23875117 on Build GRCh38
rs200635692 Chr.1: 23875190-23875190 on Build GRCh38
rs775303358 Chr.1: 23875464-23875464 on Build GRCh38
rs149656016 Chr.1: 23875543-23875543 on Build GRCh38
rs376344742 Chr.1: 23875570-23875570 on Build GRCh38
rs201688371 Chr.1: 23875233-23875233 on Build GRCh38
rs201087852 Chr.1: 23875143-23875143 on Build GRCh38
rs141481878 Chr.1: 23875410-23875410 on Build GRCh38
rs776353998 Chr.1: 23875348-23875348 on Build GRCh38
rs61996280 Chr.1: 23875421-23875421 on Build GRCh38
rs762556883 Chr.1: 23875278-23875278 on Build GRCh38
rs200099315 Chr.1: 23875373-23875373 on Build GRCh38
rs142134658 Chr.1: 23875575-23875575 on Build GRCh38
rs757932522 Chr.1: 23875208-23875208 on Build GRCh38
rs192526527 Chr.1: 23875453-23875453 on Build GRCh38
rs370109088 Chr.1: 23875402-23875402 on Build GRCh38
rs201821646 Chr.1: 23875529-23875529 on Build GRCh38
rs755226163 Chr.1: 23875554-23875554 on Build GRCh38
rs900368894 Chr.1: 23875560-23875560 on Build GRCh38
rs200843149 Chr.1: 23875360-23875360 on Build GRCh38
rs774664120 Chr.1: 23875236-23875236 on Build GRCh38
rs199691655 Chr.1: 23875226-23875226 on Build GRCh38
rs201896220 Chr.1: 23875411-23875411 on Build GRCh38
rs528804725 Chr.1: 23875425-23875425 on Build GRCh38
rs35761398 Chr.1: 23875429-23875430 on Build GRCh38
rs150233022 Chr.1: 23875227-23875227 on Build GRCh38
rs1008131094 Chr.1: 23875367-23875367 on Build GRCh38
rs772531845 Chr.1: 23875268-23875268 on Build GRCh38
rs749247846 Chr.1: 23875433-23875433 on Build GRCh38
rs778345235 Chr.1: 23875316-23875316 on Build GRCh38
rs754221879 Chr.1: 23875499-23875499 on Build GRCh38
rs758137578 Chr.1: 23875126-23875126 on Build GRCh38
rs950189696 Chr.1: 23875496-23875496 on Build GRCh38
rs2501431 Chr.1: 23875153-23875153 on Build GRCh38
rs761725003 Chr.1: 23875261-23875261 on Build GRCh38
rs200619419 Chr.1: 23875424-23875424 on Build GRCh38
rs747416117 Chr.1: 23875132-23875132 on Build GRCh38
rs145306214 Chr.1: 23875158-23875158 on Build GRCh38
rs768416819 Chr.1: 23875243-23875243 on Build GRCh38
rs772338354 Chr.1: 23875344-23875344 on Build GRCh38
rs142210955 Chr.1: 23875282-23875282 on Build GRCh38
rs201016971 Chr.1: 23875580-23875580 on Build GRCh38
rs775086010 Chr.1: 23875167-23875167 on Build GRCh38
rs760521443 Chr.1: 23875176-23875176 on Build GRCh38
rs527797875 Chr.1: 23875281-23875281 on Build GRCh38
rs761149846 Chr.1: 23875415-23875415 on Build GRCh38
rs201437984 Chr.1: 23875485-23875485 on Build GRCh38
rs759118298 Chr.1: 23875447-23875447 on Build GRCh38
rs754722865 Chr.1: 23875287-23875287 on Build GRCh38
rs746058590 Chr.1: 23875334-23875334 on Build GRCh38
rs201490624 Chr.1: 23875567-23875567 on Build GRCh38
rs182168081 Chr.1: 23875237-23875237 on Build GRCh38
rs200121304 Chr.1: 23875533-23875533 on Build GRCh38
rs757147880 Chr.1: 23875565-23875565 on Build GRCh38
rs781726068 Chr.1: 23875217-23875217 on Build GRCh38
rs757744382 Chr.1: 23875422-23875422 on Build GRCh38
rs201510809 Chr.1: 23875193-23875193 on Build GRCh38
rs200959861 Chr.1: 23875396-23875396 on Build GRCh38
rs930052487 Chr.1: 23875218-23875218 on Build GRCh38
rs966768377 Chr.1: 23875150-23875150 on Build GRCh38
rs769681479 Chr.1: 23875352-23875352 on Build GRCh38
rs770828039 Chr.1: 23875436-23875436 on Build GRCh38
rs773226744 Chr.1: 23875353-23875353 on Build GRCh38
rs370749734 Chr.1: 23875592-23875592 on Build GRCh38
rs767985981 Chr.1: 23875288-23875288 on Build GRCh38

View Full Product Details