Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82627104-82627565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB1
Entrez Gene ID
64506
Gene Name
cytoplasmic polyadenylation element binding protein 1
Gene Aliases
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1
UniGene
Hs.459132
Gene Chromosome Location
Chr.15: 82543201-82648016 on Build GRCh38
Transcripts
NM_001288819.1 NM_030594.4

SNP Detail

SNP ID Chromosome Location
rs971739297 Chr.15: 82627432-82627432 on Build GRCh38
rs949879994 Chr.15: 82627183-82627183 on Build GRCh38
rs1038316302 Chr.15: 82627269-82627269 on Build GRCh38
rs369500861 Chr.15: 82627209-82627209 on Build GRCh38
rs1032133476 Chr.15: 82627480-82627480 on Build GRCh38
rs376164471 Chr.15: 82627307-82627307 on Build GRCh38
rs370977022 Chr.15: 82627286-82627286 on Build GRCh38
rs759711264 Chr.15: 82627376-82627377 on Build GRCh38
rs1039853330 Chr.15: 82627538-82627538 on Build GRCh38
rs567063230 Chr.15: 82627437-82627437 on Build GRCh38
rs377521130 Chr.15: 82627268-82627268 on Build GRCh38
rs929941343 Chr.15: 82627494-82627494 on Build GRCh38
rs779904823 Chr.15: 82627406-82627406 on Build GRCh38
rs748700912 Chr.15: 82627354-82627354 on Build GRCh38
rs182186196 Chr.15: 82627442-82627442 on Build GRCh38
rs1007580391 Chr.15: 82627521-82627521 on Build GRCh38
rs748604452 Chr.15: 82627327-82627327 on Build GRCh38
rs776426811 Chr.15: 82627170-82627171 on Build GRCh38
rs767520769 Chr.15: 82627223-82627223 on Build GRCh38
rs927143091 Chr.15: 82627449-82627449 on Build GRCh38
rs755323018 Chr.15: 82627411-82627411 on Build GRCh38
rs917133557 Chr.15: 82627421-82627421 on Build GRCh38
rs767043771 Chr.15: 82627396-82627396 on Build GRCh38
rs753113802 Chr.15: 82627377-82627378 on Build GRCh38
rs746567716 Chr.15: 82627146-82627146 on Build GRCh38
rs941472138 Chr.15: 82627229-82627229 on Build GRCh38
rs369595604 Chr.15: 82627175-82627175 on Build GRCh38
rs528444931 Chr.15: 82627343-82627343 on Build GRCh38
rs201709014 Chr.15: 82627404-82627404 on Build GRCh38
rs900253384 Chr.15: 82627495-82627495 on Build GRCh38
rs963444630 Chr.15: 82627540-82627540 on Build GRCh38
rs752553777 Chr.15: 82627245-82627245 on Build GRCh38
rs868523340 Chr.15: 82627325-82627325 on Build GRCh38
rs770218142 Chr.15: 82627188-82627188 on Build GRCh38
rs774544798 Chr.15: 82627208-82627208 on Build GRCh38
rs751474815 Chr.15: 82627282-82627282 on Build GRCh38
rs374349658 Chr.15: 82627306-82627306 on Build GRCh38
rs774555678 Chr.15: 82627371-82627372 on Build GRCh38
rs373688655 Chr.15: 82627189-82627189 on Build GRCh38
rs200431699 Chr.15: 82627280-82627280 on Build GRCh38
rs769267245 Chr.15: 82627313-82627313 on Build GRCh38
rs941453651 Chr.15: 82627457-82627457 on Build GRCh38
rs190478412 Chr.15: 82627418-82627418 on Build GRCh38
rs185189989 Chr.15: 82627402-82627402 on Build GRCh38
rs766627809 Chr.15: 82627356-82627356 on Build GRCh38
rs762688770 Chr.15: 82627378-82627378 on Build GRCh38
rs749802457 Chr.15: 82627204-82627204 on Build GRCh38
rs542858157 Chr.15: 82627358-82627360 on Build GRCh38
rs149322138 Chr.15: 82627143-82627143 on Build GRCh38
rs763147503 Chr.15: 82627314-82627316 on Build GRCh38
rs769357207 Chr.15: 82627207-82627207 on Build GRCh38
rs202018031 Chr.15: 82627360-82627360 on Build GRCh38
rs551177350 Chr.15: 82627398-82627398 on Build GRCh38
rs780574053 Chr.15: 82627298-82627298 on Build GRCh38
rs760880558 Chr.15: 82627347-82627347 on Build GRCh38
rs897203190 Chr.15: 82627291-82627291 on Build GRCh38
rs753571999 Chr.15: 82627408-82627408 on Build GRCh38
rs923070859 Chr.15: 82627265-82627265 on Build GRCh38
rs999968581 Chr.15: 82627392-82627392 on Build GRCh38
rs758301379 Chr.15: 82627279-82627279 on Build GRCh38
rs757156470 Chr.15: 82627387-82627387 on Build GRCh38
rs760914474 Chr.15: 82627475-82627475 on Build GRCh38
rs946299481 Chr.15: 82627407-82627407 on Build GRCh38
rs144574454 Chr.15: 82627380-82627380 on Build GRCh38
rs202055971 Chr.15: 82627477-82627477 on Build GRCh38
rs773415266 Chr.15: 82627234-82627234 on Build GRCh38
rs185973111 Chr.15: 82627506-82627506 on Build GRCh38
rs776012986 Chr.15: 82627197-82627197 on Build GRCh38
rs760817775 Chr.15: 82627235-82627235 on Build GRCh38
rs772128934 Chr.15: 82627341-82627341 on Build GRCh38
rs745469220 Chr.15: 82627305-82627305 on Build GRCh38
rs891352361 Chr.15: 82627520-82627520 on Build GRCh38
rs1038453404 Chr.15: 82627476-82627476 on Build GRCh38
rs767895344 Chr.15: 82627378-82627378 on Build GRCh38
rs765002708 Chr.15: 82627236-82627236 on Build GRCh38
rs61733713 Chr.15: 82627322-82627322 on Build GRCh38
rs761900804 Chr.15: 82627216-82627216 on Build GRCh38
rs1051254966 Chr.15: 82627323-82627323 on Build GRCh38
rs531040238 Chr.15: 82627385-82627385 on Build GRCh38
rs756837753 Chr.15: 82627168-82627168 on Build GRCh38
rs995846709 Chr.15: 82627497-82627497 on Build GRCh38
rs1032496028 Chr.15: 82627133-82627133 on Build GRCh38
rs370569656 Chr.15: 82627312-82627312 on Build GRCh38

View Full Product Details