Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71377426-71377856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
UniGene
Hs.106857
Gene Chromosome Location
Chr.16: 71358713-71390438 on Build GRCh38
Transcripts
NM_001740.4 NM_007088.3 NR_027910.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3511822 c.294C>T p.F98F Chr.16: 71377699-71377699 on Build GRCh38
3511821 c.282C>T p.T94T Chr.16: 71377687-71377687 on Build GRCh38
4062524 c.330C>T p.A110A Chr.16: 71377735-71377735 on Build GRCh38
6145109 c.337A>G p.M113V Chr.16: 71377742-71377742 on Build GRCh38
4642051 c.266C>T p.A89V Chr.16: 71377671-71377671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915303683 Chr.16: 71377821-71377821 on Build GRCh38
rs760678290 Chr.16: 71377653-71377653 on Build GRCh38
rs769689206 Chr.16: 71377645-71377645 on Build GRCh38
rs750440280 Chr.16: 71377765-71377765 on Build GRCh38
rs369562892 Chr.16: 71377620-71377620 on Build GRCh38
rs771060964 Chr.16: 71377720-71377720 on Build GRCh38
rs754772666 Chr.16: 71377778-71377778 on Build GRCh38
rs115276994 Chr.16: 71377622-71377622 on Build GRCh38
rs533404802 Chr.16: 71377552-71377552 on Build GRCh38
rs762996634 Chr.16: 71377647-71377647 on Build GRCh38
rs868229129 Chr.16: 71377470-71377470 on Build GRCh38
rs1045987069 Chr.16: 71377742-71377742 on Build GRCh38
rs749852827 Chr.16: 71377741-71377741 on Build GRCh38
rs547555720 Chr.16: 71377469-71377469 on Build GRCh38
rs774214374 Chr.16: 71377706-71377706 on Build GRCh38
rs748994795 Chr.16: 71377688-71377688 on Build GRCh38
rs1002124184 Chr.16: 71377786-71377786 on Build GRCh38
rs1027717636 Chr.16: 71377802-71377802 on Build GRCh38
rs201815729 Chr.16: 71377672-71377672 on Build GRCh38
rs887273271 Chr.16: 71377572-71377572 on Build GRCh38
rs771299430 Chr.16: 71377553-71377553 on Build GRCh38
rs758759188 Chr.16: 71377499-71377500 on Build GRCh38
rs757836019 Chr.16: 71377736-71377736 on Build GRCh38
rs771978416 Chr.16: 71377783-71377783 on Build GRCh38
rs768426697 Chr.16: 71377648-71377648 on Build GRCh38
rs373382211 Chr.16: 71377523-71377523 on Build GRCh38
rs765330463 Chr.16: 71377725-71377725 on Build GRCh38
rs553158988 Chr.16: 71377675-71377675 on Build GRCh38
rs146835226 Chr.16: 71377735-71377735 on Build GRCh38
rs141424820 Chr.16: 71377717-71377717 on Build GRCh38
rs529476369 Chr.16: 71377465-71377465 on Build GRCh38
rs578007426 Chr.16: 71377732-71377732 on Build GRCh38
rs551647579 Chr.16: 71377571-71377571 on Build GRCh38
rs769715159 Chr.16: 71377501-71377501 on Build GRCh38
rs777719934 Chr.16: 71377521-71377521 on Build GRCh38
rs748121287 Chr.16: 71377767-71377767 on Build GRCh38
rs906081120 Chr.16: 71377515-71377515 on Build GRCh38
rs375481271 Chr.16: 71377740-71377740 on Build GRCh38
rs749119287 Chr.16: 71377759-71377759 on Build GRCh38
rs946691480 Chr.16: 71377527-71377527 on Build GRCh38
rs548839248 Chr.16: 71377639-71377639 on Build GRCh38
rs759855172 Chr.16: 71377721-71377721 on Build GRCh38
rs567321690 Chr.16: 71377667-71377667 on Build GRCh38
rs988255646 Chr.16: 71377763-71377763 on Build GRCh38
rs373286098 Chr.16: 71377621-71377621 on Build GRCh38
rs752986143 Chr.16: 71377670-71377670 on Build GRCh38
rs12445308 Chr.16: 71377543-71377543 on Build GRCh38
rs776797163 Chr.16: 71377795-71377795 on Build GRCh38
rs774761406 Chr.16: 71377556-71377556 on Build GRCh38
rs754114195 Chr.16: 71377657-71377657 on Build GRCh38
rs11545954 Chr.16: 71377733-71377733 on Build GRCh38
rs768640497 Chr.16: 71377693-71377693 on Build GRCh38
rs770739478 Chr.16: 71377632-71377632 on Build GRCh38
rs779504574 Chr.16: 71377750-71377750 on Build GRCh38
rs150938838 Chr.16: 71377669-71377669 on Build GRCh38
rs780837012 Chr.16: 71377684-71377684 on Build GRCh38
rs988612028 Chr.16: 71377495-71377495 on Build GRCh38
rs1014670775 Chr.16: 71377588-71377588 on Build GRCh38
rs569387795 Chr.16: 71377459-71377459 on Build GRCh38
rs868479855 Chr.16: 71377745-71377745 on Build GRCh38
rs1015478415 Chr.16: 71377563-71377563 on Build GRCh38
rs754822412 Chr.16: 71377762-71377762 on Build GRCh38
rs775116406 Chr.16: 71377646-71377646 on Build GRCh38
rs925706197 Chr.16: 71377713-71377713 on Build GRCh38
rs762792017 Chr.16: 71377787-71377788 on Build GRCh38
rs887864498 Chr.16: 71377831-71377831 on Build GRCh38
rs759800878 Chr.16: 71377576-71377576 on Build GRCh38
rs745602535 Chr.16: 71377687-71377687 on Build GRCh38
rs755820576 Chr.16: 71377743-71377743 on Build GRCh38
rs766599222 Chr.16: 71377655-71377655 on Build GRCh38
rs537029177 Chr.16: 71377638-71377638 on Build GRCh38
rs904170106 Chr.16: 71377447-71377448 on Build GRCh38

View Full Product Details