Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39404572-39405078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED1
Entrez Gene ID
5469
Gene Name
mediator complex subunit 1
Gene Aliases
CRSP1, CRSP200, DRIP205, DRIP230, PBP, PPARBP, PPARGBP, RB18A, TRAP220, TRIP2
UniGene
Hs.643754
Gene Chromosome Location
Chr.17: 39404285-39451281 on Build GRCh38
Transcripts
NM_004774.3 XM_005257465.3 XM_006721957.1 XM_017024779.1

SNP Detail

SNP ID Chromosome Location
rs765533859 Chr.17: 39404672-39404672 on Build GRCh38
rs1024932283 Chr.17: 39404712-39404712 on Build GRCh38
rs72825175 Chr.17: 39404878-39404878 on Build GRCh38
rs755507048 Chr.17: 39404654-39404654 on Build GRCh38
rs750949083 Chr.17: 39405005-39405006 on Build GRCh38
rs1025216305 Chr.17: 39405050-39405050 on Build GRCh38
rs953680035 Chr.17: 39404858-39404872 on Build GRCh38
rs980492413 Chr.17: 39404817-39404817 on Build GRCh38
rs559364278 Chr.17: 39404874-39404874 on Build GRCh38
rs899133800 Chr.17: 39404622-39404622 on Build GRCh38
rs945584004 Chr.17: 39404867-39404867 on Build GRCh38
rs957680228 Chr.17: 39404849-39404849 on Build GRCh38
rs181636540 Chr.17: 39404595-39404595 on Build GRCh38
rs891504590 Chr.17: 39404724-39404724 on Build GRCh38
rs189187103 Chr.17: 39404679-39404679 on Build GRCh38
rs899166647 Chr.17: 39404904-39404904 on Build GRCh38
rs912174152 Chr.17: 39404872-39404872 on Build GRCh38
rs989194841 Chr.17: 39404858-39404858 on Build GRCh38
rs985750011 Chr.17: 39404870-39404871 on Build GRCh38
rs886948038 Chr.17: 39404668-39404668 on Build GRCh38
rs972888163 Chr.17: 39404876-39404876 on Build GRCh38
rs186772439 Chr.17: 39404611-39404611 on Build GRCh38
rs191900896 Chr.17: 39404856-39404856 on Build GRCh38
rs142636174 Chr.17: 39404634-39404634 on Build GRCh38
rs960795430 Chr.17: 39404917-39404917 on Build GRCh38
rs930596898 Chr.17: 39404913-39404913 on Build GRCh38
rs781557045 Chr.17: 39404841-39404841 on Build GRCh38
rs753019086 Chr.17: 39404871-39404871 on Build GRCh38
rs777999964 Chr.17: 39404991-39404991 on Build GRCh38
rs565204487 Chr.17: 39404612-39404612 on Build GRCh38
rs1047875741 Chr.17: 39404952-39404952 on Build GRCh38
rs925857734 Chr.17: 39405026-39405026 on Build GRCh38
rs998277049 Chr.17: 39404782-39404782 on Build GRCh38
rs895753625 Chr.17: 39404623-39404623 on Build GRCh38
rs1029303096 Chr.17: 39404827-39404827 on Build GRCh38
rs971033484 Chr.17: 39404814-39404814 on Build GRCh38
rs1039431360 Chr.17: 39404616-39404616 on Build GRCh38
rs1031844486 Chr.17: 39404681-39404681 on Build GRCh38
rs1026362005 Chr.17: 39404655-39404655 on Build GRCh38
rs750750242 Chr.17: 39405023-39405023 on Build GRCh38
rs913621842 Chr.17: 39404863-39404863 on Build GRCh38
rs147618332 Chr.17: 39404768-39404768 on Build GRCh38
rs749548765 Chr.17: 39405014-39405014 on Build GRCh38
rs181827831 Chr.17: 39404882-39404882 on Build GRCh38

View Full Product Details