Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65902046-65902549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
UniGene
Hs.408676
Gene Chromosome Location
Chr.17: 65635540-66192131 on Build GRCh38
Transcripts
NM_001199165.2 NM_001302891.1 XM_005257119.4 XM_005257125.3 XM_005257126.4 XM_006721740.2 XM_006721744.3 XM_011524461.2 XM_011524462.2 XM_011524463.2 XM_011524464.2 XM_011524465.2 XM_011524466.2 XM_011524467.2 XM_017024302.1 XM_017024303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3717541 c.2077G>A p.E693K Chr.17: 65902238-65902238 on Build GRCh38
3717540 c.2077G>A p.E693K Chr.17: 65902238-65902238 on Build GRCh38
6885541 c.2004T>G p.H668Q Chr.17: 65902311-65902311 on Build GRCh38
2796863 c.2037T>C p.H679H Chr.17: 65902278-65902278 on Build GRCh38
2796864 c.2037T>C p.H679H Chr.17: 65902278-65902278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772137559 Chr.17: 65902350-65902350 on Build GRCh38
rs776632088 Chr.17: 65902245-65902245 on Build GRCh38
rs374621153 Chr.17: 65902209-65902209 on Build GRCh38
rs566285272 Chr.17: 65902479-65902479 on Build GRCh38
rs954853093 Chr.17: 65902099-65902099 on Build GRCh38
rs755176722 Chr.17: 65902305-65902305 on Build GRCh38
rs758846950 Chr.17: 65902379-65902379 on Build GRCh38
rs148746783 Chr.17: 65902378-65902378 on Build GRCh38
rs751353344 Chr.17: 65902162-65902162 on Build GRCh38
rs977260749 Chr.17: 65902356-65902356 on Build GRCh38
rs865863224 Chr.17: 65902423-65902423 on Build GRCh38
rs781455707 Chr.17: 65902313-65902313 on Build GRCh38
rs759312321 Chr.17: 65902163-65902163 on Build GRCh38
rs931999211 Chr.17: 65902100-65902100 on Build GRCh38
rs760615774 Chr.17: 65902354-65902354 on Build GRCh38
rs748223257 Chr.17: 65902315-65902315 on Build GRCh38
rs946691767 Chr.17: 65902331-65902331 on Build GRCh38
rs751913737 Chr.17: 65902386-65902386 on Build GRCh38
rs770893972 Chr.17: 65902247-65902247 on Build GRCh38
rs750850539 Chr.17: 65902280-65902280 on Build GRCh38
rs1030345405 Chr.17: 65902468-65902468 on Build GRCh38
rs764616838 Chr.17: 65902377-65902377 on Build GRCh38
rs779356668 Chr.17: 65902208-65902208 on Build GRCh38
rs981905213 Chr.17: 65902234-65902234 on Build GRCh38
rs773362493 Chr.17: 65902334-65902334 on Build GRCh38
rs559140469 Chr.17: 65902157-65902157 on Build GRCh38
rs370129214 Chr.17: 65902191-65902191 on Build GRCh38
rs935819319 Chr.17: 65902376-65902376 on Build GRCh38
rs1003951950 Chr.17: 65902451-65902451 on Build GRCh38
rs998981140 Chr.17: 65902076-65902078 on Build GRCh38
rs772335011 Chr.17: 65902122-65902122 on Build GRCh38
rs772544083 Chr.17: 65902119-65902119 on Build GRCh38
rs767166589 Chr.17: 65902168-65902168 on Build GRCh38
rs780332003 Chr.17: 65902292-65902292 on Build GRCh38
rs372070681 Chr.17: 65902291-65902291 on Build GRCh38
rs745751336 Chr.17: 65902342-65902342 on Build GRCh38
rs749761864 Chr.17: 65902103-65902103 on Build GRCh38
rs557300941 Chr.17: 65902104-65902104 on Build GRCh38
rs766926443 Chr.17: 65902382-65902382 on Build GRCh38
rs779280343 Chr.17: 65902110-65902110 on Build GRCh38
rs747385055 Chr.17: 65902297-65902297 on Build GRCh38
rs765756567 Chr.17: 65902279-65902279 on Build GRCh38
rs761553569 Chr.17: 65902375-65902375 on Build GRCh38
rs775413916 Chr.17: 65902353-65902353 on Build GRCh38
rs376831833 Chr.17: 65902367-65902367 on Build GRCh38
rs1003667392 Chr.17: 65902215-65902215 on Build GRCh38
rs944734870 Chr.17: 65902355-65902355 on Build GRCh38
rs760725208 Chr.17: 65902124-65902124 on Build GRCh38
rs200648869 Chr.17: 65902136-65902136 on Build GRCh38
rs777376240 Chr.17: 65902199-65902199 on Build GRCh38
rs1043440288 Chr.17: 65902339-65902339 on Build GRCh38
rs780421901 Chr.17: 65902211-65902211 on Build GRCh38
rs772585374 Chr.17: 65902523-65902523 on Build GRCh38
rs924475297 Chr.17: 65902357-65902357 on Build GRCh38
rs757769831 Chr.17: 65902201-65902201 on Build GRCh38
rs200079739 Chr.17: 65902252-65902252 on Build GRCh38
rs147238839 Chr.17: 65902253-65902253 on Build GRCh38
rs906967462 Chr.17: 65902448-65902448 on Build GRCh38
rs763898618 Chr.17: 65902267-65902267 on Build GRCh38
rs758742223 Chr.17: 65902210-65902210 on Build GRCh38
rs144307988 Chr.17: 65902330-65902330 on Build GRCh38
rs777571631 Chr.17: 65902374-65902374 on Build GRCh38
rs189286288 Chr.17: 65902123-65902123 on Build GRCh38
rs948406212 Chr.17: 65902398-65902398 on Build GRCh38
rs768676888 Chr.17: 65902125-65902125 on Build GRCh38
rs986643719 Chr.17: 65902102-65902102 on Build GRCh38
rs1015293799 Chr.17: 65902469-65902469 on Build GRCh38
rs762552462 Chr.17: 65902122-65902122 on Build GRCh38
rs867484198 Chr.17: 65902221-65902221 on Build GRCh38
rs1007641850 Chr.17: 65902521-65902521 on Build GRCh38
rs149561928 Chr.17: 65902241-65902241 on Build GRCh38
rs775454965 Chr.17: 65902255-65902255 on Build GRCh38
rs920625447 Chr.17: 65902083-65902083 on Build GRCh38
rs201577288 Chr.17: 65902274-65902274 on Build GRCh38
rs766237557 Chr.17: 65902150-65902150 on Build GRCh38
rs774191517 Chr.17: 65902248-65902248 on Build GRCh38
rs370915298 Chr.17: 65902120-65902120 on Build GRCh38
rs973382579 Chr.17: 65902075-65902075 on Build GRCh38
rs74004953 Chr.17: 65902235-65902235 on Build GRCh38
rs140758713 Chr.17: 65902275-65902275 on Build GRCh38
rs1030897781 Chr.17: 65902097-65902097 on Build GRCh38
rs760335740 Chr.17: 65902262-65902262 on Build GRCh38
rs894381332 Chr.17: 65902383-65902383 on Build GRCh38
rs762879948 Chr.17: 65902141-65902141 on Build GRCh38
rs180805977 Chr.17: 65902190-65902190 on Build GRCh38
rs763809049 Chr.17: 65902364-65902364 on Build GRCh38
rs1045362043 Chr.17: 65902427-65902427 on Build GRCh38
rs1021668300 Chr.17: 65902070-65902078 on Build GRCh38

View Full Product Details