Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67692868-67693407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITPNC1
Entrez Gene ID
26207
Gene Name
phosphatidylinositol transfer protein, cytoplasmic 1
Gene Aliases
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1
UniGene
Hs.591185
Gene Chromosome Location
Chr.17: 67377281-67697263 on Build GRCh38
Transcripts
NM_012417.3 NM_181671.2 XM_005257216.2 XM_017024442.1 XM_017024443.1 XM_017024444.1 XM_017024445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3820321 c.994G>C p.E332Q Chr.17: 67692883-67692883 on Build GRCh38
3820320 c.994G>C p.E332Q Chr.17: 67692883-67692883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775974697 Chr.17: 67693355-67693355 on Build GRCh38
rs773832575 Chr.17: 67692936-67692937 on Build GRCh38
rs576096667 Chr.17: 67692938-67692939 on Build GRCh38
rs887299554 Chr.17: 67693284-67693284 on Build GRCh38
rs1051481994 Chr.17: 67693102-67693102 on Build GRCh38
rs772744545 Chr.17: 67693174-67693174 on Build GRCh38
rs959832570 Chr.17: 67693059-67693059 on Build GRCh38
rs991540974 Chr.17: 67693104-67693104 on Build GRCh38
rs1003032363 Chr.17: 67693317-67693317 on Build GRCh38
rs1007938383 Chr.17: 67692959-67692959 on Build GRCh38
rs951286164 Chr.17: 67693381-67693381 on Build GRCh38
rs765673355 Chr.17: 67692895-67692895 on Build GRCh38
rs964281446 Chr.17: 67692990-67692990 on Build GRCh38
rs750872640 Chr.17: 67692925-67692925 on Build GRCh38
rs946454739 Chr.17: 67693038-67693038 on Build GRCh38
rs995415318 Chr.17: 67693002-67693002 on Build GRCh38
rs573337833 Chr.17: 67692938-67692938 on Build GRCh38
rs192287230 Chr.17: 67693138-67693138 on Build GRCh38
rs763372147 Chr.17: 67692937-67692937 on Build GRCh38
rs928337417 Chr.17: 67693058-67693058 on Build GRCh38
rs548189345 Chr.17: 67693020-67693020 on Build GRCh38
rs923809287 Chr.17: 67693077-67693077 on Build GRCh38
rs770192380 Chr.17: 67692934-67692935 on Build GRCh38
rs145595114 Chr.17: 67693251-67693251 on Build GRCh38
rs182429463 Chr.17: 67693363-67693363 on Build GRCh38
rs553392605 Chr.17: 67693266-67693266 on Build GRCh38
rs780525412 Chr.17: 67692938-67692938 on Build GRCh38
rs898468274 Chr.17: 67693165-67693165 on Build GRCh38
rs1039549696 Chr.17: 67693332-67693332 on Build GRCh38
rs140392622 Chr.17: 67693009-67693009 on Build GRCh38
rs113322475 Chr.17: 67693036-67693036 on Build GRCh38
rs928887603 Chr.17: 67692953-67692953 on Build GRCh38
rs367988456 Chr.17: 67692946-67692947 on Build GRCh38
rs569927742 Chr.17: 67693101-67693101 on Build GRCh38
rs1043307502 Chr.17: 67693208-67693208 on Build GRCh38
rs147971105 Chr.17: 67692937-67692938 on Build GRCh38
rs552136990 Chr.17: 67693157-67693157 on Build GRCh38
rs1017617388 Chr.17: 67692971-67692971 on Build GRCh38
rs762468901 Chr.17: 67693179-67693179 on Build GRCh38
rs760284229 Chr.17: 67692925-67692926 on Build GRCh38
rs915561319 Chr.17: 67693133-67693133 on Build GRCh38
rs942598938 Chr.17: 67693128-67693128 on Build GRCh38
rs1007660811 Chr.17: 67693325-67693325 on Build GRCh38
rs1025776890 Chr.17: 67692938-67692940 on Build GRCh38
rs1043486516 Chr.17: 67693068-67693068 on Build GRCh38
rs995278698 Chr.17: 67693361-67693361 on Build GRCh38
rs776241483 Chr.17: 67693000-67693000 on Build GRCh38
rs960682254 Chr.17: 67692987-67692987 on Build GRCh38
rs559298577 Chr.17: 67692940-67692940 on Build GRCh38
rs935160519 Chr.17: 67693086-67693087 on Build GRCh38
rs890511558 Chr.17: 67693308-67693308 on Build GRCh38
rs756781745 Chr.17: 67692893-67692893 on Build GRCh38
rs951184977 Chr.17: 67693017-67693017 on Build GRCh38
rs1006878349 Chr.17: 67692906-67692906 on Build GRCh38
rs1051886399 Chr.17: 67693248-67693248 on Build GRCh38
rs758823596 Chr.17: 67692937-67692937 on Build GRCh38
rs924907146 Chr.17: 67693226-67693226 on Build GRCh38
rs774954163 Chr.17: 67692926-67692928 on Build GRCh38
rs78871228 Chr.17: 67693235-67693235 on Build GRCh38
rs999013699 Chr.17: 67692935-67692935 on Build GRCh38

View Full Product Details