Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73350159-73350663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDK2
Entrez Gene ID
54549
Gene Name
sidekick cell adhesion molecule 2
Gene Aliases
-
UniGene
Hs.435719
Gene Chromosome Location
Chr.17: 73334384-73644482 on Build GRCh38
Transcripts
NM_001144952.1 XM_011524914.2 XM_011524915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1710720 c.5990G>C p.R1997P Chr.17: 73350285-73350285 on Build GRCh38
6081580 c.5932A>T p.S1978C Chr.17: 73350343-73350343 on Build GRCh38
3742418 c.5989C>T p.R1997W Chr.17: 73350286-73350286 on Build GRCh38
5812494 c.5927G>A p.G1976D Chr.17: 73350348-73350348 on Build GRCh38
4724446 c.5933G>T p.S1978I Chr.17: 73350342-73350342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754307392 Chr.17: 73350219-73350219 on Build GRCh38
rs767237872 Chr.17: 73350363-73350363 on Build GRCh38
rs747218544 Chr.17: 73350329-73350329 on Build GRCh38
rs765775983 Chr.17: 73350259-73350259 on Build GRCh38
rs978960368 Chr.17: 73350517-73350517 on Build GRCh38
rs202068153 Chr.17: 73350602-73350602 on Build GRCh38
rs570393817 Chr.17: 73350552-73350552 on Build GRCh38
rs147802955 Chr.17: 73350430-73350430 on Build GRCh38
rs959680519 Chr.17: 73350450-73350450 on Build GRCh38
rs953425430 Chr.17: 73350547-73350547 on Build GRCh38
rs759140095 Chr.17: 73350348-73350348 on Build GRCh38
rs1036379176 Chr.17: 73350613-73350613 on Build GRCh38
rs748511818 Chr.17: 73350401-73350401 on Build GRCh38
rs979813613 Chr.17: 73350423-73350423 on Build GRCh38
rs377641221 Chr.17: 73350638-73350638 on Build GRCh38
rs149660021 Chr.17: 73350386-73350386 on Build GRCh38
rs777149158 Chr.17: 73350254-73350254 on Build GRCh38
rs761034315 Chr.17: 73350250-73350250 on Build GRCh38
rs143527749 Chr.17: 73350285-73350285 on Build GRCh38
rs548344628 Chr.17: 73350446-73350446 on Build GRCh38
rs538168947 Chr.17: 73350582-73350582 on Build GRCh38
rs921798525 Chr.17: 73350518-73350518 on Build GRCh38
rs375803616 Chr.17: 73350213-73350214 on Build GRCh38
rs780265782 Chr.17: 73350325-73350325 on Build GRCh38
rs750105039 Chr.17: 73350212-73350213 on Build GRCh38
rs1995163 Chr.17: 73350611-73350611 on Build GRCh38
rs534579286 Chr.17: 73350579-73350579 on Build GRCh38
rs556777830 Chr.17: 73350614-73350614 on Build GRCh38
rs777905116 Chr.17: 73350316-73350316 on Build GRCh38
rs141196259 Chr.17: 73350503-73350503 on Build GRCh38
rs1041851047 Chr.17: 73350283-73350283 on Build GRCh38
rs779624996 Chr.17: 73350225-73350225 on Build GRCh38
rs747154765 Chr.17: 73350387-73350387 on Build GRCh38
rs369215097 Chr.17: 73350606-73350606 on Build GRCh38
rs369870973 Chr.17: 73350286-73350286 on Build GRCh38
rs913110150 Chr.17: 73350492-73350492 on Build GRCh38
rs771236746 Chr.17: 73350188-73350189 on Build GRCh38
rs749740859 Chr.17: 73350219-73350225 on Build GRCh38
rs781453694 Chr.17: 73350394-73350394 on Build GRCh38
rs138003419 Chr.17: 73350278-73350278 on Build GRCh38
rs1045761406 Chr.17: 73350626-73350626 on Build GRCh38
rs764649017 Chr.17: 73350206-73350206 on Build GRCh38
rs796165915 Chr.17: 73350223-73350224 on Build GRCh38
rs116738350 Chr.17: 73350317-73350317 on Build GRCh38
rs748106343 Chr.17: 73350233-73350233 on Build GRCh38
rs760646905 Chr.17: 73350209-73350210 on Build GRCh38
rs775891133 Chr.17: 73350200-73350200 on Build GRCh38
rs752640012 Chr.17: 73350296-73350296 on Build GRCh38
rs774602104 Chr.17: 73350240-73350240 on Build GRCh38
rs753840920 Chr.17: 73350383-73350383 on Build GRCh38
rs754800221 Chr.17: 73350230-73350230 on Build GRCh38
rs761719987 Chr.17: 73350211-73350212 on Build GRCh38
rs773777944 Chr.17: 73350341-73350341 on Build GRCh38
rs200095133 Chr.17: 73350232-73350232 on Build GRCh38
rs756085035 Chr.17: 73350306-73350306 on Build GRCh38
rs988463989 Chr.17: 73350487-73350487 on Build GRCh38
rs905366621 Chr.17: 73350627-73350627 on Build GRCh38
rs751171693 Chr.17: 73350345-73350345 on Build GRCh38
rs775632022 Chr.17: 73350245-73350245 on Build GRCh38
rs760994064 Chr.17: 73350205-73350205 on Build GRCh38
rs61494348 Chr.17: 73350214-73350215 on Build GRCh38
rs764130031 Chr.17: 73350210-73350211 on Build GRCh38
rs900559101 Chr.17: 73350224-73350224 on Build GRCh38
rs149822544 Chr.17: 73350190-73350191 on Build GRCh38
rs35317925 Chr.17: 73350374-73350374 on Build GRCh38
rs376004218 Chr.17: 73350384-73350384 on Build GRCh38
rs770153144 Chr.17: 73350416-73350416 on Build GRCh38
rs186667264 Chr.17: 73350494-73350494 on Build GRCh38
rs995102833 Chr.17: 73350620-73350620 on Build GRCh38
rs752517423 Chr.17: 73350370-73350370 on Build GRCh38
rs569602552 Chr.17: 73350344-73350344 on Build GRCh38
rs759857459 Chr.17: 73350189-73350189 on Build GRCh38
rs771082663 Chr.17: 73350234-73350234 on Build GRCh38
rs772383473 Chr.17: 73350222-73350223 on Build GRCh38
rs867413079 Chr.17: 73350367-73350367 on Build GRCh38
rs765259732 Chr.17: 73350233-73350236 on Build GRCh38
rs762340608 Chr.17: 73350258-73350258 on Build GRCh38
rs774573777 Chr.17: 73350208-73350209 on Build GRCh38
rs370418227 Chr.17: 73350217-73350218 on Build GRCh38
rs189944854 Chr.17: 73350546-73350546 on Build GRCh38
rs758017688 Chr.17: 73350223-73350223 on Build GRCh38
rs62071382 Chr.17: 73350190-73350190 on Build GRCh38
rs372386732 Chr.17: 73350337-73350337 on Build GRCh38
rs779319498 Chr.17: 73350220-73350221 on Build GRCh38
rs767953091 Chr.17: 73350196-73350196 on Build GRCh38
rs139480135 Chr.17: 73350340-73350340 on Build GRCh38
rs541528366 Chr.17: 73350242-73350242 on Build GRCh38
rs767062535 Chr.17: 73350635-73350635 on Build GRCh38
rs532789356 Chr.17: 73350358-73350358 on Build GRCh38
rs370472124 Chr.17: 73350218-73350219 on Build GRCh38
rs768917020 Chr.17: 73350251-73350251 on Build GRCh38
rs749778543 Chr.17: 73350604-73350604 on Build GRCh38
rs375027080 Chr.17: 73350221-73350221 on Build GRCh38
rs375316369 Chr.17: 73350277-73350277 on Build GRCh38
rs769026772 Chr.17: 73350335-73350335 on Build GRCh38
rs761775372 Chr.17: 73350229-73350230 on Build GRCh38
rs756699553 Chr.17: 73350220-73350220 on Build GRCh38
rs147120981 Chr.17: 73350191-73350192 on Build GRCh38
rs749916035 Chr.17: 73350264-73350264 on Build GRCh38
rs754407828 Chr.17: 73350215-73350216 on Build GRCh38
rs912006473 Chr.17: 73350578-73350578 on Build GRCh38
rs776722908 Chr.17: 73350226-73350227 on Build GRCh38
rs746208643 Chr.17: 73350223-73350224 on Build GRCh38

View Full Product Details