Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 58315855-58316389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEDD4L
Entrez Gene ID
23327
Gene Name
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
UniGene
Hs.185677
Gene Chromosome Location
Chr.18: 58044362-58401540 on Build GRCh38
Transcripts
NM_001144964.1 NM_001144965.1 NM_001144966.2 NM_001144967.2 NM_001144968.1 NM_001144969.1 NM_001144970.2 NM_001144971.1 NM_001243960.1 NM_015277.5 XM_005266658.4 XM_005266660.4 XM_005266663.4 XM_006722421.3 XM_006722424.3 XM_006722425.3 XM_006722426.3 XM_006722428.3 XM_006722430.3 XM_011525887.2 XM_017025676.1 XM_017025677.1 XM_017025678.1 XM_017025679.1 XM_017025680.1 XM_017025681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3526688 c.343C>T p.L115F Chr.18: 58316027-58316027 on Build GRCh38
5424805 c.302G>A p.R101Q Chr.18: 58315986-58315986 on Build GRCh38
4487007 c.312C>T p.F104F Chr.18: 58315996-58315996 on Build GRCh38
94809 c.329T>A p.V110E Chr.18: 58316013-58316013 on Build GRCh38
5699791 c.326A>T p.D109V Chr.18: 58316010-58316010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005893193 Chr.18: 58316271-58316271 on Build GRCh38
rs745369151 Chr.18: 58316038-58316038 on Build GRCh38
rs200559702 Chr.18: 58315934-58315934 on Build GRCh38
rs749302499 Chr.18: 58316249-58316249 on Build GRCh38
rs190673965 Chr.18: 58316219-58316219 on Build GRCh38
rs765817235 Chr.18: 58315991-58315991 on Build GRCh38
rs755683700 Chr.18: 58315963-58315964 on Build GRCh38
rs771260662 Chr.18: 58316069-58316069 on Build GRCh38
rs146524660 Chr.18: 58316218-58316218 on Build GRCh38
rs751163365 Chr.18: 58316040-58316040 on Build GRCh38
rs938603801 Chr.18: 58316236-58316236 on Build GRCh38
rs938595447 Chr.18: 58315911-58315911 on Build GRCh38
rs1003424018 Chr.18: 58315910-58315910 on Build GRCh38
rs758747799 Chr.18: 58316011-58316011 on Build GRCh38
rs756725290 Chr.18: 58316025-58316025 on Build GRCh38
rs749840819 Chr.18: 58315956-58315957 on Build GRCh38
rs766498556 Chr.18: 58315965-58315965 on Build GRCh38
rs934978732 Chr.18: 58316217-58316217 on Build GRCh38
rs773308533 Chr.18: 58316060-58316060 on Build GRCh38
rs1057233817 Chr.18: 58315949-58315949 on Build GRCh38
rs775322435 Chr.18: 58315938-58315938 on Build GRCh38
rs549531 Chr.18: 58316088-58316088 on Build GRCh38
rs369076553 Chr.18: 58316248-58316248 on Build GRCh38
rs776899514 Chr.18: 58316071-58316071 on Build GRCh38
rs779527001 Chr.18: 58316050-58316050 on Build GRCh38
rs772224378 Chr.18: 58315947-58315947 on Build GRCh38
rs780748593 Chr.18: 58316035-58316035 on Build GRCh38
rs371368723 Chr.18: 58316042-58316042 on Build GRCh38
rs974790399 Chr.18: 58316327-58316327 on Build GRCh38
rs763242448 Chr.18: 58316081-58316081 on Build GRCh38
rs894162995 Chr.18: 58316247-58316247 on Build GRCh38
rs987680690 Chr.18: 58316199-58316199 on Build GRCh38
rs777837830 Chr.18: 58316244-58316244 on Build GRCh38
rs543228992 Chr.18: 58316041-58316041 on Build GRCh38
rs894106473 Chr.18: 58315950-58315950 on Build GRCh38
rs868850266 Chr.18: 58316020-58316020 on Build GRCh38
rs2288775 Chr.18: 58316132-58316132 on Build GRCh38
rs947747646 Chr.18: 58316231-58316231 on Build GRCh38
rs762787679 Chr.18: 58315939-58315939 on Build GRCh38
rs924240816 Chr.18: 58316200-58316200 on Build GRCh38
rs903129525 Chr.18: 58315907-58315907 on Build GRCh38
rs898677760 Chr.18: 58316061-58316061 on Build GRCh38
rs1020041149 Chr.18: 58316128-58316128 on Build GRCh38
rs887058378 Chr.18: 58316255-58316255 on Build GRCh38
rs975489526 Chr.18: 58316152-58316152 on Build GRCh38
rs776014085 Chr.18: 58316074-58316074 on Build GRCh38
rs527841100 Chr.18: 58315899-58315899 on Build GRCh38
rs1023452219 Chr.18: 58315891-58315891 on Build GRCh38
rs559224703 Chr.18: 58316127-58316127 on Build GRCh38
rs760174391 Chr.18: 58315977-58315977 on Build GRCh38
rs13339710 Chr.18: 58316299-58316299 on Build GRCh38
rs779541843 Chr.18: 58316026-58316026 on Build GRCh38
rs1012018130 Chr.18: 58315880-58315880 on Build GRCh38
rs1025792318 Chr.18: 58316075-58316075 on Build GRCh38
rs993244090 Chr.18: 58316285-58316285 on Build GRCh38
rs1044846644 Chr.18: 58316245-58316245 on Build GRCh38
rs760681849 Chr.18: 58315954-58315954 on Build GRCh38
rs777311647 Chr.18: 58316012-58316012 on Build GRCh38
rs765875924 Chr.18: 58316073-58316073 on Build GRCh38
rs898939376 Chr.18: 58316278-58316278 on Build GRCh38
rs933525299 Chr.18: 58316187-58316187 on Build GRCh38
rs368265913 Chr.18: 58315887-58315887 on Build GRCh38
rs753151234 Chr.18: 58316002-58316002 on Build GRCh38
rs970554451 Chr.18: 58315908-58315908 on Build GRCh38
rs781124616 Chr.18: 58315933-58315933 on Build GRCh38
rs768438187 Chr.18: 58316052-58316052 on Build GRCh38
rs924458420 Chr.18: 58316233-58316233 on Build GRCh38
rs773445999 Chr.18: 58315948-58315948 on Build GRCh38
rs887249134 Chr.18: 58316364-58316364 on Build GRCh38
rs201646504 Chr.18: 58316072-58316072 on Build GRCh38
rs2288774 Chr.18: 58316098-58316098 on Build GRCh38
rs758170250 Chr.18: 58316194-58316194 on Build GRCh38
rs753959894 Chr.18: 58315975-58315975 on Build GRCh38
rs745620701 Chr.18: 58316153-58316153 on Build GRCh38
rs572854572 Chr.18: 58316031-58316031 on Build GRCh38

View Full Product Details