Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46814986-46815483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4B1
Entrez Gene ID
1580
Gene Name
cytochrome P450 family 4 subfamily B member 1
Gene Aliases
CYPIVB1, P-450HP
UniGene
Hs.436317
Gene Chromosome Location
Chr.1: 46798995-46819349 on Build GRCh38
Transcripts
NM_000779.3 NM_001099772.1 NM_001319161.1 NM_001319162.1 NM_001319163.1 NR_135003.1 XM_011540832.1 XM_011540833.1 XM_017000466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4239025 c.881A>G p.D294G Chr.1: 46815075-46815075 on Build GRCh38
4239029 c.1039G>A p.E347K Chr.1: 46815233-46815233 on Build GRCh38
328058 c.919G>A p.A307T Chr.1: 46815113-46815113 on Build GRCh38
5053553 c.924A>G p.E308E Chr.1: 46815118-46815118 on Build GRCh38
5353154 c.902C>A p.S301* Chr.1: 46815096-46815096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985552751 Chr.1: 46815317-46815317 on Build GRCh38
rs913182822 Chr.1: 46815359-46815359 on Build GRCh38
rs375551930 Chr.1: 46815067-46815067 on Build GRCh38
rs764596873 Chr.1: 46815162-46815162 on Build GRCh38
rs562875489 Chr.1: 46815405-46815405 on Build GRCh38
rs17102592 Chr.1: 46815255-46815255 on Build GRCh38
rs985537525 Chr.1: 46815351-46815351 on Build GRCh38
rs771109140 Chr.1: 46815219-46815219 on Build GRCh38
rs746996053 Chr.1: 46815093-46815093 on Build GRCh38
rs939844636 Chr.1: 46815020-46815020 on Build GRCh38
rs201173499 Chr.1: 46815116-46815116 on Build GRCh38
rs747049267 Chr.1: 46815215-46815215 on Build GRCh38
rs766262731 Chr.1: 46815253-46815255 on Build GRCh38
rs749885609 Chr.1: 46815169-46815169 on Build GRCh38
rs12058654 Chr.1: 46815355-46815355 on Build GRCh38
rs774825270 Chr.1: 46815284-46815284 on Build GRCh38
rs752544893 Chr.1: 46815210-46815210 on Build GRCh38
rs144531409 Chr.1: 46815113-46815113 on Build GRCh38
rs375320663 Chr.1: 46815050-46815050 on Build GRCh38
rs4646491 Chr.1: 46815212-46815212 on Build GRCh38
rs759047880 Chr.1: 46815301-46815301 on Build GRCh38
rs1054716409 Chr.1: 46815361-46815361 on Build GRCh38
rs45574436 Chr.1: 46815013-46815013 on Build GRCh38
rs994417074 Chr.1: 46815458-46815458 on Build GRCh38
rs2297810 Chr.1: 46815187-46815187 on Build GRCh38
rs765891281 Chr.1: 46815221-46815221 on Build GRCh38
rs775722853 Chr.1: 46815071-46815071 on Build GRCh38
rs961240081 Chr.1: 46815234-46815234 on Build GRCh38
rs778569400 Chr.1: 46815209-46815209 on Build GRCh38
rs150836169 Chr.1: 46815410-46815410 on Build GRCh38
rs571066348 Chr.1: 46815027-46815027 on Build GRCh38
rs755652341 Chr.1: 46815171-46815171 on Build GRCh38
rs749071537 Chr.1: 46815107-46815107 on Build GRCh38
rs576671037 Chr.1: 46815360-46815360 on Build GRCh38
rs762356887 Chr.1: 46815269-46815269 on Build GRCh38
rs911219675 Chr.1: 46815318-46815318 on Build GRCh38
rs1036016277 Chr.1: 46815422-46815422 on Build GRCh38
rs778817288 Chr.1: 46815111-46815111 on Build GRCh38
rs544278601 Chr.1: 46815392-46815392 on Build GRCh38
rs55835239 Chr.1: 46815076-46815076 on Build GRCh38
rs776958699 Chr.1: 46815135-46815135 on Build GRCh38
rs1003287250 Chr.1: 46815415-46815415 on Build GRCh38
rs762334759 Chr.1: 46815232-46815232 on Build GRCh38
rs772138641 Chr.1: 46815019-46815019 on Build GRCh38
rs370191155 Chr.1: 46815285-46815285 on Build GRCh38
rs747102740 Chr.1: 46815127-46815127 on Build GRCh38
rs559148619 Chr.1: 46815017-46815017 on Build GRCh38
rs1045581531 Chr.1: 46815368-46815368 on Build GRCh38
rs777880279 Chr.1: 46815213-46815213 on Build GRCh38
rs1034465987 Chr.1: 46815225-46815225 on Build GRCh38
rs913450475 Chr.1: 46815385-46815385 on Build GRCh38
rs774809111 Chr.1: 46815259-46815259 on Build GRCh38
rs879050864 Chr.1: 46815073-46815076 on Build GRCh38
rs1017174444 Chr.1: 46815253-46815253 on Build GRCh38
rs759846966 Chr.1: 46815144-46815144 on Build GRCh38
rs755703651 Chr.1: 46815094-46815094 on Build GRCh38
rs765503290 Chr.1: 46815150-46815150 on Build GRCh38
rs199691504 Chr.1: 46815231-46815231 on Build GRCh38
rs894824005 Chr.1: 46815442-46815442 on Build GRCh38
rs372816291 Chr.1: 46815086-46815086 on Build GRCh38
rs753456688 Chr.1: 46815292-46815292 on Build GRCh38
rs762373084 Chr.1: 46815391-46815391 on Build GRCh38
rs767801042 Chr.1: 46815408-46815408 on Build GRCh38
rs12094024 Chr.1: 46815180-46815180 on Build GRCh38
rs45467195 Chr.1: 46815158-46815158 on Build GRCh38
rs754700084 Chr.1: 46815110-46815110 on Build GRCh38
rs972924736 Chr.1: 46815256-46815256 on Build GRCh38
rs773363257 Chr.1: 46815124-46815124 on Build GRCh38
rs972851891 Chr.1: 46815037-46815037 on Build GRCh38
rs3215983 Chr.1: 46815075-46815076 on Build GRCh38
rs555752365 Chr.1: 46815412-46815412 on Build GRCh38
rs56180718 Chr.1: 46815230-46815230 on Build GRCh38
rs763330238 Chr.1: 46815245-46815245 on Build GRCh38
rs771233725 Chr.1: 46815131-46815131 on Build GRCh38
rs375248128 Chr.1: 46815102-46815102 on Build GRCh38
rs867826742 Chr.1: 46815073-46815074 on Build GRCh38
rs867329257 Chr.1: 46815091-46815091 on Build GRCh38
rs1033571671 Chr.1: 46815010-46815010 on Build GRCh38
rs754719484 Chr.1: 46815208-46815208 on Build GRCh38
rs753120838 Chr.1: 46815061-46815061 on Build GRCh38
rs935974628 Chr.1: 46815326-46815326 on Build GRCh38
rs764958518 Chr.1: 46815315-46815315 on Build GRCh38
rs759843798 Chr.1: 46815026-46815026 on Build GRCh38
rs879813088 Chr.1: 46815435-46815435 on Build GRCh38
rs775987211 Chr.1: 46815157-46815157 on Build GRCh38
rs142539703 Chr.1: 46815121-46815121 on Build GRCh38

View Full Product Details