Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47023820-47024322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4X1
Entrez Gene ID
260293
Gene Name
cytochrome P450 family 4 subfamily X member 1
Gene Aliases
CYPIVX1
UniGene
Hs.439760
Gene Chromosome Location
Chr.1: 46961364-47055432 on Build GRCh38
Transcripts
NM_001320289.1 NM_001320290.1 NM_178033.1 XM_017000973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310453 c.129G>T p.L43L Chr.1: 47023946-47023946 on Build GRCh38
3805473 c.174G>A p.Q58Q Chr.1: 47023991-47023991 on Build GRCh38
6371916 c.7T>G p.F3V Chr.1: 47023824-47023824 on Build GRCh38
4676035 c.138delC p.P47fs*57 Chr.1: 47023955-47023955 on Build GRCh38
4008618 c.82C>T p.Q28* Chr.1: 47023899-47023899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001760627 Chr.1: 47024222-47024222 on Build GRCh38
rs2036462 Chr.1: 47024114-47024114 on Build GRCh38
rs879680156 Chr.1: 47024156-47024156 on Build GRCh38
rs749773055 Chr.1: 47023878-47023878 on Build GRCh38
rs914205029 Chr.1: 47024083-47024083 on Build GRCh38
rs900509402 Chr.1: 47023936-47023936 on Build GRCh38
rs765481296 Chr.1: 47024005-47024006 on Build GRCh38
rs1049648008 Chr.1: 47024127-47024127 on Build GRCh38
rs979847430 Chr.1: 47024107-47024107 on Build GRCh38
rs568738179 Chr.1: 47024124-47024124 on Build GRCh38
rs367790270 Chr.1: 47023970-47023970 on Build GRCh38
rs769006787 Chr.1: 47023884-47023884 on Build GRCh38
rs1054525042 Chr.1: 47023863-47023863 on Build GRCh38
rs891878276 Chr.1: 47024165-47024165 on Build GRCh38
rs886338020 Chr.1: 47023872-47023872 on Build GRCh38
rs370355060 Chr.1: 47023931-47023931 on Build GRCh38
rs766676475 Chr.1: 47024034-47024034 on Build GRCh38
rs1037623078 Chr.1: 47024288-47024288 on Build GRCh38
rs776164387 Chr.1: 47024001-47024001 on Build GRCh38
rs745581131 Chr.1: 47023851-47023862 on Build GRCh38
rs200133350 Chr.1: 47024016-47024016 on Build GRCh38
rs761891775 Chr.1: 47024033-47024033 on Build GRCh38
rs774426082 Chr.1: 47024032-47024032 on Build GRCh38
rs750652797 Chr.1: 47023964-47023964 on Build GRCh38
rs774948210 Chr.1: 47023890-47023890 on Build GRCh38
rs909624593 Chr.1: 47024064-47024064 on Build GRCh38
rs748724526 Chr.1: 47023991-47023991 on Build GRCh38
rs887976484 Chr.1: 47024268-47024268 on Build GRCh38
rs775600612 Chr.1: 47023967-47023967 on Build GRCh38
rs372846355 Chr.1: 47023871-47023871 on Build GRCh38
rs766226102 Chr.1: 47024292-47024292 on Build GRCh38
rs972427115 Chr.1: 47024108-47024108 on Build GRCh38
rs765055118 Chr.1: 47023922-47023922 on Build GRCh38
rs762596066 Chr.1: 47024022-47024022 on Build GRCh38
rs761958336 Chr.1: 47024002-47024022 on Build GRCh38
rs935805186 Chr.1: 47024140-47024140 on Build GRCh38
rs770410664 Chr.1: 47024000-47024000 on Build GRCh38
rs780418220 Chr.1: 47023845-47023845 on Build GRCh38
rs757338909 Chr.1: 47023949-47023949 on Build GRCh38
rs760124136 Chr.1: 47023895-47023895 on Build GRCh38
rs961372960 Chr.1: 47024080-47024080 on Build GRCh38
rs371579159 Chr.1: 47024037-47024037 on Build GRCh38
rs112563041 Chr.1: 47024243-47024243 on Build GRCh38
rs531717187 Chr.1: 47024088-47024088 on Build GRCh38
rs1031154053 Chr.1: 47024001-47024011 on Build GRCh38
rs1034494415 Chr.1: 47023984-47023984 on Build GRCh38
rs267598627 Chr.1: 47023979-47023979 on Build GRCh38
rs560078689 Chr.1: 47023976-47023976 on Build GRCh38
rs373809441 Chr.1: 47023842-47023842 on Build GRCh38
rs568407658 Chr.1: 47024280-47024280 on Build GRCh38
rs558253703 Chr.1: 47024122-47024122 on Build GRCh38
rs958280970 Chr.1: 47024011-47024011 on Build GRCh38
rs763886896 Chr.1: 47023944-47023944 on Build GRCh38
rs752472908 Chr.1: 47023929-47023929 on Build GRCh38
rs1034546446 Chr.1: 47024236-47024236 on Build GRCh38
rs1045740559 Chr.1: 47024173-47024173 on Build GRCh38
rs750515947 Chr.1: 47024006-47024019 on Build GRCh38
rs560385073 Chr.1: 47023866-47023866 on Build GRCh38
rs535058800 Chr.1: 47023851-47023851 on Build GRCh38
rs539638964 Chr.1: 47023962-47023962 on Build GRCh38
rs781480815 Chr.1: 47023957-47023957 on Build GRCh38
rs779329616 Chr.1: 47023988-47023988 on Build GRCh38
rs771694180 Chr.1: 47023945-47023946 on Build GRCh38
rs759241090 Chr.1: 47023921-47023921 on Build GRCh38
rs781505792 Chr.1: 47023862-47023862 on Build GRCh38
rs763313704 Chr.1: 47024029-47024030 on Build GRCh38
rs745486385 Chr.1: 47024014-47024014 on Build GRCh38
rs770435494 Chr.1: 47023905-47023905 on Build GRCh38
rs773034674 Chr.1: 47024196-47024196 on Build GRCh38
rs757434709 Chr.1: 47023860-47023860 on Build GRCh38
rs775308109 Chr.1: 47023961-47023962 on Build GRCh38
rs111783250 Chr.1: 47024170-47024170 on Build GRCh38
rs557731759 Chr.1: 47024123-47024124 on Build GRCh38
rs970146163 Chr.1: 47024086-47024086 on Build GRCh38
rs763953294 Chr.1: 47024026-47024026 on Build GRCh38
rs7495119 Chr.1: 47024015-47024015 on Build GRCh38
rs527743703 Chr.1: 47023914-47023914 on Build GRCh38
rs748068507 Chr.1: 47024104-47024104 on Build GRCh38
rs911079421 Chr.1: 47024128-47024128 on Build GRCh38
rs1010232269 Chr.1: 47023930-47023930 on Build GRCh38
rs760336516 Chr.1: 47024214-47024214 on Build GRCh38
rs529533653 Chr.1: 47024166-47024166 on Build GRCh38
rs768661114 Chr.1: 47024000-47024001 on Build GRCh38
rs751647509 Chr.1: 47023859-47023859 on Build GRCh38

View Full Product Details