Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47260437-47260948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STIL
Entrez Gene ID
6491
Gene Name
SCL/TAL1 interrupting locus
Gene Aliases
MCPH7, SIL
UniGene
Hs.525198
Gene Chromosome Location
Chr.1: 47250139-47314787 on Build GRCh38
Transcripts
NM_001048166.1 NM_001282936.1 NM_001282937.1 NM_001282938.1 NM_001282939.1 NM_003035.2 XM_006710834.3 XM_011541991.2 XM_011541992.2 XM_011541994.2 XM_011541996.2 XM_011541998.2 XM_011542001.2 XM_017002123.1 XM_017002124.1 XM_017002125.1 XM_017002126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3930802 c.2858C>G p.S953C Chr.1: 47260508-47260508 on Build GRCh38
5789458 c.2831A>G p.Q944R Chr.1: 47260535-47260535 on Build GRCh38
5491114 c.2843T>G p.L948R Chr.1: 47260523-47260523 on Build GRCh38
5846907 c.2926G>A p.V976I Chr.1: 47260440-47260440 on Build GRCh38
4827464 c.2844A>T p.L948L Chr.1: 47260522-47260522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747434600 Chr.1: 47260922-47260924 on Build GRCh38
rs1041456479 Chr.1: 47260899-47260899 on Build GRCh38
rs551855961 Chr.1: 47260576-47260576 on Build GRCh38
rs761623433 Chr.1: 47260492-47260492 on Build GRCh38
rs1046315091 Chr.1: 47260485-47260485 on Build GRCh38
rs535732677 Chr.1: 47260732-47260732 on Build GRCh38
rs560131105 Chr.1: 47260731-47260731 on Build GRCh38
rs1026838859 Chr.1: 47260767-47260767 on Build GRCh38
rs759462016 Chr.1: 47260581-47260582 on Build GRCh38
rs779782062 Chr.1: 47260476-47260476 on Build GRCh38
rs199422204 Chr.1: 47260511-47260511 on Build GRCh38
rs963916757 Chr.1: 47260817-47260817 on Build GRCh38
rs765585891 Chr.1: 47260558-47260558 on Build GRCh38
rs748944081 Chr.1: 47260578-47260578 on Build GRCh38
rs1030668907 Chr.1: 47260708-47260708 on Build GRCh38
rs1057314040 Chr.1: 47260687-47260687 on Build GRCh38
rs745331343 Chr.1: 47260464-47260464 on Build GRCh38
rs894695688 Chr.1: 47260703-47260703 on Build GRCh38
rs1013125711 Chr.1: 47260718-47260718 on Build GRCh38
rs772094091 Chr.1: 47260496-47260496 on Build GRCh38
rs914240475 Chr.1: 47260849-47260849 on Build GRCh38
rs748958516 Chr.1: 47260477-47260477 on Build GRCh38
rs778548961 Chr.1: 47260585-47260585 on Build GRCh38
rs764767965 Chr.1: 47260550-47260551 on Build GRCh38
rs1039830675 Chr.1: 47260924-47260924 on Build GRCh38
rs113486760 Chr.1: 47260696-47260696 on Build GRCh38
rs768557739 Chr.1: 47260478-47260478 on Build GRCh38
rs755623551 Chr.1: 47260573-47260573 on Build GRCh38
rs929948863 Chr.1: 47260845-47260845 on Build GRCh38
rs983068330 Chr.1: 47260850-47260851 on Build GRCh38
rs771861493 Chr.1: 47260632-47260632 on Build GRCh38
rs967688692 Chr.1: 47260653-47260653 on Build GRCh38
rs879375940 Chr.1: 47260550-47260550 on Build GRCh38
rs776777269 Chr.1: 47260538-47260538 on Build GRCh38
rs773094005 Chr.1: 47260557-47260558 on Build GRCh38
rs531221797 Chr.1: 47260621-47260621 on Build GRCh38
rs556538999 Chr.1: 47260662-47260662 on Build GRCh38
rs888442644 Chr.1: 47260559-47260559 on Build GRCh38
rs1023316884 Chr.1: 47260763-47260763 on Build GRCh38
rs769217306 Chr.1: 47260470-47260470 on Build GRCh38
rs376840068 Chr.1: 47260908-47260908 on Build GRCh38
rs148721559 Chr.1: 47260635-47260635 on Build GRCh38
rs146333610 Chr.1: 47260878-47260878 on Build GRCh38
rs766451599 Chr.1: 47260522-47260522 on Build GRCh38
rs35447382 Chr.1: 47260514-47260514 on Build GRCh38
rs114760199 Chr.1: 47260593-47260593 on Build GRCh38
rs757492545 Chr.1: 47260690-47260690 on Build GRCh38
rs773046719 Chr.1: 47260504-47260504 on Build GRCh38
rs148193936 Chr.1: 47260463-47260463 on Build GRCh38
rs201148192 Chr.1: 47260584-47260584 on Build GRCh38
rs553744455 Chr.1: 47260741-47260741 on Build GRCh38
rs142251133 Chr.1: 47260794-47260794 on Build GRCh38
rs576598109 Chr.1: 47260816-47260816 on Build GRCh38
rs774354721 Chr.1: 47260489-47260489 on Build GRCh38
rs533307267 Chr.1: 47260559-47260559 on Build GRCh38
rs938464405 Chr.1: 47260912-47260912 on Build GRCh38
rs993539175 Chr.1: 47260755-47260755 on Build GRCh38
rs1057473875 Chr.1: 47260925-47260925 on Build GRCh38
rs191072430 Chr.1: 47260749-47260749 on Build GRCh38
rs752010409 Chr.1: 47260571-47260571 on Build GRCh38
rs540520514 Chr.1: 47260652-47260652 on Build GRCh38
rs766314697 Chr.1: 47260558-47260559 on Build GRCh38
rs764618787 Chr.1: 47260560-47260560 on Build GRCh38
rs574815683 Chr.1: 47260840-47260840 on Build GRCh38
rs751428385 Chr.1: 47260580-47260582 on Build GRCh38
rs767787674 Chr.1: 47260795-47260795 on Build GRCh38

View Full Product Details