Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54834823-54835333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR2DS4
Entrez Gene ID
3809
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
Gene Aliases
CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
UniGene
Hs.654608
Gene Chromosome Location
Chr.19: 54832676-54848569 on Build GRCh38
Transcripts
NM_001281971.1 NM_001281972.1 NM_012314.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3539069 c.35-1G>A p.? Chr.19: 54835078-54835078 on Build GRCh38
1001242 c.39C>A p.F13L Chr.19: 54835083-54835083 on Build GRCh38
1001244 c.46C>A p.L16M Chr.19: 54835090-54835090 on Build GRCh38
5009558 c.56C>G p.A19G Chr.19: 54835100-54835100 on Build GRCh38
4132557 c.66G>T p.Q22H Chr.19: 54835110-54835110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763935469 Chr.19: 54835057-54835057 on Build GRCh38
rs145598651 Chr.19: 54835020-54835020 on Build GRCh38
rs111595618 Chr.19: 54835204-54835204 on Build GRCh38
rs755509351 Chr.19: 54835095-54835095 on Build GRCh38
rs148171453 Chr.19: 54835019-54835019 on Build GRCh38
rs571225473 Chr.19: 54835249-54835249 on Build GRCh38
rs367562122 Chr.19: 54834905-54834905 on Build GRCh38
rs758746278 Chr.19: 54835104-54835104 on Build GRCh38
rs116088042 Chr.19: 54835280-54835280 on Build GRCh38
rs373284780 Chr.19: 54834988-54834988 on Build GRCh38
rs943382441 Chr.19: 54835092-54835092 on Build GRCh38
rs770620066 Chr.19: 54835042-54835042 on Build GRCh38
rs139015764 Chr.19: 54834888-54834888 on Build GRCh38
rs139958085 Chr.19: 54835174-54835174 on Build GRCh38
rs761878936 Chr.19: 54835066-54835066 on Build GRCh38
rs373466969 Chr.19: 54835065-54835065 on Build GRCh38
rs773458154 Chr.19: 54835154-54835154 on Build GRCh38
rs779078299 Chr.19: 54835097-54835097 on Build GRCh38
rs945189845 Chr.19: 54835016-54835016 on Build GRCh38
rs374758431 Chr.19: 54835270-54835270 on Build GRCh38
rs553770893 Chr.19: 54835136-54835136 on Build GRCh38
rs368915789 Chr.19: 54835008-54835008 on Build GRCh38
rs143816115 Chr.19: 54835054-54835054 on Build GRCh38
rs148903134 Chr.19: 54835218-54835218 on Build GRCh38
rs745987291 Chr.19: 54835223-54835223 on Build GRCh38
rs749201992 Chr.19: 54835129-54835129 on Build GRCh38
rs143472745 Chr.19: 54834859-54834859 on Build GRCh38
rs373262836 Chr.19: 54835161-54835161 on Build GRCh38
rs144268808 Chr.19: 54834899-54834899 on Build GRCh38
rs1130478 Chr.19: 54835110-54835110 on Build GRCh38
rs754830500 Chr.19: 54834886-54834886 on Build GRCh38
rs745547102 Chr.19: 54835112-54835112 on Build GRCh38
rs143336635 Chr.19: 54835047-54835047 on Build GRCh38
rs753119279 Chr.19: 54835098-54835098 on Build GRCh38
rs769408167 Chr.19: 54835114-54835114 on Build GRCh38
rs745370068 Chr.19: 54835046-54835046 on Build GRCh38
rs111384927 Chr.19: 54835268-54835268 on Build GRCh38
rs76344177 Chr.19: 54835310-54835310 on Build GRCh38
rs112522228 Chr.19: 54835079-54835079 on Build GRCh38
rs59333104 Chr.19: 54835203-54835203 on Build GRCh38
rs570251088 Chr.19: 54835263-54835263 on Build GRCh38
rs550408561 Chr.19: 54835254-54835254 on Build GRCh38
rs906761464 Chr.19: 54835211-54835211 on Build GRCh38
rs535663558 Chr.19: 54835028-54835028 on Build GRCh38
rs993618760 Chr.19: 54835177-54835177 on Build GRCh38
rs879219526 Chr.19: 54835272-54835272 on Build GRCh38
rs4806585 Chr.19: 54834969-54834969 on Build GRCh38
rs146702057 Chr.19: 54835035-54835035 on Build GRCh38
rs554369463 Chr.19: 54834855-54834855 on Build GRCh38
rs771714427 Chr.19: 54835293-54835293 on Build GRCh38
rs370536807 Chr.19: 54834931-54834931 on Build GRCh38
rs386810985 Chr.19: 54834899-54834900 on Build GRCh38
rs144473427 Chr.19: 54834900-54834900 on Build GRCh38
rs186270835 Chr.19: 54834959-54834959 on Build GRCh38
rs878997591 Chr.19: 54835273-54835273 on Build GRCh38
rs181032470 Chr.19: 54834929-54834929 on Build GRCh38
rs767797896 Chr.19: 54835076-54835076 on Build GRCh38
rs375560081 Chr.19: 54834996-54834996 on Build GRCh38
rs560221946 Chr.19: 54834895-54834895 on Build GRCh38
rs574628008 Chr.19: 54834861-54834861 on Build GRCh38
rs749543000 Chr.19: 54834957-54834957 on Build GRCh38
rs200101952 Chr.19: 54835088-54835088 on Build GRCh38
rs4806584 Chr.19: 54834954-54834954 on Build GRCh38
rs761659732 Chr.19: 54835141-54835141 on Build GRCh38
rs894823729 Chr.19: 54835197-54835197 on Build GRCh38
rs4806583 Chr.19: 54834918-54834918 on Build GRCh38
rs141689303 Chr.19: 54835089-54835089 on Build GRCh38
rs962931153 Chr.19: 54834910-54834910 on Build GRCh38
rs751338642 Chr.19: 54834883-54834883 on Build GRCh38
rs763035815 Chr.19: 54835055-54835055 on Build GRCh38
rs112892367 Chr.19: 54835176-54835176 on Build GRCh38
rs541653542 Chr.19: 54835198-54835198 on Build GRCh38
rs79131460 Chr.19: 54835235-54835235 on Build GRCh38
rs540088615 Chr.19: 54835011-54835011 on Build GRCh38
rs369561024 Chr.19: 54834972-54834972 on Build GRCh38
rs533174931 Chr.19: 54835248-54835248 on Build GRCh38
rs770869305 Chr.19: 54835023-54835023 on Build GRCh38
rs547364737 Chr.19: 54834974-54834974 on Build GRCh38
rs754290458 Chr.19: 54835093-54835093 on Build GRCh38
rs139861291 Chr.19: 54835085-54835085 on Build GRCh38
rs202012905 Chr.19: 54835192-54835192 on Build GRCh38
rs746601108 Chr.19: 54835036-54835036 on Build GRCh38
rs959309740 Chr.19: 54834924-54834924 on Build GRCh38
rs779734808 Chr.19: 54835117-54835117 on Build GRCh38
rs771799255 Chr.19: 54835153-54835153 on Build GRCh38
rs551212537 Chr.19: 54835286-54835286 on Build GRCh38
rs111366340 Chr.19: 54835213-54835213 on Build GRCh38
rs760907130 Chr.19: 54835159-54835159 on Build GRCh38
rs1026512388 Chr.19: 54835189-54835189 on Build GRCh38
rs111740515 Chr.19: 54835137-54835137 on Build GRCh38
rs775945114 Chr.19: 54835044-54835044 on Build GRCh38
rs982172735 Chr.19: 54834852-54834852 on Build GRCh38

View Full Product Details