Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54865713-54866220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR3DL2
Entrez Gene ID
3812
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
Gene Aliases
3DL2, CD158K, KIR-3DL2, NKAT-4, NKAT4, NKAT4B, p140
UniGene
Hs.645228
Gene Chromosome Location
Chr.19: 54850319-54867215 on Build GRCh38
Transcripts
NM_001242867.1 NM_006737.3 XM_011526940.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390722 c.1077C>T p.L359L Chr.19: 54865881-54865881 on Build GRCh38
1305099 c.1015C>T p.L339L Chr.19: 54865819-54865819 on Build GRCh38
5017957 c.1018C>T p.H340Y Chr.19: 54865822-54865822 on Build GRCh38
128335 c.1032G>A p.G344G Chr.19: 54865836-54865836 on Build GRCh38
5533426 c.1049T>A p.F350Y Chr.19: 54865853-54865853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956565738 Chr.19: 54866118-54866118 on Build GRCh38
rs757555683 Chr.19: 54865898-54865898 on Build GRCh38
rs774769920 Chr.19: 54865954-54865954 on Build GRCh38
rs371490654 Chr.19: 54865800-54865800 on Build GRCh38
rs770864062 Chr.19: 54865871-54865871 on Build GRCh38
rs921935079 Chr.19: 54866157-54866157 on Build GRCh38
rs138402445 Chr.19: 54865888-54865888 on Build GRCh38
rs200958705 Chr.19: 54865765-54865765 on Build GRCh38
rs575648653 Chr.19: 54865962-54865962 on Build GRCh38
rs753489436 Chr.19: 54865900-54865900 on Build GRCh38
rs751892996 Chr.19: 54865823-54865823 on Build GRCh38
rs369829524 Chr.19: 54865921-54865921 on Build GRCh38
rs759606315 Chr.19: 54865801-54865801 on Build GRCh38
rs10417848 Chr.19: 54865972-54865972 on Build GRCh38
rs762334487 Chr.19: 54865955-54865955 on Build GRCh38
rs184108639 Chr.19: 54865973-54865973 on Build GRCh38
rs796734062 Chr.19: 54865794-54865794 on Build GRCh38
rs145658103 Chr.19: 54866184-54866184 on Build GRCh38
rs1019572724 Chr.19: 54866146-54866146 on Build GRCh38
rs952154758 Chr.19: 54866179-54866179 on Build GRCh38
rs748902707 Chr.19: 54865826-54865826 on Build GRCh38
rs3745901 Chr.19: 54866144-54866144 on Build GRCh38
rs187054652 Chr.19: 54865958-54865958 on Build GRCh38
rs1030832934 Chr.19: 54866111-54866111 on Build GRCh38
rs199859855 Chr.19: 54865772-54865772 on Build GRCh38
rs773352847 Chr.19: 54865862-54865862 on Build GRCh38
rs779666933 Chr.19: 54865824-54865824 on Build GRCh38
rs374190020 Chr.19: 54865930-54865930 on Build GRCh38
rs796782943 Chr.19: 54865825-54865825 on Build GRCh38
rs769878891 Chr.19: 54865944-54865944 on Build GRCh38
rs746789540 Chr.19: 54865865-54865865 on Build GRCh38
rs766706988 Chr.19: 54865791-54865791 on Build GRCh38
rs376801942 Chr.19: 54865920-54865920 on Build GRCh38
rs113681719 Chr.19: 54865803-54865803 on Build GRCh38
rs896124863 Chr.19: 54866045-54866045 on Build GRCh38
rs201433203 Chr.19: 54866060-54866060 on Build GRCh38
rs563465026 Chr.19: 54865953-54865953 on Build GRCh38
rs767331311 Chr.19: 54865915-54865915 on Build GRCh38
rs567259315 Chr.19: 54865836-54865836 on Build GRCh38
rs796530954 Chr.19: 54865989-54865989 on Build GRCh38
rs760593377 Chr.19: 54865790-54865790 on Build GRCh38
rs2915987 Chr.19: 54865885-54865885 on Build GRCh38
rs3826878 Chr.19: 54865756-54865756 on Build GRCh38
rs200003478 Chr.19: 54865757-54865757 on Build GRCh38
rs764169765 Chr.19: 54865949-54865949 on Build GRCh38
rs746948969 Chr.19: 54865924-54865924 on Build GRCh38
rs3745899 Chr.19: 54866022-54866022 on Build GRCh38
rs141013090 Chr.19: 54865889-54865889 on Build GRCh38
rs3745900 Chr.19: 54866128-54866128 on Build GRCh38
rs2915985 Chr.19: 54865799-54865799 on Build GRCh38
rs1001031830 Chr.19: 54866105-54866105 on Build GRCh38
rs772826353 Chr.19: 54865801-54865801 on Build GRCh38
rs974731742 Chr.19: 54866153-54866153 on Build GRCh38
rs956577097 Chr.19: 54865933-54865933 on Build GRCh38
rs201761696 Chr.19: 54866017-54866017 on Build GRCh38
rs2915988 Chr.19: 54865908-54865908 on Build GRCh38
rs191856811 Chr.19: 54865974-54865974 on Build GRCh38
rs918154447 Chr.19: 54866193-54866193 on Build GRCh38
rs966095366 Chr.19: 54865966-54865966 on Build GRCh38
rs200943273 Chr.19: 54865992-54865992 on Build GRCh38
rs748067982 Chr.19: 54865846-54865846 on Build GRCh38
rs745729222 Chr.19: 54865940-54865940 on Build GRCh38
rs754444084 Chr.19: 54865832-54865832 on Build GRCh38
rs2915986 Chr.19: 54865852-54865852 on Build GRCh38
rs984877937 Chr.19: 54866180-54866180 on Build GRCh38
rs775850447 Chr.19: 54865945-54865945 on Build GRCh38
rs3745898 Chr.19: 54865975-54865975 on Build GRCh38
rs553210593 Chr.19: 54865902-54865902 on Build GRCh38
rs78856890 Chr.19: 54866197-54866197 on Build GRCh38
rs373869955 Chr.19: 54866010-54866010 on Build GRCh38
rs1045150761 Chr.19: 54866070-54866070 on Build GRCh38
rs775413394 Chr.19: 54865882-54865882 on Build GRCh38
rs745475879 Chr.19: 54865755-54865755 on Build GRCh38
rs73618370 Chr.19: 54865770-54865770 on Build GRCh38
rs866190123 Chr.19: 54866005-54866005 on Build GRCh38
rs964319031 Chr.19: 54866152-54866152 on Build GRCh38
rs748825928 Chr.19: 54865759-54865759 on Build GRCh38
rs770809042 Chr.19: 54865929-54865929 on Build GRCh38
rs150217832 Chr.19: 54865893-54865893 on Build GRCh38
rs753209249 Chr.19: 54865805-54865805 on Build GRCh38
rs369288132 Chr.19: 54865818-54865818 on Build GRCh38
rs375258915 Chr.19: 54865809-54865809 on Build GRCh38
rs1039157477 Chr.19: 54865746-54865746 on Build GRCh38
rs759712947 Chr.19: 54865880-54865880 on Build GRCh38
rs562948215 Chr.19: 54866191-54866191 on Build GRCh38
rs201226349 Chr.19: 54866096-54866096 on Build GRCh38
rs72489175 Chr.19: 54865967-54865967 on Build GRCh38
rs773160511 Chr.19: 54865773-54865773 on Build GRCh38
rs763063499 Chr.19: 54865884-54865884 on Build GRCh38
rs768107785 Chr.19: 54865763-54865763 on Build GRCh38
rs765314732 Chr.19: 54865881-54865881 on Build GRCh38
rs372571108 Chr.19: 54865872-54865872 on Build GRCh38
rs908030386 Chr.19: 54866190-54866190 on Build GRCh38
rs940943305 Chr.19: 54866023-54866023 on Build GRCh38
rs796466301 Chr.19: 54865810-54865810 on Build GRCh38
rs79378214 Chr.19: 54865931-54865931 on Build GRCh38
rs3745897 Chr.19: 54865948-54865948 on Build GRCh38

View Full Product Details