Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53210688-53211184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT2
Entrez Gene ID
1376
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
CPT1, CPTASE, IIAE4
UniGene
Hs.713535
Gene Chromosome Location
Chr.1: 53196429-53214197 on Build GRCh38
Transcripts
NM_000098.2 XM_005270484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6063692 c.1489G>T p.G497C Chr.1: 53211163-53211163 on Build GRCh38
2193286 c.1396G>C p.V466L Chr.1: 53211070-53211070 on Build GRCh38
6063693 c.1251C>T p.F417F Chr.1: 53210925-53210925 on Build GRCh38
4577445 c.1438G>A p.G480R Chr.1: 53211112-53211112 on Build GRCh38
6034776 c.1489G>A p.G497S Chr.1: 53211163-53211163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985374504 Chr.1: 53210715-53210715 on Build GRCh38
rs576822710 Chr.1: 53210925-53210925 on Build GRCh38
rs748976396 Chr.1: 53211060-53211060 on Build GRCh38
rs767826599 Chr.1: 53210862-53210862 on Build GRCh38
rs201681645 Chr.1: 53211066-53211066 on Build GRCh38
rs754386565 Chr.1: 53211088-53211088 on Build GRCh38
rs140771069 Chr.1: 53211078-53211078 on Build GRCh38
rs139321501 Chr.1: 53211162-53211162 on Build GRCh38
rs772159469 Chr.1: 53211156-53211170 on Build GRCh38
rs992811256 Chr.1: 53211098-53211098 on Build GRCh38
rs775246158 Chr.1: 53210984-53210984 on Build GRCh38
rs867886479 Chr.1: 53210926-53210926 on Build GRCh38
rs143486080 Chr.1: 53211138-53211138 on Build GRCh38
rs745432304 Chr.1: 53211045-53211045 on Build GRCh38
rs781398541 Chr.1: 53210766-53210766 on Build GRCh38
rs368989271 Chr.1: 53210927-53210927 on Build GRCh38
rs757126340 Chr.1: 53211115-53211118 on Build GRCh38
rs1799821 Chr.1: 53210776-53210776 on Build GRCh38
rs755395180 Chr.1: 53211022-53211022 on Build GRCh38
rs2229291 Chr.1: 53210729-53210729 on Build GRCh38
rs544400632 Chr.1: 53210910-53210910 on Build GRCh38
rs149557870 Chr.1: 53211102-53211102 on Build GRCh38
rs757979320 Chr.1: 53210794-53210795 on Build GRCh38
rs955781566 Chr.1: 53211008-53211008 on Build GRCh38
rs201745292 Chr.1: 53210863-53210863 on Build GRCh38
rs761347926 Chr.1: 53210790-53210790 on Build GRCh38
rs745804213 Chr.1: 53210971-53210971 on Build GRCh38
rs374201361 Chr.1: 53210986-53210986 on Build GRCh38
rs938668766 Chr.1: 53210774-53210774 on Build GRCh38
rs747794860 Chr.1: 53210895-53210895 on Build GRCh38
rs555126720 Chr.1: 53211010-53211010 on Build GRCh38
rs377616144 Chr.1: 53210987-53210987 on Build GRCh38
rs778894948 Chr.1: 53211148-53211149 on Build GRCh38
rs775144404 Chr.1: 53210714-53210714 on Build GRCh38
rs192779168 Chr.1: 53211096-53211096 on Build GRCh38
rs771537809 Chr.1: 53210763-53210763 on Build GRCh38
rs375957043 Chr.1: 53210906-53210906 on Build GRCh38
rs778743524 Chr.1: 53211133-53211133 on Build GRCh38
rs368132822 Chr.1: 53211134-53211134 on Build GRCh38
rs748181550 Chr.1: 53210836-53210836 on Build GRCh38
rs575447822 Chr.1: 53210908-53210908 on Build GRCh38
rs779999942 Chr.1: 53211057-53211057 on Build GRCh38
rs560189631 Chr.1: 53211062-53211062 on Build GRCh38
rs771811692 Chr.1: 53210897-53210897 on Build GRCh38
rs756931329 Chr.1: 53211043-53211043 on Build GRCh38
rs752373512 Chr.1: 53210895-53210896 on Build GRCh38
rs777236101 Chr.1: 53211023-53211023 on Build GRCh38
rs767852112 Chr.1: 53210929-53210929 on Build GRCh38
rs398123153 Chr.1: 53210913-53210914 on Build GRCh38
rs746000404 Chr.1: 53211150-53211153 on Build GRCh38
rs753781632 Chr.1: 53211068-53211080 on Build GRCh38
rs372102993 Chr.1: 53210903-53210903 on Build GRCh38
rs749714778 Chr.1: 53210937-53210937 on Build GRCh38
rs74315295 Chr.1: 53210822-53210822 on Build GRCh38
rs151003641 Chr.1: 53210722-53210722 on Build GRCh38
rs374183980 Chr.1: 53211125-53211125 on Build GRCh38
rs755500312 Chr.1: 53210755-53210755 on Build GRCh38
rs112914907 Chr.1: 53210907-53210907 on Build GRCh38
rs767699548 Chr.1: 53211003-53211003 on Build GRCh38
rs746284903 Chr.1: 53210771-53210771 on Build GRCh38
rs772377335 Chr.1: 53211141-53211141 on Build GRCh38
rs778639977 Chr.1: 53210888-53210888 on Build GRCh38
rs746683786 Chr.1: 53211042-53211042 on Build GRCh38
rs964928101 Chr.1: 53211076-53211076 on Build GRCh38
rs757953266 Chr.1: 53210948-53210948 on Build GRCh38
rs1057517507 Chr.1: 53211033-53211036 on Build GRCh38
rs200399018 Chr.1: 53211071-53211071 on Build GRCh38
rs773474888 Chr.1: 53210883-53210883 on Build GRCh38
rs1057517525 Chr.1: 53211019-53211019 on Build GRCh38
rs74315297 Chr.1: 53211016-53211016 on Build GRCh38
rs777506825 Chr.1: 53210940-53210941 on Build GRCh38
rs770734793 Chr.1: 53211103-53211103 on Build GRCh38
rs865856450 Chr.1: 53210978-53210978 on Build GRCh38
rs750339299 Chr.1: 53210728-53210728 on Build GRCh38
rs780865183 Chr.1: 53211104-53211104 on Build GRCh38
rs760514258 Chr.1: 53210811-53210811 on Build GRCh38
rs753720446 Chr.1: 53210742-53210742 on Build GRCh38
rs397509431 Chr.1: 53210912-53210913 on Build GRCh38
rs147276580 Chr.1: 53211046-53211046 on Build GRCh38
rs376445618 Chr.1: 53210952-53210952 on Build GRCh38
rs374308679 Chr.1: 53211166-53211166 on Build GRCh38
rs515726176 Chr.1: 53210819-53210819 on Build GRCh38
rs758408111 Chr.1: 53210832-53210832 on Build GRCh38
rs771486286 Chr.1: 53210795-53210795 on Build GRCh38
rs749243696 Chr.1: 53211115-53211115 on Build GRCh38
rs143075786 Chr.1: 53211009-53211009 on Build GRCh38
rs751719701 Chr.1: 53210845-53210845 on Build GRCh38
rs746567260 Chr.1: 53210902-53210902 on Build GRCh38
rs144875040 Chr.1: 53210812-53210812 on Build GRCh38
rs201241649 Chr.1: 53211132-53211132 on Build GRCh38
rs373420308 Chr.1: 53210933-53210933 on Build GRCh38
rs146670074 Chr.1: 53210876-53210876 on Build GRCh38
rs772572668 Chr.1: 53210899-53210899 on Build GRCh38
rs74315299 Chr.1: 53211034-53211034 on Build GRCh38
rs761438840 Chr.1: 53210727-53210727 on Build GRCh38
rs772843417 Chr.1: 53210798-53210798 on Build GRCh38
rs756326862 Chr.1: 53211082-53211082 on Build GRCh38
rs749833236 Chr.1: 53210754-53210754 on Build GRCh38
rs548364005 Chr.1: 53211150-53211150 on Build GRCh38
rs996410960 Chr.1: 53210904-53210904 on Build GRCh38
rs187044944 Chr.1: 53210958-53210958 on Build GRCh38
rs761731998 Chr.1: 53211002-53211002 on Build GRCh38
rs766080131 Chr.1: 53210737-53210737 on Build GRCh38
rs746186600 Chr.1: 53210869-53210869 on Build GRCh38
rs767030994 Chr.1: 53211015-53211015 on Build GRCh38
rs762081022 Chr.1: 53211070-53211070 on Build GRCh38
rs763073192 Chr.1: 53210916-53210916 on Build GRCh38
rs776488578 Chr.1: 53210964-53210964 on Build GRCh38
rs199894581 Chr.1: 53210746-53210746 on Build GRCh38
rs369847007 Chr.1: 53210817-53210817 on Build GRCh38
rs35237240 Chr.1: 53211063-53211064 on Build GRCh38
rs148512862 Chr.1: 53210721-53210721 on Build GRCh38
rs752932153 Chr.1: 53210829-53210829 on Build GRCh38
rs1057517515 Chr.1: 53210720-53210721 on Build GRCh38
rs777711968 Chr.1: 53210842-53210842 on Build GRCh38
rs769781310 Chr.1: 53210976-53210976 on Build GRCh38
rs757683330 Chr.1: 53210849-53210849 on Build GRCh38
rs773966429 Chr.1: 53210723-53210723 on Build GRCh38
rs752737849 Chr.1: 53210753-53210753 on Build GRCh38
rs1057517492 Chr.1: 53211019-53211019 on Build GRCh38
rs201008705 Chr.1: 53210921-53210921 on Build GRCh38
rs201508063 Chr.1: 53211112-53211112 on Build GRCh38
rs761130313 Chr.1: 53211090-53211090 on Build GRCh38
rs749895856 Chr.1: 53211110-53211110 on Build GRCh38
rs774644103 Chr.1: 53211001-53211001 on Build GRCh38
rs774168365 Chr.1: 53210784-53210784 on Build GRCh38
rs370157946 Chr.1: 53211111-53211111 on Build GRCh38
rs61731996 Chr.1: 53211151-53211151 on Build GRCh38
rs1024753743 Chr.1: 53210992-53210992 on Build GRCh38
rs770491351 Chr.1: 53210962-53210962 on Build GRCh38

View Full Product Details