Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105361125-105361620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL2
Entrez Gene ID
2274
Gene Name
four and a half LIM domains 2
Gene Aliases
AAG11, DRAL, FHL-2, SLIM-3, SLIM3
UniGene
Hs.443687
Gene Chromosome Location
Chr.2: 105360826-105438773 on Build GRCh38
Transcripts
NM_001039492.2 NM_001318894.1 NM_001318895.1 NM_001318896.1 NM_001318897.1 NM_001318898.1 NM_001318899.1 NM_001450.3 NM_201555.1 NM_201557.3 XM_011510798.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4973104 c.793A>T p.T265S Chr.2: 105361330-105361330 on Build GRCh38
7125329 c.792C>T p.L264L Chr.2: 105361331-105361331 on Build GRCh38
5009938 c.744C>T p.C248C Chr.2: 105361379-105361379 on Build GRCh38
5035208 c.826G>A p.G276R Chr.2: 105361297-105361297 on Build GRCh38
177027 c.725G>A p.R242Q Chr.2: 105361398-105361398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756601082 Chr.2: 105361367-105361367 on Build GRCh38
rs945037275 Chr.2: 105361147-105361147 on Build GRCh38
rs781430211 Chr.2: 105361325-105361325 on Build GRCh38
rs115841332 Chr.2: 105361444-105361444 on Build GRCh38
rs530518744 Chr.2: 105361199-105361199 on Build GRCh38
rs397517961 Chr.2: 105361399-105361399 on Build GRCh38
rs988081108 Chr.2: 105361437-105361437 on Build GRCh38
rs763513286 Chr.2: 105361352-105361352 on Build GRCh38
rs768711688 Chr.2: 105361342-105361342 on Build GRCh38
rs2244182 Chr.2: 105361446-105361446 on Build GRCh38
rs757735612 Chr.2: 105361250-105361250 on Build GRCh38
rs1004883259 Chr.2: 105361340-105361340 on Build GRCh38
rs780524457 Chr.2: 105361373-105361373 on Build GRCh38
rs752238903 Chr.2: 105361246-105361246 on Build GRCh38
rs939760066 Chr.2: 105361182-105361182 on Build GRCh38
rs368824523 Chr.2: 105361253-105361253 on Build GRCh38
rs749472445 Chr.2: 105361299-105361299 on Build GRCh38
rs924963147 Chr.2: 105361507-105361507 on Build GRCh38
rs752907057 Chr.2: 105361481-105361481 on Build GRCh38
rs760538519 Chr.2: 105361313-105361313 on Build GRCh38
rs568647217 Chr.2: 105361434-105361434 on Build GRCh38
rs777732229 Chr.2: 105361403-105361403 on Build GRCh38
rs776246738 Chr.2: 105361460-105361460 on Build GRCh38
rs963928826 Chr.2: 105361368-105361368 on Build GRCh38
rs569400431 Chr.2: 105361543-105361543 on Build GRCh38
rs779103780 Chr.2: 105361302-105361302 on Build GRCh38
rs754242864 Chr.2: 105361467-105361467 on Build GRCh38
rs730880110 Chr.2: 105361300-105361300 on Build GRCh38
rs758574596 Chr.2: 105361483-105361483 on Build GRCh38
rs765701012 Chr.2: 105361473-105361473 on Build GRCh38
rs903741979 Chr.2: 105361183-105361183 on Build GRCh38
rs769888361 Chr.2: 105361384-105361384 on Build GRCh38
rs759380486 Chr.2: 105361443-105361443 on Build GRCh38
rs750884671 Chr.2: 105361363-105361363 on Build GRCh38
rs778092790 Chr.2: 105361170-105361170 on Build GRCh38
rs11537577 Chr.2: 105361149-105361149 on Build GRCh38
rs748104388 Chr.2: 105361492-105361492 on Build GRCh38
rs770807726 Chr.2: 105361424-105361424 on Build GRCh38
rs3087523 Chr.2: 105361319-105361319 on Build GRCh38
rs781750373 Chr.2: 105361252-105361252 on Build GRCh38
rs145070796 Chr.2: 105361303-105361303 on Build GRCh38
rs775963548 Chr.2: 105361234-105361235 on Build GRCh38
rs11124029 Chr.2: 105361304-105361304 on Build GRCh38
rs755463731 Chr.2: 105361472-105361472 on Build GRCh38
rs780294130 Chr.2: 105361295-105361295 on Build GRCh38
rs773266937 Chr.2: 105361308-105361308 on Build GRCh38
rs764915908 Chr.2: 105361329-105361329 on Build GRCh38
rs879204828 Chr.2: 105361167-105361167 on Build GRCh38
rs188279857 Chr.2: 105361398-105361398 on Build GRCh38
rs1000798237 Chr.2: 105361233-105361233 on Build GRCh38
rs756430758 Chr.2: 105361267-105361267 on Build GRCh38
rs767041110 Chr.2: 105361464-105361464 on Build GRCh38
rs564693249 Chr.2: 105361549-105361549 on Build GRCh38
rs762522824 Chr.2: 105361455-105361455 on Build GRCh38
rs770706113 Chr.2: 105361318-105361318 on Build GRCh38
rs763610637 Chr.2: 105361459-105361459 on Build GRCh38
rs866614289 Chr.2: 105361336-105361336 on Build GRCh38
rs199691250 Chr.2: 105361270-105361270 on Build GRCh38
rs371759004 Chr.2: 105361341-105361341 on Build GRCh38
rs762587215 Chr.2: 105361345-105361345 on Build GRCh38
rs869025433 Chr.2: 105361408-105361408 on Build GRCh38
rs761212075 Chr.2: 105361461-105361461 on Build GRCh38
rs772078127 Chr.2: 105361495-105361495 on Build GRCh38
rs755230829 Chr.2: 105361385-105361385 on Build GRCh38
rs746919278 Chr.2: 105361421-105361421 on Build GRCh38

View Full Product Details