Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105363203-105363701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL2
Entrez Gene ID
2274
Gene Name
four and a half LIM domains 2
Gene Aliases
AAG11, DRAL, FHL-2, SLIM-3, SLIM3
UniGene
Hs.443687
Gene Chromosome Location
Chr.2: 105360826-105438773 on Build GRCh38
Transcripts
NM_001039492.2 NM_001318894.1 NM_001318895.1 NM_001318896.1 NM_001318897.1 NM_001318898.1 NM_001318899.1 NM_001450.3 NM_201555.1 NM_201557.3 XM_011510798.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859811 c.572G>T p.C191F Chr.2: 105363401-105363401 on Build GRCh38
3797813 c.609C>T p.R203R Chr.2: 105363364-105363364 on Build GRCh38
1004772 c.595C>T p.R199C Chr.2: 105363378-105363378 on Build GRCh38
1481807 c.584T>A p.L195Q Chr.2: 105363389-105363389 on Build GRCh38
4083953 c.607C>T p.R203C Chr.2: 105363366-105363366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751652242 Chr.2: 105363341-105363341 on Build GRCh38
rs945828540 Chr.2: 105363666-105363666 on Build GRCh38
rs1018740778 Chr.2: 105363460-105363460 on Build GRCh38
rs1009986435 Chr.2: 105363440-105363440 on Build GRCh38
rs1050970163 Chr.2: 105363366-105363366 on Build GRCh38
rs781258347 Chr.2: 105363352-105363352 on Build GRCh38
rs371939143 Chr.2: 105363253-105363253 on Build GRCh38
rs373259092 Chr.2: 105363378-105363378 on Build GRCh38
rs727504674 Chr.2: 105363461-105363461 on Build GRCh38
rs142473444 Chr.2: 105363529-105363529 on Build GRCh38
rs763291334 Chr.2: 105363444-105363444 on Build GRCh38
rs766470646 Chr.2: 105363550-105363550 on Build GRCh38
rs986329672 Chr.2: 105363456-105363456 on Build GRCh38
rs753039196 Chr.2: 105363319-105363319 on Build GRCh38
rs773944178 Chr.2: 105363394-105363394 on Build GRCh38
rs113236359 Chr.2: 105363579-105363579 on Build GRCh38
rs727504994 Chr.2: 105363302-105363302 on Build GRCh38
rs778784485 Chr.2: 105363499-105363499 on Build GRCh38
rs753993422 Chr.2: 105363565-105363565 on Build GRCh38
rs113188481 Chr.2: 105363443-105363443 on Build GRCh38
rs918582461 Chr.2: 105363294-105363294 on Build GRCh38
rs942612730 Chr.2: 105363583-105363583 on Build GRCh38
rs765888176 Chr.2: 105363441-105363441 on Build GRCh38
rs35564285 Chr.2: 105363610-105363611 on Build GRCh38
rs2278502 Chr.2: 105363273-105363273 on Build GRCh38
rs755085628 Chr.2: 105363625-105363625 on Build GRCh38
rs572884587 Chr.2: 105363545-105363545 on Build GRCh38
rs771650916 Chr.2: 105363406-105363406 on Build GRCh38
rs534279240 Chr.2: 105363285-105363285 on Build GRCh38
rs765651831 Chr.2: 105363318-105363318 on Build GRCh38
rs555828631 Chr.2: 105363262-105363262 on Build GRCh38
rs369765217 Chr.2: 105363286-105363286 on Build GRCh38
rs986732726 Chr.2: 105363578-105363578 on Build GRCh38
rs906250700 Chr.2: 105363363-105363363 on Build GRCh38
rs779913648 Chr.2: 105363481-105363481 on Build GRCh38
rs555422474 Chr.2: 105363612-105363612 on Build GRCh38
rs200116659 Chr.2: 105363450-105363450 on Build GRCh38
rs144442113 Chr.2: 105363452-105363452 on Build GRCh38
rs570903247 Chr.2: 105363414-105363414 on Build GRCh38
rs918629814 Chr.2: 105363549-105363549 on Build GRCh38
rs376689560 Chr.2: 105363383-105363383 on Build GRCh38
rs756076159 Chr.2: 105363377-105363377 on Build GRCh38
rs764275604 Chr.2: 105363332-105363332 on Build GRCh38
rs750695509 Chr.2: 105363482-105363482 on Build GRCh38
rs539301452 Chr.2: 105363466-105363466 on Build GRCh38
rs567289417 Chr.2: 105363304-105363304 on Build GRCh38
rs535364376 Chr.2: 105363305-105363305 on Build GRCh38
rs916111449 Chr.2: 105363573-105363573 on Build GRCh38
rs143604095 Chr.2: 105363380-105363380 on Build GRCh38
rs750539692 Chr.2: 105363464-105363464 on Build GRCh38
rs777181609 Chr.2: 105363361-105363361 on Build GRCh38
rs747844229 Chr.2: 105363397-105363397 on Build GRCh38
rs754663221 Chr.2: 105363484-105363484 on Build GRCh38
rs745753003 Chr.2: 105363364-105363364 on Build GRCh38
rs137869171 Chr.2: 105363295-105363295 on Build GRCh38
rs907390644 Chr.2: 105363640-105363640 on Build GRCh38
rs768230517 Chr.2: 105363278-105363278 on Build GRCh38
rs575300538 Chr.2: 105363255-105363255 on Build GRCh38
rs777871895 Chr.2: 105363250-105363250 on Build GRCh38
rs892899914 Chr.2: 105363415-105363415 on Build GRCh38
rs897607536 Chr.2: 105363676-105363676 on Build GRCh38
rs769517060 Chr.2: 105363271-105363271 on Build GRCh38
rs990198824 Chr.2: 105363572-105363572 on Build GRCh38
rs944795008 Chr.2: 105363233-105363233 on Build GRCh38
rs747101940 Chr.2: 105363252-105363252 on Build GRCh38
rs1036101854 Chr.2: 105363675-105363675 on Build GRCh38
rs889685727 Chr.2: 105363389-105363389 on Build GRCh38
rs758618755 Chr.2: 105363329-105363329 on Build GRCh38
rs1045935903 Chr.2: 105363322-105363322 on Build GRCh38
rs940196138 Chr.2: 105363641-105363641 on Build GRCh38

View Full Product Details