Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112528272-112528802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTL
Entrez Gene ID
150465
Gene Name
tubulin tyrosine ligase
Gene Aliases
-
UniGene
Hs.358997
Gene Chromosome Location
Chr.2: 112482127-112532646 on Build GRCh38
Transcripts
NM_153712.4 XM_005263599.3 XM_011510665.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5394146 c.1054G>A p.D352N Chr.2: 112528715-112528715 on Build GRCh38
5354263 c.1058T>C p.I353T Chr.2: 112528719-112528719 on Build GRCh38
4084238 c.1050C>T p.I350I Chr.2: 112528711-112528711 on Build GRCh38
4084237 c.1028A>G p.Y343C Chr.2: 112528689-112528689 on Build GRCh38
3039689 c.1118C>T p.A373V Chr.2: 112528779-112528779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904403834 Chr.2: 112528708-112528708 on Build GRCh38
rs377061169 Chr.2: 112528697-112528697 on Build GRCh38
rs375130874 Chr.2: 112528634-112528634 on Build GRCh38
rs944024753 Chr.2: 112528542-112528542 on Build GRCh38
rs778183817 Chr.2: 112528632-112528632 on Build GRCh38
rs745431139 Chr.2: 112528443-112528443 on Build GRCh38
rs968432145 Chr.2: 112528400-112528400 on Build GRCh38
rs549319256 Chr.2: 112528763-112528763 on Build GRCh38
rs200640449 Chr.2: 112528722-112528722 on Build GRCh38
rs745576016 Chr.2: 112528630-112528630 on Build GRCh38
rs1039694498 Chr.2: 112528576-112528576 on Build GRCh38
rs373897008 Chr.2: 112528687-112528687 on Build GRCh38
rs774763102 Chr.2: 112528607-112528607 on Build GRCh38
rs541808726 Chr.2: 112528369-112528369 on Build GRCh38
rs1004425909 Chr.2: 112528621-112528621 on Build GRCh38
rs112280013 Chr.2: 112528337-112528337 on Build GRCh38
rs371537879 Chr.2: 112528712-112528712 on Build GRCh38
rs931498769 Chr.2: 112528490-112528490 on Build GRCh38
rs750577590 Chr.2: 112528665-112528665 on Build GRCh38
rs761973593 Chr.2: 112528655-112528655 on Build GRCh38
rs181785370 Chr.2: 112528618-112528618 on Build GRCh38
rs768363386 Chr.2: 112528521-112528522 on Build GRCh38
rs775306697 Chr.2: 112528636-112528636 on Build GRCh38
rs544941620 Chr.2: 112528558-112528558 on Build GRCh38
rs970981752 Chr.2: 112528425-112528425 on Build GRCh38
rs758604499 Chr.2: 112528674-112528674 on Build GRCh38
rs377432688 Chr.2: 112528751-112528751 on Build GRCh38
rs6732299 Chr.2: 112528550-112528550 on Build GRCh38
rs768633270 Chr.2: 112528748-112528748 on Build GRCh38
rs6748148 Chr.2: 112528771-112528771 on Build GRCh38
rs1035793887 Chr.2: 112528319-112528319 on Build GRCh38
rs776692261 Chr.2: 112528749-112528749 on Build GRCh38
rs377186855 Chr.2: 112528658-112528658 on Build GRCh38
rs201658370 Chr.2: 112528723-112528723 on Build GRCh38
rs751889682 Chr.2: 112528696-112528696 on Build GRCh38
rs762952387 Chr.2: 112528774-112528774 on Build GRCh38
rs899954638 Chr.2: 112528587-112528587 on Build GRCh38
rs369676615 Chr.2: 112528648-112528648 on Build GRCh38
rs781662408 Chr.2: 112528699-112528699 on Build GRCh38
rs980046168 Chr.2: 112528416-112528416 on Build GRCh38
rs199740251 Chr.2: 112528740-112528740 on Build GRCh38
rs972967320 Chr.2: 112528426-112528426 on Build GRCh38
rs139582155 Chr.2: 112528295-112528296 on Build GRCh38
rs115916418 Chr.2: 112528327-112528327 on Build GRCh38
rs745962835 Chr.2: 112528436-112528437 on Build GRCh38
rs762249314 Chr.2: 112528617-112528617 on Build GRCh38
rs546916808 Chr.2: 112528646-112528646 on Build GRCh38
rs760560694 Chr.2: 112528637-112528637 on Build GRCh38
rs995602172 Chr.2: 112528589-112528589 on Build GRCh38
rs746798177 Chr.2: 112528741-112528741 on Build GRCh38
rs992510939 Chr.2: 112528394-112528394 on Build GRCh38
rs748710136 Chr.2: 112528702-112528702 on Build GRCh38
rs375376633 Chr.2: 112528768-112528768 on Build GRCh38
rs12471089 Chr.2: 112528442-112528442 on Build GRCh38
rs531193120 Chr.2: 112528468-112528468 on Build GRCh38
rs553581163 Chr.2: 112528711-112528711 on Build GRCh38
rs370528673 Chr.2: 112528765-112528765 on Build GRCh38
rs912797087 Chr.2: 112528524-112528524 on Build GRCh38
rs1012569600 Chr.2: 112528649-112528649 on Build GRCh38
rs1048729052 Chr.2: 112528616-112528616 on Build GRCh38
rs67178025 Chr.2: 112528446-112528446 on Build GRCh38
rs1041598942 Chr.2: 112528705-112528705 on Build GRCh38
rs925454447 Chr.2: 112528423-112528423 on Build GRCh38
rs904352885 Chr.2: 112528300-112528302 on Build GRCh38
rs959716147 Chr.2: 112528322-112528322 on Build GRCh38
rs369214464 Chr.2: 112528609-112528609 on Build GRCh38
rs745392240 Chr.2: 112528718-112528718 on Build GRCh38
rs572895001 Chr.2: 112528365-112528365 on Build GRCh38

View Full Product Details