Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29173425-29173951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2J2
Entrez Gene ID
26707
Gene Name
olfactory receptor family 2 subfamily J member 2
Gene Aliases
OR6-19, OR6-8, OR6.3.8, ORL684, dJ80I19.4, hs6M1-6
UniGene
Hs.163518
Gene Chromosome Location
Chr.6: 29173534-29174574 on Build GRCh38
Transcripts
NM_030905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1663234 c.35T>A p.F12Y Chr.6: 29173670-29173670 on Build GRCh38
4006234 c.241C>T p.Q81* Chr.6: 29173876-29173876 on Build GRCh38
1567878 c.190C>A p.L64I Chr.6: 29173825-29173825 on Build GRCh38
6500969 c.227C>A p.T76N Chr.6: 29173862-29173862 on Build GRCh38
4155151 c.90G>T p.V30V Chr.6: 29173725-29173725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200829397 Chr.6: 29173661-29173661 on Build GRCh38
rs575458407 Chr.6: 29173916-29173916 on Build GRCh38
rs370723822 Chr.6: 29173662-29173662 on Build GRCh38
rs986904037 Chr.6: 29173898-29173898 on Build GRCh38
rs530829408 Chr.6: 29173469-29173469 on Build GRCh38
rs757074784 Chr.6: 29173633-29173633 on Build GRCh38
rs772924943 Chr.6: 29173908-29173908 on Build GRCh38
rs369003322 Chr.6: 29173894-29173894 on Build GRCh38
rs374520427 Chr.6: 29173754-29173754 on Build GRCh38
rs769592780 Chr.6: 29173592-29173593 on Build GRCh38
rs1032667016 Chr.6: 29173798-29173798 on Build GRCh38
rs184607370 Chr.6: 29173909-29173909 on Build GRCh38
rs201438710 Chr.6: 29173645-29173645 on Build GRCh38
rs763225482 Chr.6: 29173901-29173901 on Build GRCh38
rs758040981 Chr.6: 29173874-29173874 on Build GRCh38
rs752387719 Chr.6: 29173871-29173871 on Build GRCh38
rs748416076 Chr.6: 29173764-29173764 on Build GRCh38
rs367717828 Chr.6: 29173723-29173723 on Build GRCh38
rs267600934 Chr.6: 29173759-29173759 on Build GRCh38
rs375318764 Chr.6: 29173678-29173678 on Build GRCh38
rs773776399 Chr.6: 29173776-29173776 on Build GRCh38
rs556940268 Chr.6: 29173645-29173645 on Build GRCh38
rs750278136 Chr.6: 29173839-29173839 on Build GRCh38
rs775603734 Chr.6: 29173758-29173758 on Build GRCh38
rs868815525 Chr.6: 29173708-29173708 on Build GRCh38
rs189491464 Chr.6: 29173917-29173917 on Build GRCh38
rs141015719 Chr.6: 29173789-29173789 on Build GRCh38
rs751383956 Chr.6: 29173622-29173622 on Build GRCh38
rs759692008 Chr.6: 29173803-29173803 on Build GRCh38
rs192953819 Chr.6: 29173528-29173528 on Build GRCh38
rs778222704 Chr.6: 29173877-29173877 on Build GRCh38
rs148108002 Chr.6: 29173570-29173570 on Build GRCh38
rs759770156 Chr.6: 29173705-29173705 on Build GRCh38
rs755988197 Chr.6: 29173852-29173852 on Build GRCh38
rs74863731 Chr.6: 29173724-29173724 on Build GRCh38
rs9257565 Chr.6: 29173613-29173613 on Build GRCh38
rs138969453 Chr.6: 29173895-29173895 on Build GRCh38
rs775243759 Chr.6: 29173607-29173610 on Build GRCh38
rs537704908 Chr.6: 29173679-29173679 on Build GRCh38
rs768477116 Chr.6: 29173678-29173678 on Build GRCh38
rs3116855 Chr.6: 29173855-29173855 on Build GRCh38
rs41317595 Chr.6: 29173477-29173477 on Build GRCh38
rs374674840 Chr.6: 29173703-29173703 on Build GRCh38
rs1024011658 Chr.6: 29173610-29173610 on Build GRCh38
rs958723054 Chr.6: 29173583-29173583 on Build GRCh38
rs777412699 Chr.6: 29173619-29173619 on Build GRCh38
rs377394296 Chr.6: 29173730-29173730 on Build GRCh38
rs995090761 Chr.6: 29173537-29173537 on Build GRCh38
rs754287734 Chr.6: 29173704-29173704 on Build GRCh38
rs998536574 Chr.6: 29173791-29173791 on Build GRCh38
rs370775103 Chr.6: 29173788-29173788 on Build GRCh38
rs376610216 Chr.6: 29173815-29173815 on Build GRCh38
rs376476162 Chr.6: 29173699-29173699 on Build GRCh38
rs1036237267 Chr.6: 29173511-29173511 on Build GRCh38
rs778338352 Chr.6: 29173870-29173870 on Build GRCh38
rs371659935 Chr.6: 29173862-29173862 on Build GRCh38
rs373372174 Chr.6: 29173587-29173587 on Build GRCh38
rs748554260 Chr.6: 29173685-29173685 on Build GRCh38
rs188320028 Chr.6: 29173470-29173470 on Build GRCh38
rs774525822 Chr.6: 29173682-29173682 on Build GRCh38
rs766312489 Chr.6: 29173853-29173853 on Build GRCh38
rs746409306 Chr.6: 29173887-29173887 on Build GRCh38
rs767341980 Chr.6: 29173837-29173837 on Build GRCh38
rs781219101 Chr.6: 29173644-29173644 on Build GRCh38
rs763126521 Chr.6: 29173833-29173833 on Build GRCh38
rs750331547 Chr.6: 29173748-29173748 on Build GRCh38
rs748446669 Chr.6: 29173512-29173512 on Build GRCh38
rs765500589 Chr.6: 29173719-29173719 on Build GRCh38
rs755601407 Chr.6: 29173652-29173652 on Build GRCh38
rs774216085 Chr.6: 29173769-29173771 on Build GRCh38
rs142201939 Chr.6: 29173902-29173902 on Build GRCh38
rs566154666 Chr.6: 29173582-29173582 on Build GRCh38
rs1024063695 Chr.6: 29173779-29173779 on Build GRCh38
rs780725104 Chr.6: 29173485-29173485 on Build GRCh38
rs199886529 Chr.6: 29173702-29173702 on Build GRCh38
rs754079241 Chr.6: 29173603-29173603 on Build GRCh38
rs771524317 Chr.6: 29173882-29173882 on Build GRCh38
rs975690687 Chr.6: 29173827-29173827 on Build GRCh38
rs755187820 Chr.6: 29173760-29173760 on Build GRCh38
rs767262944 Chr.6: 29173742-29173742 on Build GRCh38
rs754524360 Chr.6: 29173757-29173757 on Build GRCh38
rs772463705 Chr.6: 29173765-29173765 on Build GRCh38
rs371177388 Chr.6: 29173752-29173752 on Build GRCh38
rs781770046 Chr.6: 29173884-29173884 on Build GRCh38
rs199509944 Chr.6: 29173749-29173749 on Build GRCh38
rs772267947 Chr.6: 29173727-29173727 on Build GRCh38
rs761648641 Chr.6: 29173813-29173813 on Build GRCh38
rs374676477 Chr.6: 29173869-29173869 on Build GRCh38
rs752158529 Chr.6: 29173923-29173923 on Build GRCh38

View Full Product Details