Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30061972-30062464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNRD1
Entrez Gene ID
30834
Gene Name
zinc ribbon domain containing 1
Gene Aliases
HTEX-6, HTEX6, Rpa12, TCTEX6, TEX6, ZR14, hZR14, tctex-6
UniGene
Hs.57813
Gene Chromosome Location
Chr.6: 30061240-30064909 on Build GRCh38
Transcripts
NM_001278785.1 NM_001278786.1 NM_014596.5 NM_170783.3 NR_103864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6351546 c.303G>A p.Q101Q Chr.6: 30062280-30062280 on Build GRCh38
3861343 c.257G>A p.R86H Chr.6: 30062234-30062234 on Build GRCh38
5627370 c.321A>G p.E107E Chr.6: 30062298-30062298 on Build GRCh38
4825220 c.316G>T p.D106Y Chr.6: 30062293-30062293 on Build GRCh38
4957303 c.316G>A p.D106N Chr.6: 30062293-30062293 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs9261270 Chr.6: 30062358-30062358 on Build GRCh38
rs766686472 Chr.6: 30062002-30062002 on Build GRCh38
rs748500484 Chr.6: 30062289-30062289 on Build GRCh38
rs376269985 Chr.6: 30062282-30062282 on Build GRCh38
rs368504853 Chr.6: 30062420-30062420 on Build GRCh38
rs776774508 Chr.6: 30062222-30062222 on Build GRCh38
rs373543360 Chr.6: 30062184-30062184 on Build GRCh38
rs767431311 Chr.6: 30062251-30062251 on Build GRCh38
rs902430005 Chr.6: 30062268-30062268 on Build GRCh38
rs781135182 Chr.6: 30062182-30062182 on Build GRCh38
rs1036858587 Chr.6: 30062044-30062044 on Build GRCh38
rs149510733 Chr.6: 30062427-30062427 on Build GRCh38
rs779844597 Chr.6: 30062069-30062071 on Build GRCh38
rs201936252 Chr.6: 30062233-30062233 on Build GRCh38
rs995991746 Chr.6: 30062328-30062328 on Build GRCh38
rs201371274 Chr.6: 30062321-30062321 on Build GRCh38
rs527495192 Chr.6: 30062004-30062004 on Build GRCh38
rs772677373 Chr.6: 30062206-30062206 on Build GRCh38
rs748576936 Chr.6: 30062202-30062202 on Build GRCh38
rs931121150 Chr.6: 30062105-30062105 on Build GRCh38
rs749450660 Chr.6: 30062369-30062372 on Build GRCh38
rs939122720 Chr.6: 30062172-30062172 on Build GRCh38
rs928767181 Chr.6: 30062032-30062032 on Build GRCh38
rs771201250 Chr.6: 30062217-30062217 on Build GRCh38
rs9261269 Chr.6: 30062337-30062337 on Build GRCh38
rs773904814 Chr.6: 30062207-30062207 on Build GRCh38
rs755683238 Chr.6: 30062186-30062186 on Build GRCh38
rs547246315 Chr.6: 30062189-30062189 on Build GRCh38
rs761594044 Chr.6: 30062377-30062377 on Build GRCh38
rs761649008 Chr.6: 30062246-30062246 on Build GRCh38
rs778669782 Chr.6: 30062286-30062286 on Build GRCh38
rs918427048 Chr.6: 30062054-30062054 on Build GRCh38
rs9261271 Chr.6: 30062412-30062412 on Build GRCh38
rs1043781069 Chr.6: 30062243-30062243 on Build GRCh38
rs894884605 Chr.6: 30062211-30062211 on Build GRCh38
rs757134212 Chr.6: 30062174-30062174 on Build GRCh38
rs777135024 Chr.6: 30062317-30062317 on Build GRCh38
rs769872721 Chr.6: 30062375-30062375 on Build GRCh38
rs756053671 Chr.6: 30062271-30062271 on Build GRCh38
rs763726809 Chr.6: 30062015-30062015 on Build GRCh38
rs775842288 Chr.6: 30062376-30062376 on Build GRCh38
rs1049626258 Chr.6: 30062155-30062155 on Build GRCh38
rs1011716239 Chr.6: 30062259-30062259 on Build GRCh38
rs952107670 Chr.6: 30062353-30062353 on Build GRCh38
rs762803270 Chr.6: 30062008-30062008 on Build GRCh38
rs114533592 Chr.6: 30062242-30062242 on Build GRCh38
rs751380242 Chr.6: 30062041-30062041 on Build GRCh38
rs865974875 Chr.6: 30062152-30062152 on Build GRCh38
rs747069882 Chr.6: 30062213-30062213 on Build GRCh38
rs10745 Chr.6: 30062280-30062280 on Build GRCh38
rs778419367 Chr.6: 30062334-30062334 on Build GRCh38
rs754548771 Chr.6: 30062284-30062284 on Build GRCh38
rs766171005 Chr.6: 30062003-30062003 on Build GRCh38
rs36144515 Chr.6: 30062440-30062440 on Build GRCh38
rs948823722 Chr.6: 30062361-30062361 on Build GRCh38
rs771478740 Chr.6: 30062341-30062341 on Build GRCh38
rs768370574 Chr.6: 30062195-30062195 on Build GRCh38
rs776520229 Chr.6: 30062354-30062354 on Build GRCh38
rs931169153 Chr.6: 30062344-30062344 on Build GRCh38
rs948754387 Chr.6: 30062183-30062183 on Build GRCh38
rs1015306050 Chr.6: 30062407-30062407 on Build GRCh38
rs570751747 Chr.6: 30062198-30062198 on Build GRCh38

View Full Product Details