Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109465860-109466369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICAL1
Entrez Gene ID
64780
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Gene Aliases
MICAL, MICAL-1, NICAL
UniGene
Hs.33476
Gene Chromosome Location
Chr.6: 109444062-109465968 on Build GRCh38
Transcripts
NM_001286613.1 NM_014797.2

Gene Symbol
ZBTB24
Entrez Gene ID
9841
Gene Name
zinc finger and BTB domain containing 24
Gene Aliases
BIF1, ICF2, PATZ2, ZNF450
UniGene
Hs.409876
Gene Chromosome Location
Chr.6: 109462516-109483237 on Build GRCh38
Transcripts
NM_001286613.1 NM_014797.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
739712 c.2081A>T p.Q694L Chr.6: 109465864-109465864 on Build GRCh38
739711 c.1823C>G p.S608* Chr.6: 109466122-109466122 on Build GRCh38
3374011 c.1627C>G p.Q543E Chr.6: 109466318-109466318 on Build GRCh38
1071995 c.1641G>A p.S547S Chr.6: 109466304-109466304 on Build GRCh38
1166285 c.2037G>A p.P679P Chr.6: 109465908-109465908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145894539 Chr.6: 109466052-109466052 on Build GRCh38
rs963257288 Chr.6: 109466104-109466104 on Build GRCh38
rs777970263 Chr.6: 109465959-109465959 on Build GRCh38
rs761490454 Chr.6: 109465916-109465916 on Build GRCh38
rs772738232 Chr.6: 109466008-109466008 on Build GRCh38
rs938946665 Chr.6: 109465937-109465937 on Build GRCh38
rs768190156 Chr.6: 109465992-109465992 on Build GRCh38
rs750694717 Chr.6: 109466174-109466174 on Build GRCh38
rs775547572 Chr.6: 109466245-109466245 on Build GRCh38
rs769178596 Chr.6: 109466083-109466083 on Build GRCh38
rs568879322 Chr.6: 109465891-109465891 on Build GRCh38
rs766496267 Chr.6: 109465949-109465949 on Build GRCh38
rs142189675 Chr.6: 109465908-109465908 on Build GRCh38
rs765559975 Chr.6: 109466034-109466034 on Build GRCh38
rs753127074 Chr.6: 109466211-109466211 on Build GRCh38
rs777180797 Chr.6: 109466214-109466214 on Build GRCh38
rs751242825 Chr.6: 109466013-109466013 on Build GRCh38
rs532563272 Chr.6: 109466304-109466304 on Build GRCh38
rs763532224 Chr.6: 109465960-109465960 on Build GRCh38
rs755334302 Chr.6: 109466199-109466199 on Build GRCh38
rs139726759 Chr.6: 109466230-109466230 on Build GRCh38
rs770667826 Chr.6: 109466096-109466096 on Build GRCh38
rs747489610 Chr.6: 109465989-109465989 on Build GRCh38
rs554040213 Chr.6: 109466184-109466184 on Build GRCh38
rs554507506 Chr.6: 109465933-109465933 on Build GRCh38
rs1056081342 Chr.6: 109465945-109465945 on Build GRCh38
rs61731736 Chr.6: 109466273-109466273 on Build GRCh38
rs757215770 Chr.6: 109466023-109466023 on Build GRCh38
rs755693994 Chr.6: 109466212-109466212 on Build GRCh38
rs552304421 Chr.6: 109466313-109466313 on Build GRCh38
rs200378080 Chr.6: 109465895-109465895 on Build GRCh38
rs755495654 Chr.6: 109466068-109466068 on Build GRCh38
rs560810060 Chr.6: 109466327-109466327 on Build GRCh38
rs748919456 Chr.6: 109466091-109466091 on Build GRCh38
rs894715576 Chr.6: 109465979-109465979 on Build GRCh38
rs751534136 Chr.6: 109465956-109465956 on Build GRCh38
rs746246532 Chr.6: 109465980-109465980 on Build GRCh38
rs202007110 Chr.6: 109466282-109466282 on Build GRCh38
rs775909074 Chr.6: 109465897-109465897 on Build GRCh38
rs748759921 Chr.6: 109466217-109466217 on Build GRCh38
rs751770502 Chr.6: 109466274-109466274 on Build GRCh38
rs765948250 Chr.6: 109465890-109465890 on Build GRCh38
rs762867915 Chr.6: 109466010-109466010 on Build GRCh38
rs778764491 Chr.6: 109466331-109466331 on Build GRCh38
rs750596422 Chr.6: 109466036-109466036 on Build GRCh38
rs1025951778 Chr.6: 109466259-109466259 on Build GRCh38
rs142830104 Chr.6: 109466257-109466257 on Build GRCh38
rs574657385 Chr.6: 109465961-109465961 on Build GRCh38
rs775492598 Chr.6: 109465904-109465904 on Build GRCh38
rs745492894 Chr.6: 109466343-109466343 on Build GRCh38
rs1030593144 Chr.6: 109466078-109466078 on Build GRCh38
rs373326176 Chr.6: 109465889-109465889 on Build GRCh38
rs780595455 Chr.6: 109465987-109465987 on Build GRCh38
rs765207865 Chr.6: 109466158-109466158 on Build GRCh38
rs762591757 Chr.6: 109465944-109465944 on Build GRCh38
rs753404186 Chr.6: 109466320-109466320 on Build GRCh38
rs774471077 Chr.6: 109466014-109466014 on Build GRCh38
rs554644990 Chr.6: 109466126-109466126 on Build GRCh38
rs760497855 Chr.6: 109466254-109466254 on Build GRCh38
rs376297475 Chr.6: 109465958-109465958 on Build GRCh38
rs1056358692 Chr.6: 109466246-109466246 on Build GRCh38
rs139491074 Chr.6: 109466195-109466195 on Build GRCh38
rs778943402 Chr.6: 109465971-109465971 on Build GRCh38
rs963456092 Chr.6: 109466324-109466324 on Build GRCh38
rs2232449 Chr.6: 109466244-109466244 on Build GRCh38
rs1047223806 Chr.6: 109465898-109465898 on Build GRCh38
rs772969544 Chr.6: 109466089-109466089 on Build GRCh38
rs769494892 Chr.6: 109466000-109466000 on Build GRCh38
rs35366835 Chr.6: 109465907-109465907 on Build GRCh38
rs983462746 Chr.6: 109465900-109465900 on Build GRCh38
rs144754453 Chr.6: 109466243-109466243 on Build GRCh38
rs781012540 Chr.6: 109466082-109466082 on Build GRCh38
rs778202357 Chr.6: 109465883-109465883 on Build GRCh38
rs371830905 Chr.6: 109466157-109466157 on Build GRCh38
rs758756991 Chr.6: 109465984-109465984 on Build GRCh38
rs776148069 Chr.6: 109465996-109465996 on Build GRCh38
rs138493459 Chr.6: 109466109-109466109 on Build GRCh38
rs767566292 Chr.6: 109466134-109466134 on Build GRCh38
rs370775216 Chr.6: 109466048-109466048 on Build GRCh38
rs775282835 Chr.6: 109466142-109466142 on Build GRCh38
rs759946599 Chr.6: 109466275-109466275 on Build GRCh38
rs776310741 Chr.6: 109465909-109465909 on Build GRCh38
rs369925142 Chr.6: 109466262-109466262 on Build GRCh38
rs759292188 Chr.6: 109466026-109466027 on Build GRCh38
rs149379400 Chr.6: 109466238-109466238 on Build GRCh38
rs757524152 Chr.6: 109465965-109465965 on Build GRCh38
rs916775744 Chr.6: 109466004-109466004 on Build GRCh38
rs373951478 Chr.6: 109465893-109465893 on Build GRCh38
rs546872315 Chr.6: 109466280-109466280 on Build GRCh38
rs201984127 Chr.6: 109466095-109466095 on Build GRCh38
rs767515335 Chr.6: 109466022-109466022 on Build GRCh38
rs371855790 Chr.6: 109466267-109466267 on Build GRCh38
rs747559831 Chr.6: 109465999-109465999 on Build GRCh38
rs758810180 Chr.6: 109466045-109466045 on Build GRCh38
rs754717708 Chr.6: 109466020-109466020 on Build GRCh38
rs764753566 Chr.6: 109465922-109465922 on Build GRCh38
rs954596203 Chr.6: 109466079-109466079 on Build GRCh38
rs368528986 Chr.6: 109466088-109466088 on Build GRCh38
rs781753593 Chr.6: 109466210-109466210 on Build GRCh38
rs577099858 Chr.6: 109466190-109466190 on Build GRCh38
rs763528433 Chr.6: 109466176-109466187 on Build GRCh38
rs781490833 Chr.6: 109466080-109466080 on Build GRCh38
rs759348141 Chr.6: 109466113-109466113 on Build GRCh38
rs778110573 Chr.6: 109466279-109466279 on Build GRCh38

View Full Product Details