Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1544012-1544518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM184A
Entrez Gene ID
202915
Gene Name
transmembrane protein 184A
Gene Aliases
-
UniGene
Hs.592174
Gene Chromosome Location
Chr.7: 1542235-1560821 on Build GRCh38
Transcripts
NM_001097620.1 XM_017011815.1

SNP Detail

SNP ID Chromosome Location
rs986064216 Chr.7: 1544173-1544173 on Build GRCh38
rs554601400 Chr.7: 1544057-1544057 on Build GRCh38
rs111266255 Chr.7: 1544218-1544218 on Build GRCh38
rs112009579 Chr.7: 1544477-1544477 on Build GRCh38
rs1045372226 Chr.7: 1544296-1544296 on Build GRCh38
rs376509247 Chr.7: 1544245-1544245 on Build GRCh38
rs541139941 Chr.7: 1544075-1544075 on Build GRCh38
rs1023955018 Chr.7: 1544187-1544187 on Build GRCh38
rs891566346 Chr.7: 1544169-1544169 on Build GRCh38
rs966139402 Chr.7: 1544244-1544244 on Build GRCh38
rs10273193 Chr.7: 1544254-1544254 on Build GRCh38
rs73042784 Chr.7: 1544442-1544442 on Build GRCh38
rs1052907762 Chr.7: 1544282-1544282 on Build GRCh38
rs1014167507 Chr.7: 1544042-1544042 on Build GRCh38
rs567327420 Chr.7: 1544399-1544399 on Build GRCh38
rs891535325 Chr.7: 1544287-1544287 on Build GRCh38
rs949839404 Chr.7: 1544291-1544291 on Build GRCh38
rs953469695 Chr.7: 1544389-1544389 on Build GRCh38
rs1029081168 Chr.7: 1544388-1544388 on Build GRCh38
rs1015128326 Chr.7: 1544342-1544342 on Build GRCh38
rs369149971 Chr.7: 1544221-1544221 on Build GRCh38
rs1033192751 Chr.7: 1544423-1544423 on Build GRCh38
rs897179177 Chr.7: 1544119-1544119 on Build GRCh38
rs897705123 Chr.7: 1544286-1544286 on Build GRCh38
rs1000036040 Chr.7: 1544148-1544148 on Build GRCh38
rs1039767321 Chr.7: 1544486-1544486 on Build GRCh38
rs925905451 Chr.7: 1544259-1544259 on Build GRCh38
rs905628264 Chr.7: 1544456-1544456 on Build GRCh38
rs888052265 Chr.7: 1544326-1544326 on Build GRCh38
rs1018288424 Chr.7: 1544322-1544322 on Build GRCh38
rs907964759 Chr.7: 1544035-1544035 on Build GRCh38
rs575552678 Chr.7: 1544463-1544463 on Build GRCh38
rs966108769 Chr.7: 1544351-1544351 on Build GRCh38
rs759554515 Chr.7: 1544047-1544047 on Build GRCh38
rs965538000 Chr.7: 1544336-1544336 on Build GRCh38
rs952275397 Chr.7: 1544220-1544220 on Build GRCh38
rs1049572350 Chr.7: 1544115-1544115 on Build GRCh38
rs936981206 Chr.7: 1544474-1544474 on Build GRCh38
rs988598019 Chr.7: 1544441-1544441 on Build GRCh38
rs1028291400 Chr.7: 1544311-1544311 on Build GRCh38
rs979475723 Chr.7: 1544395-1544395 on Build GRCh38
rs10273306 Chr.7: 1544337-1544337 on Build GRCh38
rs376212994 Chr.7: 1544455-1544455 on Build GRCh38
rs572872128 Chr.7: 1544068-1544068 on Build GRCh38
rs949888511 Chr.7: 1544443-1544443 on Build GRCh38
rs904393548 Chr.7: 1544139-1544139 on Build GRCh38
rs901233172 Chr.7: 1544489-1544489 on Build GRCh38
rs76082160 Chr.7: 1544152-1544152 on Build GRCh38
rs529949075 Chr.7: 1544420-1544420 on Build GRCh38
rs1018393529 Chr.7: 1544204-1544204 on Build GRCh38
rs1002235205 Chr.7: 1544199-1544199 on Build GRCh38
rs118081424 Chr.7: 1544377-1544377 on Build GRCh38
rs1014990830 Chr.7: 1544222-1544222 on Build GRCh38
rs1006965040 Chr.7: 1544316-1544316 on Build GRCh38
rs994154660 Chr.7: 1544134-1544134 on Build GRCh38
rs1048745928 Chr.7: 1544476-1544476 on Build GRCh38
rs543736176 Chr.7: 1544206-1544206 on Build GRCh38
rs4268033 Chr.7: 1544037-1544037 on Build GRCh38
rs1013857639 Chr.7: 1544170-1544170 on Build GRCh38
rs561288028 Chr.7: 1544081-1544081 on Build GRCh38
rs888470212 Chr.7: 1544464-1544464 on Build GRCh38
rs922525944 Chr.7: 1544080-1544080 on Build GRCh38
rs112188946 Chr.7: 1544306-1544306 on Build GRCh38
rs563685055 Chr.7: 1544246-1544246 on Build GRCh38
rs935813473 Chr.7: 1544278-1544278 on Build GRCh38
rs532361934 Chr.7: 1544253-1544253 on Build GRCh38
rs1005172897 Chr.7: 1544497-1544497 on Build GRCh38
rs935950252 Chr.7: 1544426-1544426 on Build GRCh38
rs11760769 Chr.7: 1544305-1544305 on Build GRCh38
rs949866077 Chr.7: 1544439-1544439 on Build GRCh38
rs1007281154 Chr.7: 1544484-1544484 on Build GRCh38
rs1036298093 Chr.7: 1544283-1544283 on Build GRCh38
rs970329924 Chr.7: 1544190-1544190 on Build GRCh38
rs888564950 Chr.7: 1544310-1544310 on Build GRCh38

View Full Product Details