Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143353321-143353826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131B
Entrez Gene ID
9715
Gene Name
family with sequence similarity 131 member B
Gene Aliases
-
UniGene
Hs.648908
Gene Chromosome Location
Chr.7: 143353400-143382304 on Build GRCh38
Transcripts
NM_001031690.2 NM_001278297.1 NM_014690.4 XM_005250073.3 XM_006716186.2

SNP Detail

SNP ID Chromosome Location
rs541451424 Chr.7: 143353433-143353433 on Build GRCh38
rs1046434415 Chr.7: 143353349-143353349 on Build GRCh38
rs199498222 Chr.7: 143353473-143353473 on Build GRCh38
rs994792009 Chr.7: 143353718-143353718 on Build GRCh38
rs151006165 Chr.7: 143353604-143353604 on Build GRCh38
rs986807749 Chr.7: 143353489-143353489 on Build GRCh38
rs1055531678 Chr.7: 143353602-143353602 on Build GRCh38
rs1017212821 Chr.7: 143353385-143353385 on Build GRCh38
rs140959798 Chr.7: 143353719-143353719 on Build GRCh38
rs563345273 Chr.7: 143353725-143353725 on Build GRCh38
rs907342653 Chr.7: 143353398-143353398 on Build GRCh38
rs375040372 Chr.7: 143353566-143353566 on Build GRCh38
rs894110304 Chr.7: 143353360-143353360 on Build GRCh38
rs755892110 Chr.7: 143353600-143353600 on Build GRCh38
rs867641342 Chr.7: 143353637-143353637 on Build GRCh38
rs1045961153 Chr.7: 143353781-143353781 on Build GRCh38
rs945953085 Chr.7: 143353558-143353558 on Build GRCh38
rs983338269 Chr.7: 143353531-143353531 on Build GRCh38
rs578250159 Chr.7: 143353701-143353701 on Build GRCh38
rs1025758991 Chr.7: 143353459-143353459 on Build GRCh38
rs537598006 Chr.7: 143353453-143353453 on Build GRCh38
rs557204983 Chr.7: 143353707-143353707 on Build GRCh38
rs945731460 Chr.7: 143353736-143353736 on Build GRCh38
rs964049463 Chr.7: 143353509-143353509 on Build GRCh38
rs374820383 Chr.7: 143353495-143353495 on Build GRCh38
rs557739206 Chr.7: 143353368-143353368 on Build GRCh38
rs928420568 Chr.7: 143353592-143353592 on Build GRCh38
rs951388673 Chr.7: 143353512-143353512 on Build GRCh38
rs371266691 Chr.7: 143353472-143353472 on Build GRCh38
rs1022038502 Chr.7: 143353785-143353785 on Build GRCh38
rs1012934286 Chr.7: 143353378-143353378 on Build GRCh38
rs1018342724 Chr.7: 143353759-143353759 on Build GRCh38
rs751034808 Chr.7: 143353478-143353478 on Build GRCh38
rs1011618053 Chr.7: 143353784-143353784 on Build GRCh38
rs780793914 Chr.7: 143353556-143353556 on Build GRCh38
rs964458047 Chr.7: 143353759-143353759 on Build GRCh38
rs569941358 Chr.7: 143353425-143353425 on Build GRCh38
rs911816458 Chr.7: 143353537-143353537 on Build GRCh38
rs775599689 Chr.7: 143353620-143353620 on Build GRCh38
rs1008278908 Chr.7: 143353754-143353754 on Build GRCh38
rs555556326 Chr.7: 143353487-143353487 on Build GRCh38
rs965866330 Chr.7: 143353581-143353581 on Build GRCh38
rs898722401 Chr.7: 143353421-143353421 on Build GRCh38
rs143617823 Chr.7: 143353594-143353594 on Build GRCh38
rs1037769877 Chr.7: 143353671-143353671 on Build GRCh38
rs754622371 Chr.7: 143353494-143353494 on Build GRCh38

View Full Product Details