Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22690938-22691439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR3
Entrez Gene ID
1960
Gene Name
early growth response 3
Gene Aliases
EGR-3, PILOT
UniGene
Hs.534313
Gene Chromosome Location
Chr.8: 22687661-22693302 on Build GRCh38
Transcripts
NM_001199880.1 NM_001199881.1 NM_004430.2 XM_005273425.3 XM_005273426.3 XM_011544429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652358 c.648C>T p.D216D Chr.8: 22690989-22690989 on Build GRCh38
4680430 c.438delC p.Y147fs*44 Chr.8: 22691199-22691199 on Build GRCh38
1098151 c.542C>T p.P181L Chr.8: 22691095-22691095 on Build GRCh38
1098150 c.579C>T p.D193D Chr.8: 22691058-22691058 on Build GRCh38
3432276 c.306C>T p.I102I Chr.8: 22691331-22691331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748093043 Chr.8: 22691006-22691006 on Build GRCh38
rs773024571 Chr.8: 22691143-22691143 on Build GRCh38
rs979115535 Chr.8: 22691292-22691292 on Build GRCh38
rs966229526 Chr.8: 22691201-22691201 on Build GRCh38
rs752241262 Chr.8: 22691279-22691279 on Build GRCh38
rs748027200 Chr.8: 22691125-22691125 on Build GRCh38
rs577590553 Chr.8: 22691385-22691385 on Build GRCh38
rs35051431 Chr.8: 22691403-22691403 on Build GRCh38
rs754227276 Chr.8: 22691259-22691259 on Build GRCh38
rs775736661 Chr.8: 22691247-22691247 on Build GRCh38
rs887439024 Chr.8: 22691052-22691052 on Build GRCh38
rs571918724 Chr.8: 22691066-22691066 on Build GRCh38
rs759841065 Chr.8: 22691239-22691239 on Build GRCh38
rs998071058 Chr.8: 22691213-22691213 on Build GRCh38
rs762146677 Chr.8: 22691265-22691265 on Build GRCh38
rs201203457 Chr.8: 22691072-22691072 on Build GRCh38
rs902024202 Chr.8: 22691183-22691183 on Build GRCh38
rs766546348 Chr.8: 22691023-22691023 on Build GRCh38
rs779641186 Chr.8: 22691084-22691084 on Build GRCh38
rs967310066 Chr.8: 22691358-22691358 on Build GRCh38
rs978041994 Chr.8: 22691388-22691388 on Build GRCh38
rs370729228 Chr.8: 22691241-22691241 on Build GRCh38
rs756070397 Chr.8: 22691297-22691297 on Build GRCh38
rs573987115 Chr.8: 22691039-22691039 on Build GRCh38
rs199865032 Chr.8: 22691013-22691013 on Build GRCh38
rs749261726 Chr.8: 22691313-22691313 on Build GRCh38
rs769194373 Chr.8: 22691401-22691401 on Build GRCh38
rs774703272 Chr.8: 22691225-22691225 on Build GRCh38
rs764549547 Chr.8: 22691252-22691252 on Build GRCh38
rs771170208 Chr.8: 22691168-22691168 on Build GRCh38
rs139824831 Chr.8: 22691030-22691030 on Build GRCh38
rs758037552 Chr.8: 22691073-22691073 on Build GRCh38
rs1015558176 Chr.8: 22691123-22691123 on Build GRCh38
rs953490227 Chr.8: 22691238-22691238 on Build GRCh38
rs373775159 Chr.8: 22691172-22691172 on Build GRCh38
rs371366100 Chr.8: 22691404-22691404 on Build GRCh38
rs763210546 Chr.8: 22691017-22691017 on Build GRCh38
rs748482371 Chr.8: 22691116-22691117 on Build GRCh38
rs746512708 Chr.8: 22691094-22691094 on Build GRCh38
rs140392305 Chr.8: 22691391-22691391 on Build GRCh38
rs749100210 Chr.8: 22691166-22691166 on Build GRCh38
rs750955614 Chr.8: 22691204-22691204 on Build GRCh38
rs761224501 Chr.8: 22691250-22691250 on Build GRCh38
rs754205481 Chr.8: 22691069-22691069 on Build GRCh38
rs754431253 Chr.8: 22691096-22691096 on Build GRCh38
rs765604099 Chr.8: 22691271-22691271 on Build GRCh38
rs778880883 Chr.8: 22691361-22691361 on Build GRCh38
rs760733870 Chr.8: 22691393-22691393 on Build GRCh38
rs369402604 Chr.8: 22691145-22691145 on Build GRCh38
rs774657186 Chr.8: 22691228-22691228 on Build GRCh38
rs753186062 Chr.8: 22691049-22691049 on Build GRCh38
rs776611838 Chr.8: 22690986-22690986 on Build GRCh38
rs1014766122 Chr.8: 22691103-22691103 on Build GRCh38
rs780687434 Chr.8: 22691112-22691112 on Build GRCh38
rs767717937 Chr.8: 22691040-22691040 on Build GRCh38
rs770564582 Chr.8: 22691343-22691343 on Build GRCh38
rs769665881 Chr.8: 22691135-22691135 on Build GRCh38
rs3750192 Chr.8: 22691277-22691277 on Build GRCh38
rs375652664 Chr.8: 22691242-22691242 on Build GRCh38
rs376536749 Chr.8: 22691053-22691053 on Build GRCh38

View Full Product Details