Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83660264-83660771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBQLN1
Entrez Gene ID
29979
Gene Name
ubiquilin 1
Gene Aliases
DA41, DSK2, PLIC-1, UBQN, XDRP1
UniGene
Hs.9589
Gene Chromosome Location
Chr.9: 83659963-83708253 on Build GRCh38
Transcripts
NM_013438.4 NM_053067.2

SNP Detail

SNP ID Chromosome Location
rs773146502 Chr.9: 83660629-83660629 on Build GRCh38
rs577030778 Chr.9: 83660360-83660360 on Build GRCh38
rs879273066 Chr.9: 83660436-83660437 on Build GRCh38
rs1000822613 Chr.9: 83660321-83660321 on Build GRCh38
rs1035681019 Chr.9: 83660664-83660664 on Build GRCh38
rs111470483 Chr.9: 83660392-83660392 on Build GRCh38
rs944272762 Chr.9: 83660514-83660514 on Build GRCh38
rs931758100 Chr.9: 83660637-83660637 on Build GRCh38
rs533749865 Chr.9: 83660329-83660329 on Build GRCh38
rs924287417 Chr.9: 83660540-83660540 on Build GRCh38
rs932949411 Chr.9: 83660541-83660541 on Build GRCh38
rs1013495102 Chr.9: 83660731-83660731 on Build GRCh38
rs959142244 Chr.9: 83660454-83660454 on Build GRCh38
rs943098324 Chr.9: 83660584-83660584 on Build GRCh38
rs908802453 Chr.9: 83660503-83660503 on Build GRCh38
rs143588139 Chr.9: 83660319-83660319 on Build GRCh38
rs929271114 Chr.9: 83660632-83660632 on Build GRCh38
rs905025039 Chr.9: 83660747-83660747 on Build GRCh38
rs73476336 Chr.9: 83660393-83660393 on Build GRCh38
rs1024784378 Chr.9: 83660375-83660375 on Build GRCh38
rs1025687489 Chr.9: 83660438-83660438 on Build GRCh38
rs879580672 Chr.9: 83660445-83660446 on Build GRCh38
rs147220367 Chr.9: 83660549-83660549 on Build GRCh38
rs1046285231 Chr.9: 83660732-83660732 on Build GRCh38
rs936334802 Chr.9: 83660706-83660706 on Build GRCh38
rs1010346197 Chr.9: 83660665-83660665 on Build GRCh38
rs542996371 Chr.9: 83660486-83660486 on Build GRCh38
rs977635781 Chr.9: 83660539-83660539 on Build GRCh38
rs560669649 Chr.9: 83660409-83660409 on Build GRCh38
rs540246883 Chr.9: 83660670-83660670 on Build GRCh38
rs888951287 Chr.9: 83660386-83660386 on Build GRCh38
rs1050589985 Chr.9: 83660643-83660643 on Build GRCh38
rs879688592 Chr.9: 83660438-83660439 on Build GRCh38
rs969488506 Chr.9: 83660376-83660376 on Build GRCh38
rs34702839 Chr.9: 83660656-83660657 on Build GRCh38
rs879657889 Chr.9: 83660448-83660449 on Build GRCh38
rs894584063 Chr.9: 83660671-83660671 on Build GRCh38
rs977645459 Chr.9: 83660748-83660748 on Build GRCh38
rs879768458 Chr.9: 83660439-83660439 on Build GRCh38
rs1052083724 Chr.9: 83660722-83660722 on Build GRCh38
rs955595315 Chr.9: 83660678-83660678 on Build GRCh38
rs982769807 Chr.9: 83660485-83660485 on Build GRCh38
rs1029658040 Chr.9: 83660673-83660673 on Build GRCh38
rs1037562485 Chr.9: 83660618-83660618 on Build GRCh38
rs910273587 Chr.9: 83660566-83660566 on Build GRCh38
rs1035755894 Chr.9: 83660446-83660446 on Build GRCh38
rs899110394 Chr.9: 83660405-83660405 on Build GRCh38
rs1013417617 Chr.9: 83660672-83660672 on Build GRCh38
rs1002964000 Chr.9: 83660445-83660445 on Build GRCh38
rs533419273 Chr.9: 83660575-83660575 on Build GRCh38
rs1031341703 Chr.9: 83660323-83660323 on Build GRCh38
rs372553195 Chr.9: 83660296-83660296 on Build GRCh38
rs192520920 Chr.9: 83660347-83660347 on Build GRCh38
rs952816252 Chr.9: 83660451-83660451 on Build GRCh38
rs368215423 Chr.9: 83660657-83660657 on Build GRCh38
rs1038035305 Chr.9: 83660404-83660404 on Build GRCh38
rs879850498 Chr.9: 83660442-83660443 on Build GRCh38
rs1044214 Chr.9: 83660644-83660644 on Build GRCh38
rs1013442889 Chr.9: 83660366-83660366 on Build GRCh38
rs969037725 Chr.9: 83660508-83660508 on Build GRCh38
rs991702450 Chr.9: 83660502-83660502 on Build GRCh38
rs892400179 Chr.9: 83660724-83660724 on Build GRCh38

View Full Product Details