Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127611965-127612459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STXBP1
Entrez Gene ID
6812
Gene Name
syntaxin binding protein 1
Gene Aliases
MUNC18-1, NSEC1, P67, RBSEC1, UNC18
UniGene
Hs.288229
Gene Chromosome Location
Chr.9: 127612207-127692716 on Build GRCh38
Transcripts
NM_001032221.3 NM_003165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3375088 c.9C>G p.P3P Chr.9: 127612412-127612412 on Build GRCh38
3375089 c.9C>G p.P3P Chr.9: 127612412-127612412 on Build GRCh38
4407780 c.37+10G>T p.? Chr.9: 127612450-127612450 on Build GRCh38
4407781 c.37+10G>T p.? Chr.9: 127612450-127612450 on Build GRCh38
5572052 c.27T>C p.V9V Chr.9: 127612430-127612430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557340115 Chr.9: 127612259-127612259 on Build GRCh38
rs901998024 Chr.9: 127612006-127612006 on Build GRCh38
rs961365319 Chr.9: 127612101-127612101 on Build GRCh38
rs961049606 Chr.9: 127612225-127612225 on Build GRCh38
rs759317831 Chr.9: 127612360-127612360 on Build GRCh38
rs548211260 Chr.9: 127612109-127612109 on Build GRCh38
rs762014956 Chr.9: 127612376-127612376 on Build GRCh38
rs889378001 Chr.9: 127612042-127612042 on Build GRCh38
rs994243367 Chr.9: 127612231-127612231 on Build GRCh38
rs750653236 Chr.9: 127612383-127612383 on Build GRCh38
rs769479404 Chr.9: 127612365-127612365 on Build GRCh38
rs943599518 Chr.9: 127612160-127612160 on Build GRCh38
rs374864600 Chr.9: 127612038-127612038 on Build GRCh38
rs886063463 Chr.9: 127612247-127612247 on Build GRCh38
rs556763394 Chr.9: 127612242-127612243 on Build GRCh38
rs1040396114 Chr.9: 127612054-127612054 on Build GRCh38
rs1040672659 Chr.9: 127612163-127612163 on Build GRCh38
rs766679205 Chr.9: 127612400-127612400 on Build GRCh38
rs372842480 Chr.9: 127612433-127612433 on Build GRCh38
rs546892436 Chr.9: 127612168-127612168 on Build GRCh38
rs943043546 Chr.9: 127612183-127612198 on Build GRCh38
rs553143598 Chr.9: 127612280-127612280 on Build GRCh38
rs534702903 Chr.9: 127612092-127612092 on Build GRCh38
rs776554092 Chr.9: 127612358-127612358 on Build GRCh38
rs923536041 Chr.9: 127612287-127612287 on Build GRCh38
rs1026702229 Chr.9: 127612235-127612235 on Build GRCh38
rs1018049697 Chr.9: 127612391-127612391 on Build GRCh38
rs1032197129 Chr.9: 127612078-127612078 on Build GRCh38
rs893212187 Chr.9: 127612195-127612195 on Build GRCh38
rs952436060 Chr.9: 127612385-127612385 on Build GRCh38
rs754349925 Chr.9: 127612401-127612401 on Build GRCh38
rs567922860 Chr.9: 127612114-127612114 on Build GRCh38
rs533627946 Chr.9: 127612254-127612254 on Build GRCh38
rs868522587 Chr.9: 127612079-127612079 on Build GRCh38
rs1011373205 Chr.9: 127612205-127612205 on Build GRCh38
rs910758664 Chr.9: 127612131-127612131 on Build GRCh38
rs539677319 Chr.9: 127612273-127612273 on Build GRCh38
rs914610793 Chr.9: 127612313-127612313 on Build GRCh38
rs886063465 Chr.9: 127612313-127612313 on Build GRCh38
rs1056940334 Chr.9: 127612333-127612333 on Build GRCh38
rs886063462 Chr.9: 127612209-127612209 on Build GRCh38
rs537761958 Chr.9: 127612210-127612210 on Build GRCh38
rs1049285524 Chr.9: 127612009-127612009 on Build GRCh38
rs1002735357 Chr.9: 127612224-127612224 on Build GRCh38
rs577441836 Chr.9: 127612272-127612272 on Build GRCh38
rs934639040 Chr.9: 127612299-127612299 on Build GRCh38
rs1002681605 Chr.9: 127612333-127612333 on Build GRCh38
rs1022804365 Chr.9: 127612080-127612080 on Build GRCh38
rs910706283 Chr.9: 127612256-127612256 on Build GRCh38
rs952562705 Chr.9: 127612247-127612247 on Build GRCh38
rs930823492 Chr.9: 127612002-127612002 on Build GRCh38
rs764134954 Chr.9: 127612370-127612370 on Build GRCh38
rs762819229 Chr.9: 127612368-127612368 on Build GRCh38
rs756463699 Chr.9: 127612393-127612393 on Build GRCh38
rs780924917 Chr.9: 127612414-127612414 on Build GRCh38
rs910270526 Chr.9: 127612114-127612132 on Build GRCh38
rs889404037 Chr.9: 127612072-127612072 on Build GRCh38
rs541877421 Chr.9: 127612315-127612315 on Build GRCh38
rs532931946 Chr.9: 127612028-127612028 on Build GRCh38
rs144094037 Chr.9: 127612409-127612409 on Build GRCh38
rs985247778 Chr.9: 127612254-127612254 on Build GRCh38
rs371754728 Chr.9: 127612007-127612007 on Build GRCh38
rs1006632067 Chr.9: 127612387-127612387 on Build GRCh38
rs972482723 Chr.9: 127612107-127612108 on Build GRCh38
rs976302006 Chr.9: 127612277-127612277 on Build GRCh38
rs934670348 Chr.9: 127612179-127612179 on Build GRCh38
rs886063464 Chr.9: 127612287-127612287 on Build GRCh38
rs755479737 Chr.9: 127612403-127612403 on Build GRCh38
rs565816006 Chr.9: 127612077-127612077 on Build GRCh38
rs573050142 Chr.9: 127612307-127612307 on Build GRCh38
rs562125563 Chr.9: 127612351-127612351 on Build GRCh38
rs1035313614 Chr.9: 127612099-127612099 on Build GRCh38
rs575515332 Chr.9: 127612411-127612411 on Build GRCh38
rs1022743772 Chr.9: 127612209-127612209 on Build GRCh38
rs767780730 Chr.9: 127612382-127612382 on Build GRCh38
rs919413105 Chr.9: 127611989-127611989 on Build GRCh38
rs943311406 Chr.9: 127612043-127612043 on Build GRCh38
rs998106896 Chr.9: 127612013-127612013 on Build GRCh38
rs905678849 Chr.9: 127612330-127612330 on Build GRCh38

View Full Product Details