Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55484937-55485438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP51
Entrez Gene ID
158880
Gene Name
ubiquitin specific peptidase 51
Gene Aliases
-
UniGene
Hs.40061
Gene Chromosome Location
Chr.X: 55484524-55489875 on Build GRCh38
Transcripts
NM_201286.3 XM_017029299.1 XM_017029300.1 XM_017029301.1

SNP Detail

SNP ID Chromosome Location
rs912783584 Chr.X: 55484968-55484968 on Build GRCh38
rs1033344709 Chr.X: 55485334-55485334 on Build GRCh38
rs1000902230 Chr.X: 55485031-55485031 on Build GRCh38
rs779014562 Chr.X: 55485217-55485217 on Build GRCh38
rs899848295 Chr.X: 55485029-55485029 on Build GRCh38
rs1049086207 Chr.X: 55485002-55485002 on Build GRCh38
rs1053831522 Chr.X: 55485046-55485046 on Build GRCh38
rs763766806 Chr.X: 55485285-55485285 on Build GRCh38
rs756836624 Chr.X: 55485373-55485373 on Build GRCh38
rs944316686 Chr.X: 55485001-55485001 on Build GRCh38
rs182584769 Chr.X: 55485017-55485017 on Build GRCh38
rs1012932579 Chr.X: 55485084-55485084 on Build GRCh38
rs753291776 Chr.X: 55485320-55485320 on Build GRCh38
rs1002133142 Chr.X: 55485271-55485271 on Build GRCh38
rs1023850775 Chr.X: 55485138-55485138 on Build GRCh38
rs754072655 Chr.X: 55485051-55485051 on Build GRCh38
rs758432980 Chr.X: 55485371-55485371 on Build GRCh38
rs757442365 Chr.X: 55485052-55485052 on Build GRCh38
rs1024824533 Chr.X: 55485206-55485206 on Build GRCh38
rs750525233 Chr.X: 55485366-55485366 on Build GRCh38
rs912395357 Chr.X: 55485391-55485391 on Build GRCh38
rs968126030 Chr.X: 55485346-55485346 on Build GRCh38
rs1009838850 Chr.X: 55485054-55485054 on Build GRCh38
rs765628091 Chr.X: 55485395-55485395 on Build GRCh38
rs963076098 Chr.X: 55485335-55485335 on Build GRCh38
rs1030621776 Chr.X: 55485370-55485370 on Build GRCh38
rs942522581 Chr.X: 55485414-55485414 on Build GRCh38
rs896624521 Chr.X: 55485060-55485060 on Build GRCh38

View Full Product Details