Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230357218-230357728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGBD5
Entrez Gene ID
79605
Gene Name
piggyBac transposable element derived 5
Gene Aliases
-
UniGene
Hs.520463
Gene Chromosome Location
Chr.1: 230321646-230425928 on Build GRCh38
Transcripts
NM_001258311.1 XM_017002330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3356536 c.430C>T p.R144* Chr.1: 230357313-230357313 on Build GRCh38
3484991 c.482C>T p.P161L Chr.1: 230357261-230357261 on Build GRCh38
5789657 c.465C>T p.F155F Chr.1: 230357278-230357278 on Build GRCh38
400425 c.449C>G p.A150G Chr.1: 230357294-230357294 on Build GRCh38
6744570 c.444delC p.S149fs*44 Chr.1: 230357299-230357299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929683134 Chr.1: 230357496-230357496 on Build GRCh38
rs376716982 Chr.1: 230357312-230357312 on Build GRCh38
rs549098413 Chr.1: 230357377-230357377 on Build GRCh38
rs41494754 Chr.1: 230357699-230357699 on Build GRCh38
rs929926261 Chr.1: 230357563-230357563 on Build GRCh38
rs958954972 Chr.1: 230357420-230357420 on Build GRCh38
rs144759508 Chr.1: 230357429-230357429 on Build GRCh38
rs545476552 Chr.1: 230357605-230357605 on Build GRCh38
rs767804134 Chr.1: 230357295-230357295 on Build GRCh38
rs564237443 Chr.1: 230357296-230357296 on Build GRCh38
rs899968368 Chr.1: 230357627-230357627 on Build GRCh38
rs1047085853 Chr.1: 230357565-230357565 on Build GRCh38
rs368703002 Chr.1: 230357382-230357382 on Build GRCh38
rs998318812 Chr.1: 230357376-230357376 on Build GRCh38
rs540482771 Chr.1: 230357340-230357340 on Build GRCh38
rs556492646 Chr.1: 230357564-230357564 on Build GRCh38
rs539884072 Chr.1: 230357504-230357504 on Build GRCh38
rs773136056 Chr.1: 230357269-230357269 on Build GRCh38
rs755815295 Chr.1: 230357342-230357342 on Build GRCh38
rs891390956 Chr.1: 230357407-230357407 on Build GRCh38
rs1000167258 Chr.1: 230357644-230357644 on Build GRCh38
rs771981074 Chr.1: 230357363-230357363 on Build GRCh38
rs374594858 Chr.1: 230357357-230357357 on Build GRCh38
rs186465943 Chr.1: 230357516-230357516 on Build GRCh38
rs571244027 Chr.1: 230357517-230357517 on Build GRCh38
rs75257307 Chr.1: 230357698-230357698 on Build GRCh38
rs923875940 Chr.1: 230357334-230357334 on Build GRCh38
rs556364747 Chr.1: 230357251-230357251 on Build GRCh38
rs77417958 Chr.1: 230357439-230357439 on Build GRCh38
rs560175474 Chr.1: 230357674-230357674 on Build GRCh38
rs991672095 Chr.1: 230357277-230357277 on Build GRCh38
rs780899740 Chr.1: 230357335-230357335 on Build GRCh38
rs768383159 Chr.1: 230357255-230357255 on Build GRCh38
rs967765127 Chr.1: 230357666-230357666 on Build GRCh38
rs745978060 Chr.1: 230357362-230357362 on Build GRCh38
rs1039509838 Chr.1: 230357582-230357582 on Build GRCh38
rs898467918 Chr.1: 230357370-230357370 on Build GRCh38
rs920008787 Chr.1: 230357560-230357560 on Build GRCh38
rs111271665 Chr.1: 230357352-230357352 on Build GRCh38
rs370040930 Chr.1: 230357328-230357328 on Build GRCh38
rs577906737 Chr.1: 230357645-230357645 on Build GRCh38
rs531280291 Chr.1: 230357388-230357388 on Build GRCh38
rs374332007 Chr.1: 230357286-230357286 on Build GRCh38
rs901213341 Chr.1: 230357375-230357375 on Build GRCh38
rs774108056 Chr.1: 230357361-230357361 on Build GRCh38
rs752083876 Chr.1: 230357395-230357395 on Build GRCh38
rs982251281 Chr.1: 230357658-230357658 on Build GRCh38
rs754280201 Chr.1: 230357246-230357246 on Build GRCh38
rs967662253 Chr.1: 230357462-230357462 on Build GRCh38
rs369605433 Chr.1: 230357253-230357253 on Build GRCh38
rs1046449273 Chr.1: 230357301-230357301 on Build GRCh38
rs1022004066 Chr.1: 230357664-230357664 on Build GRCh38
rs947895015 Chr.1: 230357528-230357528 on Build GRCh38
rs977947843 Chr.1: 230357319-230357319 on Build GRCh38
rs763421720 Chr.1: 230357270-230357270 on Build GRCh38
rs536229421 Chr.1: 230357553-230357553 on Build GRCh38
rs551602055 Chr.1: 230357515-230357515 on Build GRCh38
rs373961554 Chr.1: 230357264-230357264 on Build GRCh38
rs144928166 Chr.1: 230357287-230357287 on Build GRCh38
rs918235348 Chr.1: 230357490-230357490 on Build GRCh38
rs576623690 Chr.1: 230357575-230357575 on Build GRCh38
rs747987797 Chr.1: 230357256-230357256 on Build GRCh38
rs1024888435 Chr.1: 230357653-230357653 on Build GRCh38
rs948147324 Chr.1: 230357358-230357358 on Build GRCh38
rs115949842 Chr.1: 230357607-230357607 on Build GRCh38
rs759496321 Chr.1: 230357293-230357293 on Build GRCh38
rs1039519726 Chr.1: 230357374-230357374 on Build GRCh38
rs923645480 Chr.1: 230357495-230357495 on Build GRCh38
rs1025368778 Chr.1: 230357659-230357659 on Build GRCh38
rs540563456 Chr.1: 230357665-230357665 on Build GRCh38
rs370623285 Chr.1: 230357628-230357628 on Build GRCh38
rs901039357 Chr.1: 230357590-230357590 on Build GRCh38
rs756715872 Chr.1: 230357416-230357416 on Build GRCh38
rs1018442192 Chr.1: 230357419-230357419 on Build GRCh38
rs777761759 Chr.1: 230357347-230357347 on Build GRCh38
rs746466330 Chr.1: 230357254-230357254 on Build GRCh38
rs749984070 Chr.1: 230357466-230357466 on Build GRCh38
rs77462072 Chr.1: 230357428-230357428 on Build GRCh38
rs763041867 Chr.1: 230357271-230357271 on Build GRCh38
rs533271110 Chr.1: 230357316-230357316 on Build GRCh38
rs981919941 Chr.1: 230357446-230357446 on Build GRCh38
rs764513211 Chr.1: 230357305-230357305 on Build GRCh38
rs575422476 Chr.1: 230357261-230357261 on Build GRCh38
rs373257100 Chr.1: 230357304-230357304 on Build GRCh38
rs775639591 Chr.1: 230357366-230357366 on Build GRCh38
rs971619504 Chr.1: 230357658-230357659 on Build GRCh38

View Full Product Details