Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 233375800-233376309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1804
Entrez Gene ID
84451
Gene Name
mixed lineage kinase 4
Gene Aliases
MLK4, dJ862P8.3
UniGene
Hs.547779
Gene Chromosome Location
Chr.1: 233327732-233385148 on Build GRCh38
Transcripts
NM_032435.2 XM_011544305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117926 c.1723C>T p.R575* Chr.1: 233375963-233375963 on Build GRCh38
7002206 c.1732G>T p.E578* Chr.1: 233375972-233375972 on Build GRCh38
1579039 c.1826+8T>C p.? Chr.1: 233376074-233376074 on Build GRCh38
5969245 c.1816T>C p.C606R Chr.1: 233376056-233376056 on Build GRCh38
4143523 c.1800G>T p.M600I Chr.1: 233376040-233376040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759249543 Chr.1: 233376003-233376003 on Build GRCh38
rs767641028 Chr.1: 233375876-233375876 on Build GRCh38
rs753986648 Chr.1: 233376044-233376044 on Build GRCh38
rs34984140 Chr.1: 233376030-233376030 on Build GRCh38
rs771964988 Chr.1: 233376004-233376004 on Build GRCh38
rs745862449 Chr.1: 233376110-233376110 on Build GRCh38
rs575713653 Chr.1: 233376121-233376121 on Build GRCh38
rs540738511 Chr.1: 233375931-233375931 on Build GRCh38
rs778706284 Chr.1: 233376099-233376099 on Build GRCh38
rs1047084289 Chr.1: 233376209-233376209 on Build GRCh38
rs542232476 Chr.1: 233376244-233376244 on Build GRCh38
rs765842021 Chr.1: 233376055-233376055 on Build GRCh38
rs374695182 Chr.1: 233375964-233375964 on Build GRCh38
rs35758282 Chr.1: 233375929-233375929 on Build GRCh38
rs577850250 Chr.1: 233376139-233376139 on Build GRCh38
rs759748681 Chr.1: 233375867-233375867 on Build GRCh38
rs555861371 Chr.1: 233376094-233376094 on Build GRCh38
rs200447649 Chr.1: 233375890-233375890 on Build GRCh38
rs771729620 Chr.1: 233376111-233376111 on Build GRCh38
rs764315441 Chr.1: 233375906-233375906 on Build GRCh38
rs528819698 Chr.1: 233376243-233376243 on Build GRCh38
rs1038425459 Chr.1: 233376077-233376077 on Build GRCh38
rs754592533 Chr.1: 233375927-233375927 on Build GRCh38
rs995607994 Chr.1: 233375868-233375868 on Build GRCh38
rs887228423 Chr.1: 233376213-233376213 on Build GRCh38
rs189523055 Chr.1: 233376134-233376134 on Build GRCh38
rs183925268 Chr.1: 233375839-233375839 on Build GRCh38
rs935081085 Chr.1: 233376078-233376078 on Build GRCh38
rs779369677 Chr.1: 233375916-233375916 on Build GRCh38
rs535777470 Chr.1: 233375887-233375887 on Build GRCh38
rs756830029 Chr.1: 233376098-233376098 on Build GRCh38
rs373077914 Chr.1: 233376080-233376080 on Build GRCh38
rs540174311 Chr.1: 233375970-233375970 on Build GRCh38
rs183995122 Chr.1: 233376088-233376088 on Build GRCh38
rs774131913 Chr.1: 233375998-233375998 on Build GRCh38
rs35806537 Chr.1: 233376168-233376171 on Build GRCh38
rs1045361639 Chr.1: 233376142-233376142 on Build GRCh38
rs112162717 Chr.1: 233376107-233376107 on Build GRCh38
rs988619613 Chr.1: 233376010-233376011 on Build GRCh38
rs866385858 Chr.1: 233376031-233376031 on Build GRCh38
rs370340548 Chr.1: 233375896-233375896 on Build GRCh38
rs867466724 Chr.1: 233376023-233376023 on Build GRCh38
rs758676664 Chr.1: 233376070-233376070 on Build GRCh38
rs557966895 Chr.1: 233376136-233376136 on Build GRCh38
rs777297213 Chr.1: 233375962-233375962 on Build GRCh38
rs151099146 Chr.1: 233375934-233375934 on Build GRCh38
rs760993548 Chr.1: 233376032-233376032 on Build GRCh38
rs199515472 Chr.1: 233376071-233376071 on Build GRCh38
rs182180001 Chr.1: 233376198-233376198 on Build GRCh38
rs146732260 Chr.1: 233376143-233376143 on Build GRCh38
rs913175116 Chr.1: 233376026-233376026 on Build GRCh38
rs762349872 Chr.1: 233376047-233376047 on Build GRCh38
rs71173275 Chr.1: 233376171-233376174 on Build GRCh38
rs750764987 Chr.1: 233375924-233375924 on Build GRCh38
rs774155441 Chr.1: 233375866-233375866 on Build GRCh38
rs1008275687 Chr.1: 233376248-233376248 on Build GRCh38
rs764225816 Chr.1: 233376040-233376040 on Build GRCh38
rs1028678839 Chr.1: 233376034-233376034 on Build GRCh38
rs562126234 Chr.1: 233376063-233376063 on Build GRCh38
rs1020196926 Chr.1: 233375986-233375986 on Build GRCh38
rs757868575 Chr.1: 233375907-233375907 on Build GRCh38
rs760653430 Chr.1: 233375899-233375899 on Build GRCh38

View Full Product Details