Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 46009543-46009144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA4
Entrez Gene ID
8031
Gene Name
nuclear receptor coactivator 4
Gene Aliases
ARA70, ELE1, PTC3, RFG
UniGene
Hs.643658
Gene Chromosome Location
Chr.10: 46005088-46030714 on Build GRCh38
Transcripts
NM_001145260.1 NM_001145261.1 NM_001145262.1 NM_001145263.1 NM_005437.3

View Full Product Details