Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110780720-110781224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM20
Entrez Gene ID
282996
Gene Name
RNA binding motif protein 20
Gene Aliases
-
UniGene
Hs.116630
Gene Chromosome Location
Chr.10: 110641934-110839471 on Build GRCh38
Transcripts
NM_001134363.2 XM_011539697.2 XM_017016103.1 XM_017016104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4501582 c.592C>T p.P198S Chr.10: 110781201-110781201 on Build GRCh38
5114238 c.236C>T p.P79L Chr.10: 110780845-110780845 on Build GRCh38
6560652 c.298C>T p.L100F Chr.10: 110780907-110780907 on Build GRCh38
465256 c.467C>T p.A156V Chr.10: 110781076-110781076 on Build GRCh38
6551316 c.360G>T p.L120L Chr.10: 110780969-110780969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs794729161 Chr.10: 110781141-110781141 on Build GRCh38
rs764473320 Chr.10: 110780842-110780842 on Build GRCh38
rs1010202196 Chr.10: 110780941-110780941 on Build GRCh38
rs1033979309 Chr.10: 110780945-110780945 on Build GRCh38
rs876657570 Chr.10: 110781018-110781018 on Build GRCh38
rs751872145 Chr.10: 110780846-110780846 on Build GRCh38
rs1054494691 Chr.10: 110780925-110780933 on Build GRCh38
rs898904034 Chr.10: 110780935-110780935 on Build GRCh38
rs878854253 Chr.10: 110780961-110780961 on Build GRCh38
rs866771105 Chr.10: 110780850-110780850 on Build GRCh38
rs569709951 Chr.10: 110780957-110780957 on Build GRCh38
rs748452017 Chr.10: 110781152-110781152 on Build GRCh38
rs971703468 Chr.10: 110781130-110781130 on Build GRCh38
rs932196417 Chr.10: 110780793-110780793 on Build GRCh38
rs794729160 Chr.10: 110780913-110780913 on Build GRCh38
rs186482685 Chr.10: 110780959-110780959 on Build GRCh38
rs199868951 Chr.10: 110781057-110781057 on Build GRCh38
rs375200447 Chr.10: 110781051-110781051 on Build GRCh38
rs962374472 Chr.10: 110781065-110781065 on Build GRCh38
rs397516611 Chr.10: 110780922-110780922 on Build GRCh38
rs377139558 Chr.10: 110781164-110781164 on Build GRCh38
rs543478618 Chr.10: 110781150-110781150 on Build GRCh38
rs536357058 Chr.10: 110780856-110780856 on Build GRCh38
rs751279925 Chr.10: 110781087-110781087 on Build GRCh38
rs377623370 Chr.10: 110781176-110781176 on Build GRCh38
rs752370319 Chr.10: 110780810-110780810 on Build GRCh38
rs576099332 Chr.10: 110781149-110781149 on Build GRCh38
rs182768779 Chr.10: 110780889-110780889 on Build GRCh38
rs890687647 Chr.10: 110781131-110781131 on Build GRCh38
rs183130427 Chr.10: 110781139-110781139 on Build GRCh38
rs770818336 Chr.10: 110781017-110781017 on Build GRCh38
rs794729139 Chr.10: 110781097-110781097 on Build GRCh38
rs191342808 Chr.10: 110780833-110780833 on Build GRCh38
rs532331535 Chr.10: 110780767-110780767 on Build GRCh38
rs772994077 Chr.10: 110780996-110780996 on Build GRCh38
rs921903094 Chr.10: 110781076-110781076 on Build GRCh38
rs924003900 Chr.10: 110780754-110780754 on Build GRCh38
rs767909746 Chr.10: 110781098-110781098 on Build GRCh38
rs140119703 Chr.10: 110780755-110780755 on Build GRCh38
rs397516619 Chr.10: 110781050-110781050 on Build GRCh38
rs553309602 Chr.10: 110781132-110781132 on Build GRCh38
rs115379805 Chr.10: 110781077-110781077 on Build GRCh38
rs746776894 Chr.10: 110781008-110781008 on Build GRCh38
rs727504583 Chr.10: 110780973-110780973 on Build GRCh38
rs893150326 Chr.10: 110780936-110780936 on Build GRCh38
rs762597597 Chr.10: 110781078-110781078 on Build GRCh38
rs1041725339 Chr.10: 110781172-110781172 on Build GRCh38
rs967312621 Chr.10: 110780997-110780997 on Build GRCh38
rs561085365 Chr.10: 110781142-110781142 on Build GRCh38
rs994636707 Chr.10: 110780942-110780942 on Build GRCh38
rs727504930 Chr.10: 110780972-110780972 on Build GRCh38
rs190897417 Chr.10: 110781090-110781090 on Build GRCh38
rs7908490 Chr.10: 110781126-110781126 on Build GRCh38
rs1051972948 Chr.10: 110780805-110780805 on Build GRCh38
rs776692347 Chr.10: 110781029-110781029 on Build GRCh38
rs79088940 Chr.10: 110780862-110780862 on Build GRCh38
rs587781136 Chr.10: 110780864-110780864 on Build GRCh38
rs868855645 Chr.10: 110780861-110780864 on Build GRCh38
rs1054187967 Chr.10: 110780756-110780756 on Build GRCh38
rs554738351 Chr.10: 110780743-110780743 on Build GRCh38
rs1051939668 Chr.10: 110781129-110781129 on Build GRCh38
rs997721675 Chr.10: 110780978-110780978 on Build GRCh38
rs397516620 Chr.10: 110781074-110781074 on Build GRCh38
rs1042452407 Chr.10: 110780752-110780752 on Build GRCh38
rs886046699 Chr.10: 110780914-110780914 on Build GRCh38
rs757704209 Chr.10: 110780848-110780848 on Build GRCh38
rs1024246873 Chr.10: 110780979-110780979 on Build GRCh38
rs397516622 Chr.10: 110781153-110781153 on Build GRCh38
rs977645949 Chr.10: 110781028-110781028 on Build GRCh38
rs114622030 Chr.10: 110780770-110780770 on Build GRCh38
rs746016893 Chr.10: 110780942-110780943 on Build GRCh38
rs886038415 Chr.10: 110780870-110780870 on Build GRCh38
rs727504943 Chr.10: 110781145-110781145 on Build GRCh38
rs397516621 Chr.10: 110781138-110781138 on Build GRCh38
rs373480063 Chr.10: 110780958-110780958 on Build GRCh38
rs781652701 Chr.10: 110780860-110780860 on Build GRCh38
rs565220046 Chr.10: 110780760-110780760 on Build GRCh38

View Full Product Details