Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110799400-110799919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM20
Entrez Gene ID
282996
Gene Name
RNA binding motif protein 20
Gene Aliases
-
UniGene
Hs.116630
Gene Chromosome Location
Chr.10: 110641934-110839471 on Build GRCh38
Transcripts
NM_001134363.2 XM_011539697.2 XM_017016103.1 XM_017016104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5173294 c.1669-8T>A p.? Chr.10: 110799779-110799779 on Build GRCh38
7187735 c.1682T>C p.M561T Chr.10: 110799800-110799800 on Build GRCh38
6260444 c.1699G>T p.A567S Chr.10: 110799817-110799817 on Build GRCh38
5557379 c.1763T>A p.I588N Chr.10: 110799881-110799881 on Build GRCh38
1345773 c.1770G>T p.M590I Chr.10: 110799888-110799888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192684053 Chr.10: 110799692-110799692 on Build GRCh38
rs907121925 Chr.10: 110799698-110799698 on Build GRCh38
rs755364077 Chr.10: 110799822-110799822 on Build GRCh38
rs903324770 Chr.10: 110799774-110799777 on Build GRCh38
rs968101858 Chr.10: 110799505-110799505 on Build GRCh38
rs967829624 Chr.10: 110799488-110799488 on Build GRCh38
rs890837429 Chr.10: 110799863-110799863 on Build GRCh38
rs999297099 Chr.10: 110799800-110799800 on Build GRCh38
rs1002747714 Chr.10: 110799447-110799447 on Build GRCh38
rs929075835 Chr.10: 110799573-110799573 on Build GRCh38
rs113893184 Chr.10: 110799729-110799729 on Build GRCh38
rs544635435 Chr.10: 110799672-110799672 on Build GRCh38
rs763043195 Chr.10: 110799588-110799588 on Build GRCh38
rs752100126 Chr.10: 110799723-110799723 on Build GRCh38
rs767827357 Chr.10: 110799866-110799866 on Build GRCh38
rs571381073 Chr.10: 110799481-110799481 on Build GRCh38
rs779454179 Chr.10: 110799830-110799830 on Build GRCh38
rs577569852 Chr.10: 110799654-110799654 on Build GRCh38
rs374809405 Chr.10: 110799443-110799443 on Build GRCh38
rs1009638763 Chr.10: 110799724-110799724 on Build GRCh38
rs755313638 Chr.10: 110799745-110799745 on Build GRCh38
rs976197437 Chr.10: 110799823-110799823 on Build GRCh38
rs142446622 Chr.10: 110799491-110799491 on Build GRCh38
rs559309808 Chr.10: 110799732-110799732 on Build GRCh38
rs777847060 Chr.10: 110799880-110799880 on Build GRCh38
rs919402785 Chr.10: 110799883-110799883 on Build GRCh38
rs973942393 Chr.10: 110799674-110799674 on Build GRCh38
rs530071637 Chr.10: 110799894-110799894 on Build GRCh38
rs536540514 Chr.10: 110799641-110799641 on Build GRCh38
rs929489764 Chr.10: 110799885-110799885 on Build GRCh38
rs918977242 Chr.10: 110799549-110799549 on Build GRCh38
rs986777752 Chr.10: 110799516-110799516 on Build GRCh38
rs944835746 Chr.10: 110799542-110799542 on Build GRCh38
rs576937628 Chr.10: 110799707-110799707 on Build GRCh38
rs576165971 Chr.10: 110799671-110799671 on Build GRCh38
rs915249026 Chr.10: 110799498-110799498 on Build GRCh38
rs753592290 Chr.10: 110799764-110799764 on Build GRCh38
rs794729147 Chr.10: 110799887-110799887 on Build GRCh38
rs7086886 Chr.10: 110799656-110799656 on Build GRCh38
rs184524202 Chr.10: 110799708-110799708 on Build GRCh38
rs1040526558 Chr.10: 110799544-110799544 on Build GRCh38
rs549871964 Chr.10: 110799457-110799457 on Build GRCh38
rs748640107 Chr.10: 110799852-110799852 on Build GRCh38
rs1011742607 Chr.10: 110799448-110799448 on Build GRCh38
rs1030751538 Chr.10: 110799841-110799841 on Build GRCh38
rs368716639 Chr.10: 110799884-110799884 on Build GRCh38
rs910759944 Chr.10: 110799771-110799771 on Build GRCh38
rs727504820 Chr.10: 110799879-110799879 on Build GRCh38
rs565587386 Chr.10: 110799580-110799580 on Build GRCh38
rs1022171092 Chr.10: 110799483-110799483 on Build GRCh38
rs876657973 Chr.10: 110799897-110799897 on Build GRCh38
rs557873670 Chr.10: 110799819-110799819 on Build GRCh38
rs928668102 Chr.10: 110799710-110799710 on Build GRCh38
rs1031523750 Chr.10: 110799711-110799711 on Build GRCh38
rs762010010 Chr.10: 110799479-110799479 on Build GRCh38
rs7086491 Chr.10: 110799531-110799531 on Build GRCh38
rs1035064244 Chr.10: 110799464-110799464 on Build GRCh38
rs986149118 Chr.10: 110799768-110799768 on Build GRCh38

View Full Product Details