Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 15235260-15235758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSC
Entrez Gene ID
387755
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
-
UniGene
Hs.591997
Gene Chromosome Location
Chr.11: 15111416-15269675 on Build GRCh38
Transcripts
NM_001031853.4 NM_001042536.2 NM_001278313.1 NM_001278314.1 NM_001278315.1 NM_001278316.1 XM_006718227.3 XM_011520085.1 XM_011520086.1 XM_017017697.1 XM_017017698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3445673 c.1372G>A p.E458K Chr.11: 15235662-15235662 on Build GRCh38
6028098 c.1373delA p.N459fs*34 Chr.11: 15235663-15235663 on Build GRCh38
1581050 c.1330A>G p.N444D Chr.11: 15235620-15235620 on Build GRCh38
6295368 c.1339G>T p.V447F Chr.11: 15235629-15235629 on Build GRCh38
39609 c.1333A>T p.M445L Chr.11: 15235623-15235623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909681183 Chr.11: 15235396-15235396 on Build GRCh38
rs960565715 Chr.11: 15235400-15235400 on Build GRCh38
rs751693489 Chr.11: 15235569-15235569 on Build GRCh38
rs144664939 Chr.11: 15235470-15235470 on Build GRCh38
rs749064678 Chr.11: 15235602-15235602 on Build GRCh38
rs1049584199 Chr.11: 15235492-15235492 on Build GRCh38
rs902244379 Chr.11: 15235537-15235537 on Build GRCh38
rs957835760 Chr.11: 15235307-15235307 on Build GRCh38
rs547681466 Chr.11: 15235412-15235412 on Build GRCh38
rs537533039 Chr.11: 15235478-15235478 on Build GRCh38
rs780190547 Chr.11: 15235649-15235649 on Build GRCh38
rs981742222 Chr.11: 15235340-15235340 on Build GRCh38
rs774820190 Chr.11: 15235627-15235627 on Build GRCh38
rs1028569449 Chr.11: 15235293-15235293 on Build GRCh38
rs765528584 Chr.11: 15235691-15235691 on Build GRCh38
rs550134833 Chr.11: 15235310-15235310 on Build GRCh38
rs568622651 Chr.11: 15235325-15235325 on Build GRCh38
rs981773444 Chr.11: 15235612-15235612 on Build GRCh38
rs759144849 Chr.11: 15235311-15235311 on Build GRCh38
rs1033195405 Chr.11: 15235494-15235494 on Build GRCh38
rs554111603 Chr.11: 15235564-15235564 on Build GRCh38
rs185209513 Chr.11: 15235499-15235499 on Build GRCh38
rs553824564 Chr.11: 15235480-15235480 on Build GRCh38
rs750805249 Chr.11: 15235473-15235473 on Build GRCh38
rs768170840 Chr.11: 15235667-15235667 on Build GRCh38
rs113337994 Chr.11: 15235620-15235620 on Build GRCh38
rs777217495 Chr.11: 15235616-15235616 on Build GRCh38
rs781617877 Chr.11: 15235598-15235598 on Build GRCh38
rs760253455 Chr.11: 15235646-15235646 on Build GRCh38
rs752080680 Chr.11: 15235365-15235365 on Build GRCh38
rs539456320 Chr.11: 15235462-15235462 on Build GRCh38
rs1023382081 Chr.11: 15235505-15235505 on Build GRCh38
rs757882469 Chr.11: 15235555-15235555 on Build GRCh38
rs951350510 Chr.11: 15235673-15235673 on Build GRCh38
rs556121501 Chr.11: 15235479-15235479 on Build GRCh38
rs772439385 Chr.11: 15235498-15235498 on Build GRCh38
rs755109763 Chr.11: 15235588-15235588 on Build GRCh38
rs1039774931 Chr.11: 15235409-15235409 on Build GRCh38
rs1000836271 Chr.11: 15235472-15235472 on Build GRCh38
rs1048250222 Chr.11: 15235374-15235374 on Build GRCh38
rs745731034 Chr.11: 15235623-15235623 on Build GRCh38
rs996734139 Chr.11: 15235288-15235288 on Build GRCh38
rs942772203 Chr.11: 15235397-15235397 on Build GRCh38
rs918546926 Chr.11: 15235355-15235355 on Build GRCh38
rs376351612 Chr.11: 15235603-15235603 on Build GRCh38
rs776304641 Chr.11: 15235668-15235668 on Build GRCh38
rs140600519 Chr.11: 15235421-15235421 on Build GRCh38
rs77020728 Chr.11: 15235716-15235716 on Build GRCh38
rs143252211 Chr.11: 15235691-15235692 on Build GRCh38
rs751571299 Chr.11: 15235698-15235698 on Build GRCh38
rs970444345 Chr.11: 15235521-15235521 on Build GRCh38
rs772048852 Chr.11: 15235626-15235626 on Build GRCh38
rs369300538 Chr.11: 15235700-15235700 on Build GRCh38
rs11023479 Chr.11: 15235500-15235500 on Build GRCh38
rs922714419 Chr.11: 15235723-15235723 on Build GRCh38
rs545831611 Chr.11: 15235696-15235696 on Build GRCh38
rs769100831 Chr.11: 15235543-15235543 on Build GRCh38
rs543495742 Chr.11: 15235719-15235719 on Build GRCh38
rs998225798 Chr.11: 15235413-15235413 on Build GRCh38
rs374144229 Chr.11: 15235711-15235711 on Build GRCh38
rs74771935 Chr.11: 15235601-15235601 on Build GRCh38
rs189868599 Chr.11: 15235697-15235697 on Build GRCh38
rs970328157 Chr.11: 15235332-15235332 on Build GRCh38
rs984144574 Chr.11: 15235438-15235438 on Build GRCh38
rs761185469 Chr.11: 15235539-15235539 on Build GRCh38
rs762160704 Chr.11: 15235687-15235687 on Build GRCh38
rs188802042 Chr.11: 15235373-15235373 on Build GRCh38
rs377681745 Chr.11: 15235713-15235713 on Build GRCh38
rs754022239 Chr.11: 15235580-15235580 on Build GRCh38
rs770921178 Chr.11: 15235611-15235611 on Build GRCh38
rs755168627 Chr.11: 15235593-15235593 on Build GRCh38
rs766613270 Chr.11: 15235576-15235576 on Build GRCh38
rs748171101 Chr.11: 15235656-15235656 on Build GRCh38
rs773571357 Chr.11: 15235693-15235693 on Build GRCh38
rs990501579 Chr.11: 15235319-15235319 on Build GRCh38
rs181323039 Chr.11: 15235463-15235463 on Build GRCh38

View Full Product Details