Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 16787569-16788072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHA7
Entrez Gene ID
144100
Gene Name
pleckstrin homology domain containing A7
Gene Aliases
-
UniGene
Hs.12332
Gene Chromosome Location
Chr.11: 16787153-17014423 on Build GRCh38
Transcripts
NM_175058.4 XM_017017240.1 XM_017017241.1 XM_017017242.1 XM_017017243.1

SNP Detail

SNP ID Chromosome Location
rs74463290 Chr.11: 16787641-16787641 on Build GRCh38
rs964783228 Chr.11: 16788031-16788031 on Build GRCh38
rs746289548 Chr.11: 16788030-16788030 on Build GRCh38
rs77768267 Chr.11: 16787630-16787630 on Build GRCh38
rs890281004 Chr.11: 16787621-16787621 on Build GRCh38
rs904947625 Chr.11: 16787917-16787917 on Build GRCh38
rs1026953301 Chr.11: 16788011-16788011 on Build GRCh38
rs1046257634 Chr.11: 16787907-16787907 on Build GRCh38
rs12272319 Chr.11: 16787943-16787943 on Build GRCh38
rs960181757 Chr.11: 16787760-16787760 on Build GRCh38
rs920254568 Chr.11: 16787912-16787912 on Build GRCh38
rs1019637377 Chr.11: 16787637-16787637 on Build GRCh38
rs908056079 Chr.11: 16787805-16787805 on Build GRCh38
rs1052742876 Chr.11: 16787755-16787755 on Build GRCh38
rs904513125 Chr.11: 16788041-16788041 on Build GRCh38
rs781329707 Chr.11: 16787880-16787880 on Build GRCh38
rs78456803 Chr.11: 16787931-16787931 on Build GRCh38
rs1557875 Chr.11: 16788026-16788026 on Build GRCh38
rs955018177 Chr.11: 16787650-16787650 on Build GRCh38
rs987668431 Chr.11: 16787651-16787651 on Build GRCh38
rs968750166 Chr.11: 16787690-16787690 on Build GRCh38
rs543887378 Chr.11: 16787949-16787949 on Build GRCh38
rs746243956 Chr.11: 16787901-16787901 on Build GRCh38
rs1034640068 Chr.11: 16787751-16787751 on Build GRCh38
rs577454940 Chr.11: 16787948-16787948 on Build GRCh38
rs1022007805 Chr.11: 16787668-16787668 on Build GRCh38
rs536300607 Chr.11: 16787781-16787781 on Build GRCh38
rs891857065 Chr.11: 16787639-16787639 on Build GRCh38
rs1017827145 Chr.11: 16788029-16788029 on Build GRCh38
rs550689717 Chr.11: 16787704-16787704 on Build GRCh38
rs548807382 Chr.11: 16787789-16787789 on Build GRCh38
rs779092737 Chr.11: 16787710-16787712 on Build GRCh38
rs993196390 Chr.11: 16788004-16788004 on Build GRCh38
rs36064815 Chr.11: 16787742-16787743 on Build GRCh38
rs567034064 Chr.11: 16787886-16787886 on Build GRCh38
rs752652563 Chr.11: 16787928-16787928 on Build GRCh38
rs955011098 Chr.11: 16788025-16788025 on Build GRCh38
rs116166874 Chr.11: 16787947-16787947 on Build GRCh38
rs963589703 Chr.11: 16787622-16787622 on Build GRCh38
rs556725808 Chr.11: 16787671-16787671 on Build GRCh38
rs1015922723 Chr.11: 16787591-16787591 on Build GRCh38
rs772392687 Chr.11: 16788032-16788032 on Build GRCh38
rs771122832 Chr.11: 16787988-16787988 on Build GRCh38
rs749488600 Chr.11: 16787966-16787966 on Build GRCh38
rs76600878 Chr.11: 16787750-16787750 on Build GRCh38
rs940388344 Chr.11: 16787844-16787844 on Build GRCh38

View Full Product Details