Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220103388-220103967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
UniGene
Hs.262823
Gene Chromosome Location
Chr.1: 220094113-220148041 on Build GRCh38
Transcripts
NM_018060.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
904105 c.962T>A p.V321E Chr.1: 220103458-220103458 on Build GRCh38
4576915 c.951G>A p.K317K Chr.1: 220103447-220103447 on Build GRCh38
5498768 c.999G>A p.A333A Chr.1: 220103495-220103495 on Build GRCh38
6865052 c.1046A>G p.E349G Chr.1: 220103542-220103542 on Build GRCh38
5379686 c.989A>G p.Y330C Chr.1: 220103485-220103485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147781758 Chr.1: 220103527-220103527 on Build GRCh38
rs781507223 Chr.1: 220103554-220103554 on Build GRCh38
rs769813241 Chr.1: 220103565-220103565 on Build GRCh38
rs748557971 Chr.1: 220103430-220103430 on Build GRCh38
rs12039399 Chr.1: 220103807-220103807 on Build GRCh38
rs190058588 Chr.1: 220103859-220103859 on Build GRCh38
rs774147135 Chr.1: 220103580-220103580 on Build GRCh38
rs58256586 Chr.1: 220103753-220103753 on Build GRCh38
rs771012171 Chr.1: 220103632-220103632 on Build GRCh38
rs976032472 Chr.1: 220103714-220103714 on Build GRCh38
rs767301893 Chr.1: 220103499-220103499 on Build GRCh38
rs983660383 Chr.1: 220103776-220103776 on Build GRCh38
rs922413903 Chr.1: 220103927-220103927 on Build GRCh38
rs941784889 Chr.1: 220103796-220103796 on Build GRCh38
rs146236436 Chr.1: 220103447-220103447 on Build GRCh38
rs141215374 Chr.1: 220103553-220103553 on Build GRCh38
rs755804476 Chr.1: 220103418-220103418 on Build GRCh38
rs200839272 Chr.1: 220103622-220103622 on Build GRCh38
rs754688245 Chr.1: 220103612-220103612 on Build GRCh38
rs749577151 Chr.1: 220103577-220103577 on Build GRCh38
rs747580628 Chr.1: 220103454-220103454 on Build GRCh38
rs767522177 Chr.1: 220103593-220103593 on Build GRCh38
rs959158789 Chr.1: 220103469-220103469 on Build GRCh38
rs994359483 Chr.1: 220103668-220103668 on Build GRCh38
rs760643822 Chr.1: 220103603-220103603 on Build GRCh38
rs753397811 Chr.1: 220103609-220103609 on Build GRCh38
rs148461080 Chr.1: 220103455-220103455 on Build GRCh38
rs577342087 Chr.1: 220103495-220103495 on Build GRCh38
rs778845065 Chr.1: 220103416-220103417 on Build GRCh38
rs977913379 Chr.1: 220103896-220103896 on Build GRCh38
rs1038893904 Chr.1: 220103601-220103601 on Build GRCh38
rs377623873 Chr.1: 220103486-220103486 on Build GRCh38
rs745882395 Chr.1: 220103479-220103479 on Build GRCh38
rs371011410 Chr.1: 220103523-220103523 on Build GRCh38
rs371632277 Chr.1: 220103598-220103598 on Build GRCh38
rs778351851 Chr.1: 220103537-220103537 on Build GRCh38
rs761468302 Chr.1: 220103587-220103587 on Build GRCh38
rs112037958 Chr.1: 220103618-220103618 on Build GRCh38
rs770082667 Chr.1: 220103482-220103482 on Build GRCh38
rs1033353161 Chr.1: 220103792-220103792 on Build GRCh38
rs139437119 Chr.1: 220103512-220103512 on Build GRCh38
rs1024453100 Chr.1: 220103707-220103707 on Build GRCh38
rs907426148 Chr.1: 220103885-220103885 on Build GRCh38
rs776783023 Chr.1: 220103466-220103466 on Build GRCh38
rs554814778 Chr.1: 220103819-220103819 on Build GRCh38
rs945246819 Chr.1: 220103494-220103494 on Build GRCh38
rs749970922 Chr.1: 220103509-220103509 on Build GRCh38
rs766046969 Chr.1: 220103604-220103604 on Build GRCh38
rs139443640 Chr.1: 220103440-220103440 on Build GRCh38
rs761745187 Chr.1: 220103496-220103496 on Build GRCh38
rs182690292 Chr.1: 220103874-220103874 on Build GRCh38
rs575084493 Chr.1: 220103838-220103838 on Build GRCh38
rs375400499 Chr.1: 220103575-220103575 on Build GRCh38
rs908930453 Chr.1: 220103422-220103422 on Build GRCh38
rs959336322 Chr.1: 220103801-220103801 on Build GRCh38
rs948999743 Chr.1: 220103862-220103864 on Build GRCh38
rs950596665 Chr.1: 220103761-220103761 on Build GRCh38
rs545161596 Chr.1: 220103902-220103902 on Build GRCh38
rs555827707 Chr.1: 220103661-220103661 on Build GRCh38
rs746350943 Chr.1: 220103559-220103559 on Build GRCh38
rs1053718559 Chr.1: 220103897-220103897 on Build GRCh38
rs768799338 Chr.1: 220103578-220103578 on Build GRCh38
rs540560028 Chr.1: 220103847-220103847 on Build GRCh38
rs779524957 Chr.1: 220103429-220103429 on Build GRCh38
rs201619737 Chr.1: 220103487-220103487 on Build GRCh38
rs1002158603 Chr.1: 220103782-220103782 on Build GRCh38

View Full Product Details