Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26553251-26553764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KA1
Entrez Gene ID
6195
Gene Name
ribosomal protein S6 kinase A1
Gene Aliases
HU-1, MAPKAPK1A, RSK, RSK1, p90Rsk
UniGene
Hs.149957
Gene Chromosome Location
Chr.1: 26529758-26575029 on Build GRCh38
Transcripts
NM_001006665.1 NM_002953.3 XM_005245966.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2235331 c.543T>C p.A181A Chr.1: 26553438-26553438 on Build GRCh38
6261826 c.508G>T p.E170* Chr.1: 26553430-26553430 on Build GRCh38
2235329 c.533C>T p.A178V Chr.1: 26553428-26553428 on Build GRCh38
2235332 c.516T>C p.A172A Chr.1: 26553438-26553438 on Build GRCh38
4790568 c.506C>T p.T169M Chr.1: 26553401-26553401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182006701 Chr.1: 26553645-26553645 on Build GRCh38
rs535391909 Chr.1: 26553560-26553560 on Build GRCh38
rs142081450 Chr.1: 26553664-26553664 on Build GRCh38
rs774328513 Chr.1: 26553374-26553374 on Build GRCh38
rs888242382 Chr.1: 26553632-26553632 on Build GRCh38
rs748559933 Chr.1: 26553364-26553364 on Build GRCh38
rs965195584 Chr.1: 26553659-26553659 on Build GRCh38
rs55711203 Chr.1: 26553535-26553535 on Build GRCh38
rs749627318 Chr.1: 26553481-26553481 on Build GRCh38
rs569680564 Chr.1: 26553559-26553559 on Build GRCh38
rs4970490 Chr.1: 26553429-26553429 on Build GRCh38
rs745988842 Chr.1: 26553523-26553523 on Build GRCh38
rs780807681 Chr.1: 26553716-26553716 on Build GRCh38
rs773832192 Chr.1: 26553368-26553368 on Build GRCh38
rs976643992 Chr.1: 26553275-26553275 on Build GRCh38
rs775659702 Chr.1: 26553391-26553391 on Build GRCh38
rs376383316 Chr.1: 26553402-26553402 on Build GRCh38
rs935351356 Chr.1: 26553298-26553298 on Build GRCh38
rs555744522 Chr.1: 26553596-26553596 on Build GRCh38
rs533477035 Chr.1: 26553717-26553717 on Build GRCh38
rs775490188 Chr.1: 26553533-26553533 on Build GRCh38
rs759566064 Chr.1: 26553375-26553375 on Build GRCh38
rs754698371 Chr.1: 26553347-26553347 on Build GRCh38
rs746475218 Chr.1: 26553343-26553343 on Build GRCh38
rs577738317 Chr.1: 26553737-26553737 on Build GRCh38
rs770046413 Chr.1: 26553366-26553366 on Build GRCh38
rs779372465 Chr.1: 26553502-26553502 on Build GRCh38
rs780967761 Chr.1: 26553358-26553358 on Build GRCh38
rs369247907 Chr.1: 26553372-26553372 on Build GRCh38
rs1007052188 Chr.1: 26553635-26553635 on Build GRCh38
rs772135647 Chr.1: 26553531-26553531 on Build GRCh38
rs754502526 Chr.1: 26553430-26553430 on Build GRCh38
rs368715139 Chr.1: 26553295-26553295 on Build GRCh38
rs755737010 Chr.1: 26553463-26553463 on Build GRCh38
rs772330279 Chr.1: 26553383-26553383 on Build GRCh38
rs772903320 Chr.1: 26553538-26553538 on Build GRCh38
rs528724125 Chr.1: 26553299-26553299 on Build GRCh38
rs915666963 Chr.1: 26553302-26553302 on Build GRCh38
rs758743406 Chr.1: 26553316-26553316 on Build GRCh38
rs1003764429 Chr.1: 26553443-26553443 on Build GRCh38
rs199789905 Chr.1: 26553438-26553438 on Build GRCh38
rs747454883 Chr.1: 26553468-26553468 on Build GRCh38
rs976646521 Chr.1: 26553291-26553303 on Build GRCh38
rs1036630145 Chr.1: 26553500-26553500 on Build GRCh38
rs749776573 Chr.1: 26553371-26553371 on Build GRCh38
rs1027654331 Chr.1: 26553522-26553522 on Build GRCh38
rs867512176 Chr.1: 26553576-26553576 on Build GRCh38
rs751534519 Chr.1: 26553546-26553546 on Build GRCh38
rs766264061 Chr.1: 26553542-26553542 on Build GRCh38
rs4970489 Chr.1: 26553301-26553301 on Build GRCh38
rs765500318 Chr.1: 26553451-26553452 on Build GRCh38
rs924762849 Chr.1: 26553288-26553288 on Build GRCh38
rs752388723 Chr.1: 26553455-26553455 on Build GRCh38
rs759171184 Chr.1: 26553548-26553548 on Build GRCh38
rs372793504 Chr.1: 26553417-26553417 on Build GRCh38
rs778006992 Chr.1: 26553477-26553477 on Build GRCh38
rs750164804 Chr.1: 26553407-26553407 on Build GRCh38
rs559793778 Chr.1: 26553282-26553282 on Build GRCh38
rs771365818 Chr.1: 26553495-26553495 on Build GRCh38
rs762783778 Chr.1: 26553539-26553539 on Build GRCh38
rs976613172 Chr.1: 26553660-26553660 on Build GRCh38
rs924898700 Chr.1: 26553622-26553622 on Build GRCh38
rs762827541 Chr.1: 26553426-26553426 on Build GRCh38
rs768881868 Chr.1: 26553537-26553537 on Build GRCh38

View Full Product Details