Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9653466-9653958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL1X
Entrez Gene ID
6907
Gene Name
transducin (beta)-like 1X-linked
Gene Aliases
EBI, SMAP55, TBL1
UniGene
Hs.495656
Gene Chromosome Location
Chr.X: 9463295-9719740 on Build GRCh38
Transcripts
NM_001139466.1 NM_001139467.1 NM_001139468.1 NM_005647.3 XM_011545571.2

SNP Detail

SNP ID Chromosome Location
rs997730374 Chr.X: 9653627-9653627 on Build GRCh38
rs770995288 Chr.X: 9653674-9653674 on Build GRCh38
rs774423083 Chr.X: 9653749-9653749 on Build GRCh38
rs759699146 Chr.X: 9653825-9653825 on Build GRCh38
rs374028204 Chr.X: 9653921-9653921 on Build GRCh38
rs949257514 Chr.X: 9653751-9653751 on Build GRCh38
rs200509173 Chr.X: 9653644-9653644 on Build GRCh38
rs749246712 Chr.X: 9653682-9653682 on Build GRCh38
rs751198470 Chr.X: 9653665-9653665 on Build GRCh38
rs933629994 Chr.X: 9653774-9653774 on Build GRCh38
rs768301248 Chr.X: 9653808-9653808 on Build GRCh38
rs763857065 Chr.X: 9653650-9653650 on Build GRCh38
rs982045647 Chr.X: 9653754-9653754 on Build GRCh38
rs780687675 Chr.X: 9653696-9653696 on Build GRCh38
rs775681161 Chr.X: 9653697-9653697 on Build GRCh38
rs111477873 Chr.X: 9653850-9653850 on Build GRCh38
rs918406742 Chr.X: 9653764-9653764 on Build GRCh38
rs375170775 Chr.X: 9653712-9653712 on Build GRCh38
rs150634379 Chr.X: 9653662-9653662 on Build GRCh38
rs781171485 Chr.X: 9653795-9653796 on Build GRCh38
rs967440466 Chr.X: 9653862-9653862 on Build GRCh38
rs929689043 Chr.X: 9653498-9653498 on Build GRCh38
rs202036572 Chr.X: 9653706-9653706 on Build GRCh38
rs928520815 Chr.X: 9653639-9653639 on Build GRCh38
rs148076612 Chr.X: 9653598-9653598 on Build GRCh38
rs190845497 Chr.X: 9653855-9653855 on Build GRCh38
rs1036286397 Chr.X: 9653741-9653741 on Build GRCh38
rs187562226 Chr.X: 9653829-9653829 on Build GRCh38
rs756100518 Chr.X: 9653777-9653777 on Build GRCh38
rs147021961 Chr.X: 9653596-9653596 on Build GRCh38
rs929857059 Chr.X: 9653770-9653770 on Build GRCh38
rs775611683 Chr.X: 9653868-9653868 on Build GRCh38
rs138597617 Chr.X: 9653599-9653599 on Build GRCh38
rs749306110 Chr.X: 9653673-9653673 on Build GRCh38
rs902682347 Chr.X: 9653898-9653898 on Build GRCh38
rs889321290 Chr.X: 9653814-9653814 on Build GRCh38
rs958582420 Chr.X: 9653899-9653899 on Build GRCh38
rs770962413 Chr.X: 9653723-9653723 on Build GRCh38
rs369613305 Chr.X: 9653538-9653538 on Build GRCh38
rs749144117 Chr.X: 9653849-9653849 on Build GRCh38
rs112828269 Chr.X: 9653860-9653860 on Build GRCh38
rs779687124 Chr.X: 9653626-9653626 on Build GRCh38
rs377641433 Chr.X: 9653693-9653693 on Build GRCh38
rs1051990858 Chr.X: 9653870-9653870 on Build GRCh38
rs181623324 Chr.X: 9653861-9653861 on Build GRCh38
rs41311813 Chr.X: 9653675-9653675 on Build GRCh38
rs769518255 Chr.X: 9653514-9653514 on Build GRCh38
rs374281338 Chr.X: 9653579-9653579 on Build GRCh38
rs758249725 Chr.X: 9653604-9653604 on Build GRCh38
rs748348758 Chr.X: 9653863-9653863 on Build GRCh38

View Full Product Details