Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34836026-34836526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UHRF1BP1
Entrez Gene ID
54887
Gene Name
UHRF1 binding protein 1
Gene Aliases
C6orf107, ICBP90, dJ349A12.1
UniGene
Hs.700656
Gene Chromosome Location
Chr.6: 34792017-34877514 on Build GRCh38
Transcripts
NM_017754.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3684291 c.855C>A p.A285A Chr.6: 34836170-34836170 on Build GRCh38
5947928 c.894C>T p.G298G Chr.6: 34836209-34836209 on Build GRCh38
6843116 c.961G>C p.E321Q Chr.6: 34836276-34836276 on Build GRCh38
3761998 c.855C>T p.A285A Chr.6: 34836170-34836170 on Build GRCh38
1231710 c.994G>A p.D332N Chr.6: 34836309-34836309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77408330 Chr.6: 34836105-34836105 on Build GRCh38
rs374245786 Chr.6: 34836266-34836266 on Build GRCh38
rs989687196 Chr.6: 34836380-34836380 on Build GRCh38
rs746416591 Chr.6: 34836284-34836284 on Build GRCh38
rs375961341 Chr.6: 34836370-34836370 on Build GRCh38
rs369086918 Chr.6: 34836247-34836247 on Build GRCh38
rs79483961 Chr.6: 34836454-34836454 on Build GRCh38
rs888954946 Chr.6: 34836467-34836467 on Build GRCh38
rs999205479 Chr.6: 34836313-34836313 on Build GRCh38
rs547422034 Chr.6: 34836471-34836471 on Build GRCh38
rs1021700245 Chr.6: 34836486-34836486 on Build GRCh38
rs368398705 Chr.6: 34836165-34836165 on Build GRCh38
rs192844474 Chr.6: 34836172-34836172 on Build GRCh38
rs542121660 Chr.6: 34836302-34836302 on Build GRCh38
rs753705390 Chr.6: 34836232-34836232 on Build GRCh38
rs887443169 Chr.6: 34836162-34836162 on Build GRCh38
rs565186390 Chr.6: 34836451-34836451 on Build GRCh38
rs767342542 Chr.6: 34836219-34836219 on Build GRCh38
rs190081988 Chr.6: 34836314-34836314 on Build GRCh38
rs997328376 Chr.6: 34836209-34836209 on Build GRCh38
rs766429896 Chr.6: 34836335-34836335 on Build GRCh38
rs776523207 Chr.6: 34836464-34836464 on Build GRCh38
rs758638982 Chr.6: 34836240-34836240 on Build GRCh38
rs773546723 Chr.6: 34836332-34836332 on Build GRCh38
rs1031145325 Chr.6: 34836339-34836339 on Build GRCh38
rs778065010 Chr.6: 34836241-34836241 on Build GRCh38
rs1028829291 Chr.6: 34836211-34836211 on Build GRCh38
rs761165193 Chr.6: 34836347-34836348 on Build GRCh38
rs7761391 Chr.6: 34836427-34836427 on Build GRCh38
rs997451891 Chr.6: 34836441-34836441 on Build GRCh38
rs374504572 Chr.6: 34836116-34836116 on Build GRCh38
rs377703789 Chr.6: 34836340-34836340 on Build GRCh38
rs1014830444 Chr.6: 34836188-34836188 on Build GRCh38
rs200601252 Chr.6: 34836223-34836223 on Build GRCh38
rs750525396 Chr.6: 34836392-34836392 on Build GRCh38
rs781529733 Chr.6: 34836269-34836269 on Build GRCh38
rs1022169449 Chr.6: 34836233-34836233 on Build GRCh38
rs773668214 Chr.6: 34836199-34836199 on Build GRCh38
rs766231948 Chr.6: 34836226-34836226 on Build GRCh38
rs371698183 Chr.6: 34836293-34836293 on Build GRCh38
rs901722570 Chr.6: 34836433-34836433 on Build GRCh38
rs748689536 Chr.6: 34836226-34836227 on Build GRCh38
rs761204762 Chr.6: 34836311-34836311 on Build GRCh38
rs770194208 Chr.6: 34836159-34836159 on Build GRCh38
rs368952964 Chr.6: 34836386-34836386 on Build GRCh38
rs774109340 Chr.6: 34836308-34836308 on Build GRCh38
rs772609278 Chr.6: 34836315-34836315 on Build GRCh38
rs759472947 Chr.6: 34836358-34836358 on Build GRCh38
rs771650277 Chr.6: 34836317-34836317 on Build GRCh38
rs374006337 Chr.6: 34836387-34836387 on Build GRCh38
rs772877843 Chr.6: 34836329-34836329 on Build GRCh38
rs761734650 Chr.6: 34836468-34836468 on Build GRCh38
rs774794569 Chr.6: 34836133-34836134 on Build GRCh38
rs781291621 Chr.6: 34836389-34836389 on Build GRCh38
rs901775235 Chr.6: 34836194-34836194 on Build GRCh38
rs185252636 Chr.6: 34836268-34836268 on Build GRCh38
rs911820651 Chr.6: 34836483-34836483 on Build GRCh38
rs761451294 Chr.6: 34836208-34836208 on Build GRCh38
rs760636472 Chr.6: 34836332-34836332 on Build GRCh38
rs534755535 Chr.6: 34836049-34836049 on Build GRCh38
rs753623011 Chr.6: 34836344-34836344 on Build GRCh38
rs144638972 Chr.6: 34836218-34836219 on Build GRCh38
rs947830034 Chr.6: 34836055-34836055 on Build GRCh38
rs199775314 Chr.6: 34836304-34836304 on Build GRCh38
rs4259246 Chr.6: 34836170-34836170 on Build GRCh38

View Full Product Details