Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149147233-149147745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF398
Entrez Gene ID
57541
Gene Name
zinc finger protein 398
Gene Aliases
P51, P71, ZER6
UniGene
Hs.632032
Gene Chromosome Location
Chr.7: 149126416-149183042 on Build GRCh38
Transcripts
NM_020781.3 NM_170686.2 XM_011516441.2

SNP Detail

SNP ID Chromosome Location
rs1046272214 Chr.7: 149147476-149147476 on Build GRCh38
rs899613351 Chr.7: 149147558-149147558 on Build GRCh38
rs978325314 Chr.7: 149147657-149147657 on Build GRCh38
rs1025319055 Chr.7: 149147329-149147329 on Build GRCh38
rs867198757 Chr.7: 149147579-149147579 on Build GRCh38
rs3735309 Chr.7: 149147694-149147694 on Build GRCh38
rs936721717 Chr.7: 149147418-149147418 on Build GRCh38
rs147377556 Chr.7: 149147331-149147332 on Build GRCh38
rs993802821 Chr.7: 149147284-149147284 on Build GRCh38
rs986681832 Chr.7: 149147573-149147573 on Build GRCh38
rs925373206 Chr.7: 149147660-149147660 on Build GRCh38
rs1015328459 Chr.7: 149147471-149147471 on Build GRCh38
rs962797672 Chr.7: 149147475-149147475 on Build GRCh38
rs568581784 Chr.7: 149147436-149147436 on Build GRCh38
rs1005652568 Chr.7: 149147535-149147535 on Build GRCh38
rs989562428 Chr.7: 149147444-149147444 on Build GRCh38
rs916692127 Chr.7: 149147450-149147450 on Build GRCh38
rs975353402 Chr.7: 149147337-149147338 on Build GRCh38
rs527789170 Chr.7: 149147308-149147308 on Build GRCh38
rs978023124 Chr.7: 149147394-149147394 on Build GRCh38
rs543213474 Chr.7: 149147334-149147334 on Build GRCh38
rs1055103465 Chr.7: 149147688-149147688 on Build GRCh38
rs144658556 Chr.7: 149147278-149147278 on Build GRCh38
rs989922004 Chr.7: 149147687-149147688 on Build GRCh38
rs1013860164 Chr.7: 149147358-149147358 on Build GRCh38
rs964887138 Chr.7: 149147336-149147336 on Build GRCh38
rs895377880 Chr.7: 149147354-149147354 on Build GRCh38
rs527819534 Chr.7: 149147516-149147516 on Build GRCh38
rs147482246 Chr.7: 149147636-149147636 on Build GRCh38
rs1030530254 Chr.7: 149147340-149147340 on Build GRCh38
rs950865535 Chr.7: 149147371-149147371 on Build GRCh38
rs945221313 Chr.7: 149147604-149147604 on Build GRCh38
rs906411776 Chr.7: 149147510-149147510 on Build GRCh38
rs953929440 Chr.7: 149147542-149147542 on Build GRCh38
rs868385291 Chr.7: 149147534-149147534 on Build GRCh38
rs1000860310 Chr.7: 149147292-149147292 on Build GRCh38
rs1025115899 Chr.7: 149147370-149147370 on Build GRCh38
rs916636272 Chr.7: 149147688-149147696 on Build GRCh38
rs1030459845 Chr.7: 149147632-149147632 on Build GRCh38
rs1042473954 Chr.7: 149147264-149147264 on Build GRCh38
rs947904914 Chr.7: 149147461-149147461 on Build GRCh38
rs912518621 Chr.7: 149147583-149147583 on Build GRCh38
rs920941173 Chr.7: 149147540-149147540 on Build GRCh38
rs112201471 Chr.7: 149147697-149147697 on Build GRCh38
rs139320278 Chr.7: 149147643-149147643 on Build GRCh38
rs1046309370 Chr.7: 149147257-149147257 on Build GRCh38
rs570802802 Chr.7: 149147661-149147661 on Build GRCh38
rs895157281 Chr.7: 149147690-149147690 on Build GRCh38
rs1005503593 Chr.7: 149147385-149147385 on Build GRCh38
rs548769815 Chr.7: 149147402-149147402 on Build GRCh38

View Full Product Details