Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149773174-149773681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF467
Entrez Gene ID
168544
Gene Name
zinc finger protein 467
Gene Aliases
EZI, Zfp467
UniGene
Hs.726477
Gene Chromosome Location
Chr.7: 149764175-149776323 on Build GRCh38
Transcripts
NM_207336.1 XM_006715864.3 XM_011515855.2 XM_011515856.2 XM_011515857.2 XM_011515858.1 XM_017011799.1

SNP Detail

SNP ID Chromosome Location
rs974557640 Chr.7: 149773467-149773467 on Build GRCh38
rs1016100248 Chr.7: 149773458-149773458 on Build GRCh38
rs1008395861 Chr.7: 149773233-149773233 on Build GRCh38
rs907728714 Chr.7: 149773651-149773651 on Build GRCh38
rs1054670219 Chr.7: 149773344-149773344 on Build GRCh38
rs944331665 Chr.7: 149773304-149773304 on Build GRCh38
rs540370287 Chr.7: 149773622-149773622 on Build GRCh38
rs953174596 Chr.7: 149773574-149773574 on Build GRCh38
rs983753991 Chr.7: 149773647-149773647 on Build GRCh38
rs41555 Chr.7: 149773653-149773653 on Build GRCh38
rs377542041 Chr.7: 149773486-149773490 on Build GRCh38
rs956190650 Chr.7: 149773278-149773278 on Build GRCh38
rs986064845 Chr.7: 149773583-149773583 on Build GRCh38
rs944362602 Chr.7: 149773589-149773589 on Build GRCh38
rs900230869 Chr.7: 149773410-149773410 on Build GRCh38
rs1008895562 Chr.7: 149773528-149773528 on Build GRCh38
rs894626820 Chr.7: 149773346-149773346 on Build GRCh38
rs1047592076 Chr.7: 149773361-149773361 on Build GRCh38
rs980076628 Chr.7: 149773340-149773340 on Build GRCh38
rs1004138378 Chr.7: 149773417-149773417 on Build GRCh38
rs919271806 Chr.7: 149773523-149773523 on Build GRCh38
rs990217243 Chr.7: 149773315-149773315 on Build GRCh38
rs1032220001 Chr.7: 149773254-149773254 on Build GRCh38
rs894670295 Chr.7: 149773641-149773641 on Build GRCh38
rs1035868987 Chr.7: 149773495-149773495 on Build GRCh38
rs1027519687 Chr.7: 149773556-149773556 on Build GRCh38
rs866089904 Chr.7: 149773259-149773259 on Build GRCh38
rs911585315 Chr.7: 149773295-149773295 on Build GRCh38
rs902898653 Chr.7: 149773323-149773323 on Build GRCh38
rs941954104 Chr.7: 149773514-149773514 on Build GRCh38
rs914606742 Chr.7: 149773319-149773319 on Build GRCh38
rs1050322633 Chr.7: 149773253-149773253 on Build GRCh38
rs1041502074 Chr.7: 149773311-149773311 on Build GRCh38
rs955788438 Chr.7: 149773193-149773193 on Build GRCh38
rs368152481 Chr.7: 149773486-149773495 on Build GRCh38
rs941808296 Chr.7: 149773375-149773375 on Build GRCh38
rs1024491898 Chr.7: 149773374-149773374 on Build GRCh38
rs962869277 Chr.7: 149773462-149773462 on Build GRCh38
rs924268895 Chr.7: 149773597-149773597 on Build GRCh38
rs554978459 Chr.7: 149773324-149773324 on Build GRCh38
rs960023178 Chr.7: 149773646-149773646 on Build GRCh38
rs907229296 Chr.7: 149773407-149773407 on Build GRCh38
rs911784875 Chr.7: 149773588-149773588 on Build GRCh38
rs1037400175 Chr.7: 149773389-149773389 on Build GRCh38
rs977075426 Chr.7: 149773596-149773596 on Build GRCh38
rs972003882 Chr.7: 149773517-149773517 on Build GRCh38
rs1051825739 Chr.7: 149773451-149773451 on Build GRCh38
rs980107519 Chr.7: 149773642-149773649 on Build GRCh38
rs909130393 Chr.7: 149773510-149773510 on Build GRCh38
rs1635785 Chr.7: 149773349-149773349 on Build GRCh38
rs933377438 Chr.7: 149773529-149773529 on Build GRCh38
rs907697981 Chr.7: 149773367-149773367 on Build GRCh38
rs958551390 Chr.7: 149773500-149773500 on Build GRCh38
rs991764792 Chr.7: 149773505-149773505 on Build GRCh38
rs1031864567 Chr.7: 149773534-149773534 on Build GRCh38
rs995815528 Chr.7: 149773418-149773418 on Build GRCh38
rs1051838424 Chr.7: 149773595-149773595 on Build GRCh38
rs935799790 Chr.7: 149773599-149773599 on Build GRCh38
rs943543042 Chr.7: 149773656-149773656 on Build GRCh38
rs1036093036 Chr.7: 149773645-149773646 on Build GRCh38

View Full Product Details